Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161350024-161350283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6547867 c.1363C>A p.R455S Chr.6: 161350134-161350134 on Build GRCh38
5019623 c.1310C>T p.P437L Chr.6: 161350187-161350187 on Build GRCh38
3024329 c.1311G>A p.P437P Chr.6: 161350186-161350186 on Build GRCh38
3782024 c.1391A>T p.D464V Chr.6: 161350106-161350106 on Build GRCh38
6932924 c.1341G>A p.W447* Chr.6: 161350156-161350156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949479970 Chr.6: 161350195-161350195 on Build GRCh38
rs942502004 Chr.6: 161350135-161350135 on Build GRCh38
rs772592654 Chr.6: 161350167-161350167 on Build GRCh38
rs770983577 Chr.6: 161350186-161350186 on Build GRCh38
rs201468658 Chr.6: 161350247-161350247 on Build GRCh38
rs779721381 Chr.6: 161350132-161350132 on Build GRCh38
rs149953814 Chr.6: 161350187-161350187 on Build GRCh38
rs1016493307 Chr.6: 161350255-161350255 on Build GRCh38
rs368253350 Chr.6: 161350091-161350091 on Build GRCh38
rs544295360 Chr.6: 161350158-161350158 on Build GRCh38
rs779138830 Chr.6: 161350117-161350118 on Build GRCh38
rs182893847 Chr.6: 161350125-161350125 on Build GRCh38
rs755627153 Chr.6: 161350131-161350131 on Build GRCh38
rs397514694 Chr.6: 161350205-161350205 on Build GRCh38
rs908339150 Chr.6: 161350152-161350152 on Build GRCh38
rs748955949 Chr.6: 161350133-161350133 on Build GRCh38
rs35125035 Chr.6: 161350084-161350084 on Build GRCh38
rs751485437 Chr.6: 161350121-161350121 on Build GRCh38
rs763854303 Chr.6: 161350103-161350103 on Build GRCh38
rs768939316 Chr.6: 161350062-161350062 on Build GRCh38
rs752910255 Chr.6: 161350227-161350229 on Build GRCh38
rs755354082 Chr.6: 161350143-161350143 on Build GRCh38
rs772078706 Chr.6: 161350075-161350075 on Build GRCh38
rs372111562 Chr.6: 161350251-161350251 on Build GRCh38
rs748617874 Chr.6: 161350159-161350159 on Build GRCh38
rs748710039 Chr.6: 161350071-161350071 on Build GRCh38
rs760223151 Chr.6: 161350211-161350211 on Build GRCh38
rs761791697 Chr.6: 161350202-161350202 on Build GRCh38
rs765860776 Chr.6: 161350214-161350214 on Build GRCh38
rs778305273 Chr.6: 161350176-161350176 on Build GRCh38
rs61730194 Chr.6: 161350083-161350083 on Build GRCh38
rs892646687 Chr.6: 161350053-161350053 on Build GRCh38
rs917999795 Chr.6: 161350193-161350193 on Build GRCh38
rs774913629 Chr.6: 161350068-161350068 on Build GRCh38
rs747064211 Chr.6: 161350178-161350178 on Build GRCh38
rs965138593 Chr.6: 161350086-161350086 on Build GRCh38
rs137853056 Chr.6: 161350139-161350139 on Build GRCh38
rs757263675 Chr.6: 161350123-161350123 on Build GRCh38
rs202021008 Chr.6: 161350215-161350215 on Build GRCh38
rs749641579 Chr.6: 161350060-161350060 on Build GRCh38
rs997369852 Chr.6: 161350101-161350101 on Build GRCh38
rs767539742 Chr.6: 161350203-161350203 on Build GRCh38
rs961239925 Chr.6: 161350163-161350163 on Build GRCh38
rs759692468 Chr.6: 161350188-161350188 on Build GRCh38
rs747573472 Chr.6: 161350259-161350259 on Build GRCh38
rs774317638 Chr.6: 161350197-161350197 on Build GRCh38
rs776761587 Chr.6: 161350089-161350089 on Build GRCh38
rs779201206 Chr.6: 161350218-161350218 on Build GRCh38
rs754589415 Chr.6: 161350217-161350217 on Build GRCh38
rs376430600 Chr.6: 161350234-161350234 on Build GRCh38
rs112022904 Chr.6: 161350097-161350097 on Build GRCh38
rs191486604 Chr.6: 161350208-161350208 on Build GRCh38

View Full Product Details