Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 177026312-177026585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBL1XR1
Entrez Gene ID
79718
Gene Name
transducin (beta)-like 1 X-linked receptor 1
Gene Aliases
C21, DC42, IRA1, MRD41, TBLR1
UniGene
Hs.714201
Gene Chromosome Location
Chr.3: 177019355-177197357 on Build GRCh38
Transcripts
NM_001321193.1 NM_001321194.1 NM_001321195.1 NM_024665.5 XM_005247775.2 XM_006713746.1 XM_011513141.1 XM_011513142.2 XM_011513143.2 XM_017007185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5000343 c.1508C>T p.S503L Chr.3: 177026383-177026383 on Build GRCh38
6797788 c.1480G>A p.A494T Chr.3: 177026411-177026411 on Build GRCh38
1316247 c.1457T>G p.I486R Chr.3: 177026434-177026434 on Build GRCh38
4584119 c.1424C>T p.A475V Chr.3: 177026467-177026467 on Build GRCh38
6358668 c.1461T>G p.F487L Chr.3: 177026430-177026430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769956144 Chr.3: 177026366-177026366 on Build GRCh38
rs543430470 Chr.3: 177026507-177026508 on Build GRCh38
rs867199680 Chr.3: 177026455-177026455 on Build GRCh38
rs760486288 Chr.3: 177026459-177026459 on Build GRCh38
rs1005514166 Chr.3: 177026365-177026365 on Build GRCh38
rs899195854 Chr.3: 177026472-177026472 on Build GRCh38
rs200091446 Chr.3: 177026421-177026421 on Build GRCh38
rs748134369 Chr.3: 177026359-177026359 on Build GRCh38
rs761210577 Chr.3: 177026481-177026481 on Build GRCh38
rs944049368 Chr.3: 177026343-177026343 on Build GRCh38
rs904665206 Chr.3: 177026514-177026514 on Build GRCh38
rs774132454 Chr.3: 177026373-177026373 on Build GRCh38
rs750885591 Chr.3: 177026484-177026484 on Build GRCh38
rs575951280 Chr.3: 177026344-177026344 on Build GRCh38
rs758674368 Chr.3: 177026342-177026342 on Build GRCh38
rs767060216 Chr.3: 177026507-177026507 on Build GRCh38
rs34147595 Chr.3: 177026422-177026423 on Build GRCh38
rs753533374 Chr.3: 177026467-177026467 on Build GRCh38
rs750992529 Chr.3: 177026335-177026335 on Build GRCh38
rs775209675 Chr.3: 177026454-177026454 on Build GRCh38
rs1001692890 Chr.3: 177026533-177026533 on Build GRCh38
rs771909324 Chr.3: 177026412-177026412 on Build GRCh38
rs753561692 Chr.3: 177026485-177026486 on Build GRCh38
rs971183833 Chr.3: 177026527-177026527 on Build GRCh38
rs544818049 Chr.3: 177026463-177026463 on Build GRCh38
rs780716623 Chr.3: 177026499-177026499 on Build GRCh38
rs745583165 Chr.3: 177026449-177026449 on Build GRCh38
rs768939841 Chr.3: 177026349-177026349 on Build GRCh38
rs759585840 Chr.3: 177026401-177026401 on Build GRCh38
rs758870385 Chr.3: 177026496-177026496 on Build GRCh38
rs1043243181 Chr.3: 177026506-177026506 on Build GRCh38
rs781587077 Chr.3: 177026353-177026353 on Build GRCh38
rs1031165698 Chr.3: 177026542-177026542 on Build GRCh38
rs982302832 Chr.3: 177026536-177026536 on Build GRCh38

View Full Product Details