Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134151874-134152117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR5
Entrez Gene ID
11091
Gene Name
WD repeat domain 5
Gene Aliases
BIG-3, CFAP89, SWD3
UniGene
Hs.397638
Gene Chromosome Location
Chr.9: 134135679-134159972 on Build GRCh38
Transcripts
NM_017588.2 NM_052821.3 XM_005272163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5492599 c.600C>T p.T200T Chr.9: 134151998-134151998 on Build GRCh38
1195666 c.601G>A p.A201T Chr.9: 134151999-134151999 on Build GRCh38
7141003 c.627C>A p.L209L Chr.9: 134152025-134152025 on Build GRCh38
1232606 c.586C>T p.R196C Chr.9: 134151984-134151984 on Build GRCh38
6114897 c.605C>T p.S202L Chr.9: 134152003-134152003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376262863 Chr.9: 134151975-134151975 on Build GRCh38
rs137995131 Chr.9: 134151931-134151931 on Build GRCh38
rs937958037 Chr.9: 134152084-134152084 on Build GRCh38
rs888269996 Chr.9: 134151902-134151902 on Build GRCh38
rs761647965 Chr.9: 134152072-134152072 on Build GRCh38
rs373025975 Chr.9: 134151959-134151959 on Build GRCh38
rs149771062 Chr.9: 134152025-134152025 on Build GRCh38
rs757032518 Chr.9: 134151957-134151957 on Build GRCh38
rs542135586 Chr.9: 134151978-134151978 on Build GRCh38
rs765258720 Chr.9: 134152052-134152052 on Build GRCh38
rs748739676 Chr.9: 134151998-134151998 on Build GRCh38
rs776681152 Chr.9: 134151932-134151932 on Build GRCh38
rs771961962 Chr.9: 134151979-134151979 on Build GRCh38
rs112881246 Chr.9: 134152030-134152030 on Build GRCh38
rs775676359 Chr.9: 134152055-134152055 on Build GRCh38
rs771087103 Chr.9: 134152044-134152044 on Build GRCh38
rs370007697 Chr.9: 134152028-134152028 on Build GRCh38
rs1005284744 Chr.9: 134151904-134151904 on Build GRCh38
rs540803574 Chr.9: 134152097-134152097 on Build GRCh38
rs373477577 Chr.9: 134152029-134152029 on Build GRCh38
rs1017814315 Chr.9: 134151949-134151949 on Build GRCh38
rs961086000 Chr.9: 134151914-134151914 on Build GRCh38
rs200226561 Chr.9: 134152076-134152076 on Build GRCh38
rs35709000 Chr.9: 134151930-134151930 on Build GRCh38
rs138223746 Chr.9: 134152048-134152048 on Build GRCh38
rs764028737 Chr.9: 134151934-134151934 on Build GRCh38
rs904761967 Chr.9: 134152036-134152036 on Build GRCh38
rs776245560 Chr.9: 134152074-134152074 on Build GRCh38
rs773499859 Chr.9: 134151954-134151955 on Build GRCh38
rs746872793 Chr.9: 134152037-134152053 on Build GRCh38
rs34873912 Chr.9: 134151929-134151929 on Build GRCh38
rs748579977 Chr.9: 134152022-134152022 on Build GRCh38
rs767181417 Chr.9: 134151970-134151970 on Build GRCh38
rs74312123 Chr.9: 134151911-134151911 on Build GRCh38
rs34523253 Chr.9: 134152004-134152004 on Build GRCh38
rs751486937 Chr.9: 134151956-134151956 on Build GRCh38

View Full Product Details