Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241501361-241501634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK25
Entrez Gene ID
10494
Gene Name
serine/threonine kinase 25
Gene Aliases
SOK1, YSK1
UniGene
Hs.516807
Gene Chromosome Location
Chr.2: 241494707-241509730 on Build GRCh38
Transcripts
NM_001271977.1 NM_001271978.1 NM_001271979.1 NM_001271980.1 NM_001282305.1 NM_001282306.1 NM_001282307.1 NM_001282308.1 NM_006374.4 XM_011510493.2 XM_011510494.2 XM_011510495.2 XM_011510496.2 XM_017003168.1 XM_017003169.1 XM_017003170.1 XM_017003171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4595680 c.207C>T p.V69V Chr.2: 241501532-241501532 on Build GRCh38
6279286 c.150C>T p.I50I Chr.2: 241501589-241501589 on Build GRCh38
5452136 c.193C>T p.Q65* Chr.2: 241501546-241501546 on Build GRCh38
7293228 c.139G>A p.A47T Chr.2: 241501600-241501600 on Build GRCh38
2910692 c.118A>G p.N40D Chr.2: 241501621-241501621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770438782 Chr.2: 241501509-241501509 on Build GRCh38
rs11555262 Chr.2: 241501513-241501513 on Build GRCh38
rs767684321 Chr.2: 241501541-241501541 on Build GRCh38
rs775965864 Chr.2: 241501520-241501520 on Build GRCh38
rs147850474 Chr.2: 241501571-241501571 on Build GRCh38
rs763320430 Chr.2: 241501450-241501450 on Build GRCh38
rs79414086 Chr.2: 241501582-241501582 on Build GRCh38
rs773298909 Chr.2: 241501553-241501553 on Build GRCh38
rs139615771 Chr.2: 241501507-241501507 on Build GRCh38
rs781176422 Chr.2: 241501383-241501383 on Build GRCh38
rs766308354 Chr.2: 241501561-241501561 on Build GRCh38
rs142503478 Chr.2: 241501493-241501493 on Build GRCh38
rs774488883 Chr.2: 241501527-241501527 on Build GRCh38
rs750398764 Chr.2: 241501462-241501462 on Build GRCh38
rs762062324 Chr.2: 241501529-241501529 on Build GRCh38
rs113961789 Chr.2: 241501494-241501494 on Build GRCh38
rs551223588 Chr.2: 241501459-241501459 on Build GRCh38
rs751805690 Chr.2: 241501456-241501456 on Build GRCh38
rs756074098 Chr.2: 241501463-241501463 on Build GRCh38
rs377627531 Chr.2: 241501428-241501428 on Build GRCh38
rs34819281 Chr.2: 241501433-241501433 on Build GRCh38
rs771702280 Chr.2: 241501424-241501424 on Build GRCh38
rs1002352369 Chr.2: 241501382-241501382 on Build GRCh38
rs778569973 Chr.2: 241501421-241501421 on Build GRCh38
rs768211804 Chr.2: 241501407-241501407 on Build GRCh38
rs776020923 Chr.2: 241501435-241501435 on Build GRCh38
rs747337771 Chr.2: 241501586-241501586 on Build GRCh38
rs182512698 Chr.2: 241501515-241501515 on Build GRCh38
rs764230747 Chr.2: 241501454-241501454 on Build GRCh38
rs752397232 Chr.2: 241501583-241501583 on Build GRCh38
rs754785882 Chr.2: 241501577-241501577 on Build GRCh38
rs779914368 Chr.2: 241501471-241501471 on Build GRCh38
rs187090685 Chr.2: 241501585-241501585 on Build GRCh38
rs751599599 Chr.2: 241501522-241501525 on Build GRCh38
rs978879093 Chr.2: 241501607-241501607 on Build GRCh38
rs967828363 Chr.2: 241501574-241501574 on Build GRCh38
rs746522098 Chr.2: 241501606-241501606 on Build GRCh38
rs771755240 Chr.2: 241501501-241501501 on Build GRCh38
rs34341643 Chr.2: 241501547-241501547 on Build GRCh38
rs755949534 Chr.2: 241501387-241501387 on Build GRCh38
rs753585405 Chr.2: 241501483-241501483 on Build GRCh38
rs540735668 Chr.2: 241501399-241501399 on Build GRCh38
rs778872528 Chr.2: 241501385-241501385 on Build GRCh38
rs965178011 Chr.2: 241501595-241501595 on Build GRCh38
rs576894509 Chr.2: 241501384-241501384 on Build GRCh38
rs760686339 Chr.2: 241501559-241501559 on Build GRCh38
rs747615211 Chr.2: 241501423-241501423 on Build GRCh38
rs747821996 Chr.2: 241501500-241501500 on Build GRCh38
rs368452885 Chr.2: 241501458-241501458 on Build GRCh38
rs763297657 Chr.2: 241501519-241501519 on Build GRCh38
rs754735722 Chr.2: 241501488-241501488 on Build GRCh38
rs749067319 Chr.2: 241501403-241501403 on Build GRCh38
rs375122530 Chr.2: 241501517-241501517 on Build GRCh38
rs370702495 Chr.2: 241501434-241501434 on Build GRCh38
rs756826952 Chr.2: 241501608-241501608 on Build GRCh38

View Full Product Details