Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71046557-71046786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGER3
Entrez Gene ID
5733
Gene Name
prostaglandin E receptor 3
Gene Aliases
EP3, EP3-I, EP3-II, EP3-III, EP3-IV, EP3-VI, EP3e, PGE2-R
UniGene
Hs.445000
Gene Chromosome Location
Chr.1: 70852353-71047814 on Build GRCh38
Transcripts
NM_001126044.1 NM_198714.1 NM_198715.2 NM_198716.1 NM_198717.1 NM_198718.1 NM_198719.1 NR_028292.1 NR_028293.1 NR_028294.1 XM_011541810.2 NR_038420.1

Gene Symbol
ZRANB2-AS1
Entrez Gene ID
100132618
Gene Name
ZRANB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.720513
Gene Chromosome Location
Chr.1: 71046506-71067184 on Build GRCh38
Transcripts
NM_001126044.1 NM_198714.1 NM_198715.2 NM_198716.1 NM_198717.1 NM_198718.1 NM_198719.1 NR_028292.1 NR_028293.1 NR_028294.1 XM_011541810.2 NR_038420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
377803 c.795G>C p.T265T Chr.1: 71046783-71046783 on Build GRCh38
4718730 c.867C>T p.C289C Chr.1: 71046711-71046711 on Build GRCh38
377804 c.795G>C p.T265T Chr.1: 71046783-71046783 on Build GRCh38
3492174 c.861C>T p.I287I Chr.1: 71046717-71046717 on Build GRCh38
4598683 c.797C>A p.A266E Chr.1: 71046781-71046781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778780821 Chr.1: 71046741-71046741 on Build GRCh38
rs757460013 Chr.1: 71046721-71046721 on Build GRCh38
rs765802523 Chr.1: 71046684-71046684 on Build GRCh38
rs768522767 Chr.1: 71046664-71046664 on Build GRCh38
rs1052825869 Chr.1: 71046579-71046579 on Build GRCh38
rs751617793 Chr.1: 71046720-71046720 on Build GRCh38
rs1014011412 Chr.1: 71046609-71046609 on Build GRCh38
rs929171912 Chr.1: 71046764-71046764 on Build GRCh38
rs748106085 Chr.1: 71046743-71046743 on Build GRCh38
rs114518249 Chr.1: 71046739-71046739 on Build GRCh38
rs777193036 Chr.1: 71046758-71046758 on Build GRCh38
rs757452233 Chr.1: 71046650-71046650 on Build GRCh38
rs371015154 Chr.1: 71046632-71046632 on Build GRCh38
rs1016794825 Chr.1: 71046683-71046683 on Build GRCh38
rs772786049 Chr.1: 71046668-71046668 on Build GRCh38
rs753208119 Chr.1: 71046701-71046701 on Build GRCh38
rs758846619 Chr.1: 71046710-71046710 on Build GRCh38
rs891531286 Chr.1: 71046599-71046599 on Build GRCh38
rs760281024 Chr.1: 71046670-71046670 on Build GRCh38
rs749093746 Chr.1: 71046729-71046729 on Build GRCh38
rs1024519160 Chr.1: 71046622-71046622 on Build GRCh38
rs376509801 Chr.1: 71046645-71046645 on Build GRCh38
rs368485003 Chr.1: 71046660-71046660 on Build GRCh38
rs766027408 Chr.1: 71046673-71046673 on Build GRCh38
rs779744220 Chr.1: 71046658-71046658 on Build GRCh38
rs747725164 Chr.1: 71046665-71046665 on Build GRCh38
rs771710073 Chr.1: 71046666-71046666 on Build GRCh38
rs759934649 Chr.1: 71046676-71046676 on Build GRCh38
rs199858801 Chr.1: 71046674-71046674 on Build GRCh38
rs781452899 Chr.1: 71046723-71046723 on Build GRCh38
rs753664752 Chr.1: 71046583-71046583 on Build GRCh38
rs115682069 Chr.1: 71046747-71046747 on Build GRCh38
rs988407451 Chr.1: 71046588-71046588 on Build GRCh38
rs764182070 Chr.1: 71046717-71046717 on Build GRCh38
rs981766763 Chr.1: 71046639-71046639 on Build GRCh38

View Full Product Details