Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57538068-57538327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF549
Entrez Gene ID
256051
Gene Name
zinc finger protein 549
Gene Aliases
-
UniGene
Hs.744289
Gene Chromosome Location
Chr.19: 57527321-57540876 on Build GRCh38
Transcripts
NM_001199295.1 NM_153263.2 XM_006723128.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4601007 c.1147A>G p.K383E Chr.19: 57538190-57538190 on Build GRCh38
5856917 c.1054G>A p.E352K Chr.19: 57538097-57538097 on Build GRCh38
5813181 c.1124C>A p.A375D Chr.19: 57538167-57538167 on Build GRCh38
1397106 c.1244A>G p.H415R Chr.19: 57538287-57538287 on Build GRCh38
2972633 c.1133G>A p.R378Q Chr.19: 57538137-57538137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772725582 Chr.19: 57538119-57538119 on Build GRCh38
rs138206012 Chr.19: 57538123-57538123 on Build GRCh38
rs778366058 Chr.19: 57538141-57538141 on Build GRCh38
rs184622580 Chr.19: 57538174-57538174 on Build GRCh38
rs372405681 Chr.19: 57538154-57538154 on Build GRCh38
rs769893710 Chr.19: 57538165-57538165 on Build GRCh38
rs751692437 Chr.19: 57538145-57538145 on Build GRCh38
rs746053157 Chr.19: 57538200-57538200 on Build GRCh38
rs199866043 Chr.19: 57538204-57538204 on Build GRCh38
rs760544683 Chr.19: 57538203-57538206 on Build GRCh38
rs753421481 Chr.19: 57538130-57538130 on Build GRCh38
rs531934209 Chr.19: 57538239-57538239 on Build GRCh38
rs765848473 Chr.19: 57538288-57538289 on Build GRCh38
rs764895077 Chr.19: 57538289-57538289 on Build GRCh38
rs910656722 Chr.19: 57538250-57538250 on Build GRCh38
rs781532617 Chr.19: 57538168-57538168 on Build GRCh38
rs962007980 Chr.19: 57538125-57538125 on Build GRCh38
rs768320191 Chr.19: 57538092-57538092 on Build GRCh38
rs546163846 Chr.19: 57538131-57538131 on Build GRCh38
rs747885613 Chr.19: 57538226-57538226 on Build GRCh38
rs753066253 Chr.19: 57538136-57538136 on Build GRCh38
rs771952062 Chr.19: 57538112-57538112 on Build GRCh38
rs1040849068 Chr.19: 57538276-57538276 on Build GRCh38
rs143688393 Chr.19: 57538240-57538240 on Build GRCh38
rs773133180 Chr.19: 57538230-57538230 on Build GRCh38
rs199998659 Chr.19: 57538137-57538137 on Build GRCh38
rs761704112 Chr.19: 57538103-57538103 on Build GRCh38
rs982039729 Chr.19: 57538126-57538126 on Build GRCh38
rs753323533 Chr.19: 57538296-57538298 on Build GRCh38
rs944771352 Chr.19: 57538275-57538275 on Build GRCh38
rs754838184 Chr.19: 57538216-57538216 on Build GRCh38
rs903221029 Chr.19: 57538293-57538293 on Build GRCh38
rs924843286 Chr.19: 57538127-57538127 on Build GRCh38
rs147397394 Chr.19: 57538101-57538101 on Build GRCh38
rs759144117 Chr.19: 57538265-57538265 on Build GRCh38
rs773189898 Chr.19: 57538143-57538144 on Build GRCh38
rs866319199 Chr.19: 57538228-57538228 on Build GRCh38
rs772005171 Chr.19: 57538227-57538227 on Build GRCh38
rs778600041 Chr.19: 57538223-57538223 on Build GRCh38
rs746582352 Chr.19: 57538238-57538238 on Build GRCh38

View Full Product Details