Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206444006-206444263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM23
Entrez Gene ID
8745
Gene Name
ADAM metallopeptidase domain 23
Gene Aliases
MDC-3, MDC3
UniGene
Hs.370287
Gene Chromosome Location
Chr.2: 206443544-206621130 on Build GRCh38
Transcripts
NM_003812.3 XM_005246932.3 XM_011512086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4608860 c.148C>A p.L50I Chr.2: 206444014-206444014 on Build GRCh38
4659364 c.194delG p.A67fs*102 Chr.2: 206444060-206444060 on Build GRCh38
4659365 c.194delG p.A67fs*102 Chr.2: 206444060-206444060 on Build GRCh38
6035856 c.165G>A p.P55P Chr.2: 206444031-206444031 on Build GRCh38
5733758 c.188G>A p.R63H Chr.2: 206444054-206444054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955308824 Chr.2: 206444025-206444025 on Build GRCh38
rs552796068 Chr.2: 206444174-206444174 on Build GRCh38
rs913390334 Chr.2: 206444198-206444198 on Build GRCh38
rs979037871 Chr.2: 206444168-206444168 on Build GRCh38
rs753678278 Chr.2: 206444034-206444034 on Build GRCh38
rs751855892 Chr.2: 206444153-206444153 on Build GRCh38
rs751609517 Chr.2: 206444108-206444108 on Build GRCh38
rs989391454 Chr.2: 206444196-206444196 on Build GRCh38
rs759292607 Chr.2: 206444048-206444048 on Build GRCh38
rs889778453 Chr.2: 206444065-206444065 on Build GRCh38
rs945468223 Chr.2: 206444153-206444171 on Build GRCh38
rs752901447 Chr.2: 206444056-206444056 on Build GRCh38
rs934242419 Chr.2: 206444032-206444032 on Build GRCh38
rs1448903 Chr.2: 206444237-206444237 on Build GRCh38
rs758588135 Chr.2: 206444062-206444062 on Build GRCh38
rs1006926372 Chr.2: 206444144-206444145 on Build GRCh38
rs1018379494 Chr.2: 206444113-206444113 on Build GRCh38
rs963668611 Chr.2: 206444147-206444147 on Build GRCh38
rs893243886 Chr.2: 206444235-206444235 on Build GRCh38
rs950221060 Chr.2: 206444206-206444206 on Build GRCh38
rs905894060 Chr.2: 206444232-206444232 on Build GRCh38
rs986564340 Chr.2: 206444060-206444060 on Build GRCh38
rs564726590 Chr.2: 206444215-206444215 on Build GRCh38
rs777719884 Chr.2: 206444088-206444088 on Build GRCh38
rs765508313 Chr.2: 206444050-206444050 on Build GRCh38
rs936215041 Chr.2: 206444195-206444195 on Build GRCh38
rs926223947 Chr.2: 206444177-206444177 on Build GRCh38
rs377275475 Chr.2: 206444165-206444165 on Build GRCh38

View Full Product Details