Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16433808-16434081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFTN1
Entrez Gene ID
23180
Gene Name
raftlin, lipid raft linker 1
Gene Aliases
MIG2, PIB10, PIG9, RAFTLIN
UniGene
Hs.98910
Gene Chromosome Location
Chr.3: 16315845-16513715 on Build GRCh38
Transcripts
NM_015150.1 XM_005264985.1 XM_005264986.2 XM_006713069.1 XM_011533529.1 XM_011533530.1 XM_011533531.1 XM_017006005.1 XM_017006006.1 XM_017006007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6006904 c.268C>T p.H90Y Chr.3: 16433915-16433915 on Build GRCh38
4801952 c.292G>T p.E98* Chr.3: 16433891-16433891 on Build GRCh38
6010835 c.300C>T p.I100I Chr.3: 16433883-16433883 on Build GRCh38
6006903 c.287C>T p.P96L Chr.3: 16433896-16433896 on Build GRCh38
479656 c.247C>T p.H83Y Chr.3: 16433936-16433936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750036759 Chr.3: 16433995-16433995 on Build GRCh38
rs200870463 Chr.3: 16433943-16433943 on Build GRCh38
rs375698878 Chr.3: 16433855-16433855 on Build GRCh38
rs775324927 Chr.3: 16433965-16433965 on Build GRCh38
rs771163715 Chr.3: 16434048-16434048 on Build GRCh38
rs761254385 Chr.3: 16433980-16433980 on Build GRCh38
rs6442602 Chr.3: 16434024-16434024 on Build GRCh38
rs372475268 Chr.3: 16433838-16433838 on Build GRCh38
rs780367454 Chr.3: 16433845-16433845 on Build GRCh38
rs781738635 Chr.3: 16434010-16434010 on Build GRCh38
rs746816952 Chr.3: 16434056-16434056 on Build GRCh38
rs771725465 Chr.3: 16433955-16433955 on Build GRCh38
rs776552641 Chr.3: 16433978-16433978 on Build GRCh38
rs764749726 Chr.3: 16433984-16433984 on Build GRCh38
rs755485120 Chr.3: 16433941-16433941 on Build GRCh38
rs920562874 Chr.3: 16433833-16433833 on Build GRCh38
rs932017170 Chr.3: 16433962-16433962 on Build GRCh38
rs930548591 Chr.3: 16433837-16433837 on Build GRCh38
rs910359646 Chr.3: 16433885-16433885 on Build GRCh38
rs139816101 Chr.3: 16433919-16433919 on Build GRCh38
rs764080100 Chr.3: 16433970-16433970 on Build GRCh38
rs575203250 Chr.3: 16433887-16433887 on Build GRCh38
rs184959730 Chr.3: 16433945-16433945 on Build GRCh38
rs143467684 Chr.3: 16433864-16433864 on Build GRCh38
rs373404854 Chr.3: 16433927-16433927 on Build GRCh38
rs990443598 Chr.3: 16433919-16433919 on Build GRCh38
rs1036552632 Chr.3: 16433830-16433830 on Build GRCh38
rs144679139 Chr.3: 16433949-16433949 on Build GRCh38
rs147754203 Chr.3: 16434005-16434005 on Build GRCh38
rs758639972 Chr.3: 16433921-16433921 on Build GRCh38
rs781778331 Chr.3: 16433942-16433942 on Build GRCh38
rs776737748 Chr.3: 16433856-16433856 on Build GRCh38
rs756825841 Chr.3: 16433910-16433910 on Build GRCh38
rs768451447 Chr.3: 16434058-16434058 on Build GRCh38
rs375619866 Chr.3: 16434036-16434036 on Build GRCh38
rs377763849 Chr.3: 16434042-16434042 on Build GRCh38
rs1034582934 Chr.3: 16433966-16433966 on Build GRCh38
rs373009209 Chr.3: 16433900-16433900 on Build GRCh38
rs770000558 Chr.3: 16433944-16433944 on Build GRCh38
rs746226405 Chr.3: 16433831-16433831 on Build GRCh38
rs770124165 Chr.3: 16433883-16433883 on Build GRCh38
rs747369668 Chr.3: 16433928-16433928 on Build GRCh38
rs73816455 Chr.3: 16433907-16433907 on Build GRCh38
rs764801487 Chr.3: 16433912-16433912 on Build GRCh38
rs749546322 Chr.3: 16433950-16433950 on Build GRCh38
rs765993446 Chr.3: 16433998-16433998 on Build GRCh38
rs142669277 Chr.3: 16433898-16433898 on Build GRCh38
rs1014399691 Chr.3: 16433985-16433985 on Build GRCh38
rs774497190 Chr.3: 16434054-16434054 on Build GRCh38
rs201677703 Chr.3: 16433889-16433889 on Build GRCh38
rs370730387 Chr.3: 16433996-16433996 on Build GRCh38
rs760438408 Chr.3: 16433968-16433968 on Build GRCh38
rs755361405 Chr.3: 16434008-16434008 on Build GRCh38
rs532776757 Chr.3: 16434037-16434037 on Build GRCh38
rs763978788 Chr.3: 16433908-16433908 on Build GRCh38

View Full Product Details