Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219002970-219003231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP65
Entrez Gene ID
255101
Gene Name
cilia and flagella associated protein 65
Gene Aliases
CCDC108
UniGene
Hs.147762
Gene Chromosome Location
Chr.2: 219002846-219043142 on Build GRCh38
Transcripts
NM_194302.3 XM_011510903.1 XM_011510904.2 XM_011510908.1 XM_011510909.1 XM_011510910.1 XM_011510911.1 XM_011510912.1 XM_011510914.1 XM_011510915.1 XM_011510916.1 XM_011510921.1 XM_017003752.1 XM_017003753.1 XM_017003754.1 XM_017003755.1 NR_046086.1

Gene Symbol
LOC100129175
Entrez Gene ID
100129175
Gene Name
uncharacterized LOC100129175
Gene Aliases
-
UniGene
Hs.147762
Gene Chromosome Location
Chr.2: 219002215-219015722 on Build GRCh38
Transcripts
NM_194302.3 XM_011510903.1 XM_011510904.2 XM_011510908.1 XM_011510909.1 XM_011510910.1 XM_011510911.1 XM_011510912.1 XM_011510914.1 XM_011510915.1 XM_011510916.1 XM_011510921.1 XM_017003752.1 XM_017003753.1 XM_017003754.1 XM_017003755.1 NR_046086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359689 c.5628C>A p.S1876R Chr.2: 219003200-219003200 on Build GRCh38
4133463 c.5618T>G p.V1873G Chr.2: 219003210-219003210 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150663004 Chr.2: 219003120-219003120 on Build GRCh38
rs372088760 Chr.2: 219003065-219003065 on Build GRCh38
rs199732478 Chr.2: 219003068-219003068 on Build GRCh38
rs761169245 Chr.2: 219003012-219003012 on Build GRCh38
rs902086253 Chr.2: 219003161-219003161 on Build GRCh38
rs1005526536 Chr.2: 219003191-219003191 on Build GRCh38
rs959900230 Chr.2: 219003121-219003121 on Build GRCh38
rs925201744 Chr.2: 219003010-219003010 on Build GRCh38
rs750322732 Chr.2: 219003124-219003124 on Build GRCh38
rs901348095 Chr.2: 219003007-219003007 on Build GRCh38
rs747719089 Chr.2: 219003203-219003203 on Build GRCh38
rs755976651 Chr.2: 219003125-219003125 on Build GRCh38
rs545869238 Chr.2: 219003153-219003153 on Build GRCh38
rs551755249 Chr.2: 219003097-219003097 on Build GRCh38
rs772651010 Chr.2: 219002996-219002996 on Build GRCh38
rs1050208270 Chr.2: 219003023-219003023 on Build GRCh38
rs886805557 Chr.2: 219003155-219003155 on Build GRCh38
rs771704295 Chr.2: 219003204-219003204 on Build GRCh38
rs567327535 Chr.2: 219003189-219003189 on Build GRCh38
rs899583835 Chr.2: 219003103-219003103 on Build GRCh38
rs61739963 Chr.2: 219003147-219003147 on Build GRCh38
rs756072413 Chr.2: 219003038-219003038 on Build GRCh38
rs549456927 Chr.2: 219003209-219003209 on Build GRCh38
rs916020467 Chr.2: 219003208-219003208 on Build GRCh38
rs537992850 Chr.2: 219003194-219003194 on Build GRCh38
rs73089092 Chr.2: 219002997-219002997 on Build GRCh38
rs997008021 Chr.2: 219003021-219003021 on Build GRCh38
rs150887672 Chr.2: 219003016-219003016 on Build GRCh38
rs766624456 Chr.2: 219003059-219003059 on Build GRCh38
rs369362490 Chr.2: 219003100-219003100 on Build GRCh38
rs778582401 Chr.2: 219003181-219003181 on Build GRCh38
rs890946142 Chr.2: 219003081-219003081 on Build GRCh38
rs202093549 Chr.2: 219003030-219003030 on Build GRCh38
rs549196797 Chr.2: 219003158-219003158 on Build GRCh38
rs765335761 Chr.2: 219003085-219003085 on Build GRCh38
rs979843975 Chr.2: 219003003-219003003 on Build GRCh38
rs754612567 Chr.2: 219003175-219003175 on Build GRCh38
rs1015168010 Chr.2: 219003197-219003197 on Build GRCh38
rs778975292 Chr.2: 219003026-219003026 on Build GRCh38
rs757197711 Chr.2: 219003111-219003111 on Build GRCh38
rs752434488 Chr.2: 219003031-219003032 on Build GRCh38

View Full Product Details