Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4898876-4899131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNE
Entrez Gene ID
1145
Gene Name
cholinergic receptor nicotinic epsilon subunit
Gene Aliases
ACHRE, CMS1D, CMS1E, CMS2A, CMS4A, CMS4B, CMS4C, FCCMS, SCCMS
UniGene
Hs.654535
Gene Chromosome Location
Chr.17: 4897769-4905019 on Build GRCh38
Transcripts
NM_000080.3 XM_017024115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4067522 c.1296G>T p.E432D Chr.17: 4899031-4899031 on Build GRCh38
6016418 c.1327-4C>T p.? Chr.17: 4898895-4898895 on Build GRCh38
4642135 c.1244C>G p.A415G Chr.17: 4899083-4899083 on Build GRCh38
7301128 c.1243G>A p.A415T Chr.17: 4899084-4899084 on Build GRCh38
4130368 c.1315G>T p.A439S Chr.17: 4899012-4899012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773970037 Chr.17: 4899028-4899028 on Build GRCh38
rs774197166 Chr.17: 4898907-4898908 on Build GRCh38
rs200343520 Chr.17: 4899038-4899038 on Build GRCh38
rs1028107315 Chr.17: 4898909-4898909 on Build GRCh38
rs990579970 Chr.17: 4898917-4898917 on Build GRCh38
rs755154052 Chr.17: 4899051-4899051 on Build GRCh38
rs757212314 Chr.17: 4899101-4899101 on Build GRCh38
rs55681486 Chr.17: 4899022-4899022 on Build GRCh38
rs748694848 Chr.17: 4899060-4899061 on Build GRCh38
rs753221624 Chr.17: 4898972-4898972 on Build GRCh38
rs781774131 Chr.17: 4899058-4899059 on Build GRCh38
rs748075130 Chr.17: 4898913-4898913 on Build GRCh38
rs756615085 Chr.17: 4898976-4898976 on Build GRCh38
rs778624148 Chr.17: 4899104-4899104 on Build GRCh38
rs762607579 Chr.17: 4898923-4898923 on Build GRCh38
rs758517310 Chr.17: 4899004-4899004 on Build GRCh38
rs778714163 Chr.17: 4899062-4899062 on Build GRCh38
rs952363037 Chr.17: 4898977-4898977 on Build GRCh38
rs753739289 Chr.17: 4899099-4899099 on Build GRCh38
rs769630764 Chr.17: 4899010-4899010 on Build GRCh38
rs779261310 Chr.17: 4898994-4898994 on Build GRCh38
rs34312573 Chr.17: 4898955-4898955 on Build GRCh38
rs769590956 Chr.17: 4898921-4898921 on Build GRCh38
rs370770111 Chr.17: 4899085-4899085 on Build GRCh38
rs121909517 Chr.17: 4899036-4899036 on Build GRCh38
rs939979098 Chr.17: 4898995-4898995 on Build GRCh38
rs188564977 Chr.17: 4899112-4899112 on Build GRCh38
rs56251503 Chr.17: 4899113-4899113 on Build GRCh38
rs767498448 Chr.17: 4898932-4898946 on Build GRCh38
rs908561478 Chr.17: 4898988-4898988 on Build GRCh38
rs375221757 Chr.17: 4899006-4899006 on Build GRCh38
rs545198600 Chr.17: 4899110-4899110 on Build GRCh38
rs759034839 Chr.17: 4898953-4898953 on Build GRCh38
rs753435292 Chr.17: 4898963-4898964 on Build GRCh38
rs538405166 Chr.17: 4898918-4898918 on Build GRCh38
rs749489742 Chr.17: 4898894-4898894 on Build GRCh38
rs749273134 Chr.17: 4899058-4899058 on Build GRCh38
rs777560468 Chr.17: 4899052-4899052 on Build GRCh38
rs886053122 Chr.17: 4898995-4898995 on Build GRCh38
rs763068459 Chr.17: 4898960-4898961 on Build GRCh38
rs774059152 Chr.17: 4898950-4898950 on Build GRCh38
rs756938858 Chr.17: 4898965-4898983 on Build GRCh38
rs746139069 Chr.17: 4898998-4898998 on Build GRCh38
rs767063318 Chr.17: 4898971-4898971 on Build GRCh38
rs147753790 Chr.17: 4898961-4898980 on Build GRCh38
rs369709668 Chr.17: 4899066-4899066 on Build GRCh38
rs920756174 Chr.17: 4898974-4898974 on Build GRCh38
rs538063018 Chr.17: 4898947-4898947 on Build GRCh38
rs760140721 Chr.17: 4898961-4898976 on Build GRCh38
rs898442941 Chr.17: 4899016-4899016 on Build GRCh38
rs750634265 Chr.17: 4899043-4899043 on Build GRCh38
rs745934587 Chr.17: 4898904-4898904 on Build GRCh38
rs763199413 Chr.17: 4899083-4899083 on Build GRCh38
rs759230979 Chr.17: 4899031-4899031 on Build GRCh38
rs1049573372 Chr.17: 4899026-4899026 on Build GRCh38
rs761189555 Chr.17: 4899035-4899035 on Build GRCh38
rs780122731 Chr.17: 4899048-4899048 on Build GRCh38
rs754170428 Chr.17: 4898980-4898980 on Build GRCh38
rs926162931 Chr.17: 4898946-4898946 on Build GRCh38
rs966410323 Chr.17: 4899079-4899079 on Build GRCh38
rs757445616 Chr.17: 4898899-4898899 on Build GRCh38
rs751593666 Chr.17: 4899049-4899049 on Build GRCh38
rs994146259 Chr.17: 4899061-4899061 on Build GRCh38
rs33978919 Chr.17: 4899034-4899034 on Build GRCh38
rs775001449 Chr.17: 4899070-4899070 on Build GRCh38
rs369296997 Chr.17: 4898979-4898979 on Build GRCh38
rs149892066 Chr.17: 4898905-4898905 on Build GRCh38
rs769945146 Chr.17: 4899072-4899073 on Build GRCh38
rs552229479 Chr.17: 4898922-4898922 on Build GRCh38
rs754975887 Chr.17: 4899072-4899072 on Build GRCh38
rs772995680 Chr.17: 4899013-4899013 on Build GRCh38
rs759383923 Chr.17: 4898920-4898932 on Build GRCh38
rs201740289 Chr.17: 4898968-4898968 on Build GRCh38
rs368279189 Chr.17: 4898940-4898940 on Build GRCh38
rs778012442 Chr.17: 4899102-4899103 on Build GRCh38
rs372515997 Chr.17: 4899060-4899060 on Build GRCh38
rs752019990 Chr.17: 4898978-4898981 on Build GRCh38
rs373710822 Chr.17: 4899108-4899108 on Build GRCh38
rs763619720 Chr.17: 4898962-4898962 on Build GRCh38
rs770570480 Chr.17: 4898894-4898894 on Build GRCh38
rs758716291 Chr.17: 4899045-4899045 on Build GRCh38
rs781373404 Chr.17: 4899086-4899086 on Build GRCh38
rs757760505 Chr.17: 4898981-4898981 on Build GRCh38
rs976632458 Chr.17: 4898931-4898932 on Build GRCh38
rs201553105 Chr.17: 4899111-4899111 on Build GRCh38
rs779066933 Chr.17: 4898901-4898901 on Build GRCh38
rs202051373 Chr.17: 4899082-4899082 on Build GRCh38
rs755420360 Chr.17: 4899002-4899002 on Build GRCh38
rs748069648 Chr.17: 4899009-4899009 on Build GRCh38
rs765647575 Chr.17: 4899078-4899078 on Build GRCh38
rs770772272 Chr.17: 4899025-4899025 on Build GRCh38
rs144952259 Chr.17: 4899065-4899065 on Build GRCh38
rs768258598 Chr.17: 4899077-4899077 on Build GRCh38
rs766507655 Chr.17: 4899084-4899084 on Build GRCh38

View Full Product Details