Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27055224-27055497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBOX1
Entrez Gene ID
8424
Gene Name
gamma-butyrobetaine hydroxylase 1
Gene Aliases
BBH, BBOX, G-BBH, gamma-BBH
UniGene
Hs.591996
Gene Chromosome Location
Chr.11: 27040705-27127807 on Build GRCh38
Transcripts
NM_003986.2 XM_005253159.2 XM_005253160.2 XM_005253161.2 XM_011520402.1 XM_017018403.1 NR_125768.1

Gene Symbol
BBOX1-AS1
Entrez Gene ID
103695435
Gene Name
BBOX1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.196133
Gene Chromosome Location
Chr.11: 27047186-27220113 on Build GRCh38
Transcripts
NM_003986.2 XM_005253159.2 XM_005253160.2 XM_005253161.2 XM_011520402.1 XM_017018403.1 NR_125768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357318 c.37G>T p.G13W Chr.11: 27055467-27055467 on Build GRCh38
309436 c.16C>A p.Q6K Chr.11: 27055446-27055446 on Build GRCh38
3869048 c.58C>T p.L20F Chr.11: 27055488-27055488 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050205457 Chr.11: 27055259-27055259 on Build GRCh38
rs148033507 Chr.11: 27055438-27055438 on Build GRCh38
rs905410094 Chr.11: 27055343-27055343 on Build GRCh38
rs368465428 Chr.11: 27055437-27055437 on Build GRCh38
rs779558464 Chr.11: 27055412-27055412 on Build GRCh38
rs908658616 Chr.11: 27055315-27055315 on Build GRCh38
rs1017133397 Chr.11: 27055316-27055316 on Build GRCh38
rs934005809 Chr.11: 27055289-27055289 on Build GRCh38
rs568201060 Chr.11: 27055256-27055256 on Build GRCh38
rs551267414 Chr.11: 27055429-27055429 on Build GRCh38
rs536072159 Chr.11: 27055415-27055415 on Build GRCh38
rs759343729 Chr.11: 27055440-27055440 on Build GRCh38
rs200195953 Chr.11: 27055467-27055467 on Build GRCh38
rs551897570 Chr.11: 27055257-27055257 on Build GRCh38
rs768633123 Chr.11: 27055432-27055432 on Build GRCh38
rs372006148 Chr.11: 27055390-27055390 on Build GRCh38
rs765427088 Chr.11: 27055352-27055352 on Build GRCh38
rs964314198 Chr.11: 27055258-27055258 on Build GRCh38
rs1013909822 Chr.11: 27055423-27055423 on Build GRCh38
rs2305095 Chr.11: 27055430-27055430 on Build GRCh38
rs757839184 Chr.11: 27055406-27055406 on Build GRCh38
rs975412989 Chr.11: 27055273-27055273 on Build GRCh38
rs764359575 Chr.11: 27055252-27055252 on Build GRCh38
rs570470650 Chr.11: 27055326-27055326 on Build GRCh38
rs1047168333 Chr.11: 27055313-27055313 on Build GRCh38
rs922617468 Chr.11: 27055279-27055279 on Build GRCh38
rs758517397 Chr.11: 27055425-27055425 on Build GRCh38
rs377355425 Chr.11: 27055384-27055384 on Build GRCh38
rs540166569 Chr.11: 27055471-27055471 on Build GRCh38
rs376941503 Chr.11: 27055404-27055404 on Build GRCh38
rs56349897 Chr.11: 27055342-27055342 on Build GRCh38
rs746274716 Chr.11: 27055424-27055424 on Build GRCh38

View Full Product Details