Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4953174-4953425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO3
Entrez Gene ID
2027
Gene Name
enolase 3
Gene Aliases
GSD13, MSE
UniGene
Hs.224171
Gene Chromosome Location
Chr.17: 4949182-4957131 on Build GRCh38
Transcripts
NM_001193503.1 NM_001976.4 NM_053013.3 XM_005256521.2 XM_011523729.1 XM_017024346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680985 c.275A>C p.K92T Chr.17: 4953306-4953306 on Build GRCh38
4067630 c.255G>A p.V85V Chr.17: 4953286-4953286 on Build GRCh38
4829107 c.301G>A p.E101K Chr.17: 4953332-4953332 on Build GRCh38
981057 c.241-1G>T p.? Chr.17: 4953271-4953271 on Build GRCh38
6081515 c.305A>G p.N102S Chr.17: 4953336-4953336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754154618 Chr.17: 4953330-4953330 on Build GRCh38
rs764391024 Chr.17: 4953329-4953329 on Build GRCh38
rs780926849 Chr.17: 4953343-4953343 on Build GRCh38
rs756356937 Chr.17: 4953307-4953307 on Build GRCh38
rs776135288 Chr.17: 4953227-4953227 on Build GRCh38
rs759892571 Chr.17: 4953358-4953358 on Build GRCh38
rs546785438 Chr.17: 4953194-4953194 on Build GRCh38
rs549806535 Chr.17: 4953348-4953348 on Build GRCh38
rs537421309 Chr.17: 4953403-4953403 on Build GRCh38
rs34399160 Chr.17: 4953298-4953299 on Build GRCh38
rs747034249 Chr.17: 4953235-4953235 on Build GRCh38
rs371895662 Chr.17: 4953354-4953354 on Build GRCh38
rs570595152 Chr.17: 4953281-4953281 on Build GRCh38
rs761178397 Chr.17: 4953244-4953244 on Build GRCh38
rs764620224 Chr.17: 4953368-4953368 on Build GRCh38
rs765949352 Chr.17: 4953276-4953276 on Build GRCh38
rs1029617418 Chr.17: 4953287-4953287 on Build GRCh38
rs779111454 Chr.17: 4953334-4953334 on Build GRCh38
rs747956058 Chr.17: 4953346-4953346 on Build GRCh38
rs145859308 Chr.17: 4953280-4953280 on Build GRCh38
rs768163624 Chr.17: 4953359-4953359 on Build GRCh38
rs199644334 Chr.17: 4953252-4953252 on Build GRCh38
rs200608211 Chr.17: 4953286-4953286 on Build GRCh38
rs989188780 Chr.17: 4953384-4953384 on Build GRCh38
rs930161930 Chr.17: 4953211-4953211 on Build GRCh38
rs754951263 Chr.17: 4953338-4953338 on Build GRCh38
rs753997078 Chr.17: 4953369-4953369 on Build GRCh38
rs566502858 Chr.17: 4953218-4953218 on Build GRCh38
rs757517899 Chr.17: 4953386-4953386 on Build GRCh38
rs200385923 Chr.17: 4953245-4953245 on Build GRCh38
rs761378499 Chr.17: 4953275-4953276 on Build GRCh38
rs238239 Chr.17: 4953285-4953285 on Build GRCh38
rs757279282 Chr.17: 4953332-4953332 on Build GRCh38
rs201855213 Chr.17: 4953254-4953254 on Build GRCh38
rs762343282 Chr.17: 4953273-4953273 on Build GRCh38
rs113298706 Chr.17: 4953342-4953342 on Build GRCh38
rs770333521 Chr.17: 4953352-4953352 on Build GRCh38
rs111900624 Chr.17: 4953269-4953269 on Build GRCh38
rs753114346 Chr.17: 4953293-4953293 on Build GRCh38
rs913239617 Chr.17: 4953397-4953397 on Build GRCh38
rs34935329 Chr.17: 4953252-4953253 on Build GRCh38
rs917488664 Chr.17: 4953195-4953195 on Build GRCh38
rs765166893 Chr.17: 4953390-4953390 on Build GRCh38
rs771094653 Chr.17: 4953225-4953225 on Build GRCh38
rs746866973 Chr.17: 4953337-4953337 on Build GRCh38
rs769465776 Chr.17: 4953261-4953261 on Build GRCh38
rs749036548 Chr.17: 4953351-4953351 on Build GRCh38
rs182308848 Chr.17: 4953362-4953362 on Build GRCh38

View Full Product Details