Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46615514-46615739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HARBI1
Entrez Gene ID
283254
Gene Name
harbinger transposase derived 1
Gene Aliases
C11orf77
UniGene
Hs.714463
Gene Chromosome Location
Chr.11: 46603027-46617651 on Build GRCh38
Transcripts
NM_173811.3 XM_011520025.1 XM_011520026.2 XM_011520027.1 XM_011520028.2 XM_011520029.2 XM_017017602.1 XM_017017603.1 XM_017017604.1 XM_017017605.1 XM_017017606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5144308 c.670+8T>C p.? Chr.11: 46615560-46615560 on Build GRCh38
6677618 c.503G>A p.R168Q Chr.11: 46615735-46615735 on Build GRCh38
6055352 c.564C>T p.T188T Chr.11: 46615674-46615674 on Build GRCh38
5469610 c.502C>T p.R168* Chr.11: 46615736-46615736 on Build GRCh38
6677620 c.664C>A p.L222I Chr.11: 46615574-46615574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751471391 Chr.11: 46615564-46615564 on Build GRCh38
rs775298236 Chr.11: 46615674-46615674 on Build GRCh38
rs147599879 Chr.11: 46615616-46615616 on Build GRCh38
rs770206258 Chr.11: 46615649-46615649 on Build GRCh38
rs766524874 Chr.11: 46615704-46615704 on Build GRCh38
rs767296960 Chr.11: 46615573-46615573 on Build GRCh38
rs746885615 Chr.11: 46615673-46615673 on Build GRCh38
rs199580977 Chr.11: 46615669-46615669 on Build GRCh38
rs750740861 Chr.11: 46615612-46615612 on Build GRCh38
rs750650747 Chr.11: 46615694-46615694 on Build GRCh38
rs1034125649 Chr.11: 46615702-46615702 on Build GRCh38
rs147076911 Chr.11: 46615656-46615656 on Build GRCh38
rs769072419 Chr.11: 46615638-46615638 on Build GRCh38
rs1035815398 Chr.11: 46615710-46615710 on Build GRCh38
rs758875912 Chr.11: 46615686-46615686 on Build GRCh38
rs559220162 Chr.11: 46615650-46615650 on Build GRCh38
rs1036304406 Chr.11: 46615629-46615629 on Build GRCh38
rs764019374 Chr.11: 46615679-46615679 on Build GRCh38
rs932450521 Chr.11: 46615554-46615554 on Build GRCh38
rs1023608380 Chr.11: 46615663-46615663 on Build GRCh38
rs540421345 Chr.11: 46615625-46615625 on Build GRCh38
rs779291856 Chr.11: 46615597-46615597 on Build GRCh38
rs758521139 Chr.11: 46615613-46615613 on Build GRCh38
rs751591792 Chr.11: 46615720-46615720 on Build GRCh38
rs748471285 Chr.11: 46615647-46615647 on Build GRCh38
rs201216223 Chr.11: 46615614-46615614 on Build GRCh38
rs765944145 Chr.11: 46615551-46615551 on Build GRCh38
rs112306737 Chr.11: 46615591-46615591 on Build GRCh38
rs759443202 Chr.11: 46615567-46615567 on Build GRCh38
rs200844501 Chr.11: 46615658-46615658 on Build GRCh38
rs748982160 Chr.11: 46615655-46615655 on Build GRCh38
rs141749998 Chr.11: 46615645-46615645 on Build GRCh38
rs138485412 Chr.11: 46615695-46615695 on Build GRCh38
rs752467095 Chr.11: 46615579-46615579 on Build GRCh38
rs760542239 Chr.11: 46615678-46615678 on Build GRCh38
rs376539269 Chr.11: 46615584-46615584 on Build GRCh38
rs772850336 Chr.11: 46615652-46615652 on Build GRCh38

View Full Product Details