Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58193902-58194129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPX
Entrez Gene ID
8288
Gene Name
eosinophil peroxidase
Gene Aliases
EPO, EPP, EPX-PEN, EPXD
UniGene
Hs.279259
Gene Chromosome Location
Chr.17: 58192724-58205174 on Build GRCh38
Transcripts
NM_000502.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
981735 c.540G>A p.S180S Chr.17: 58194038-58194038 on Build GRCh38
4501476 c.589C>T p.P197S Chr.17: 58194087-58194087 on Build GRCh38
981734 c.472C>G p.P158A Chr.17: 58193970-58193970 on Build GRCh38
6902785 c.468G>A p.R156R Chr.17: 58193966-58193966 on Build GRCh38
4799314 c.531T>C p.D177D Chr.17: 58194029-58194029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149915044 Chr.17: 58194063-58194063 on Build GRCh38
rs781647078 Chr.17: 58194075-58194075 on Build GRCh38
rs368394525 Chr.17: 58194033-58194033 on Build GRCh38
rs757297876 Chr.17: 58193943-58193943 on Build GRCh38
rs748905543 Chr.17: 58194003-58194003 on Build GRCh38
rs758563196 Chr.17: 58194048-58194048 on Build GRCh38
rs757921967 Chr.17: 58193929-58193930 on Build GRCh38
rs780229286 Chr.17: 58194004-58194004 on Build GRCh38
rs903413866 Chr.17: 58194069-58194069 on Build GRCh38
rs764210039 Chr.17: 58193936-58193936 on Build GRCh38
rs375023183 Chr.17: 58193982-58193982 on Build GRCh38
rs758834598 Chr.17: 58193968-58193968 on Build GRCh38
rs536094300 Chr.17: 58194017-58194017 on Build GRCh38
rs374967566 Chr.17: 58194103-58194103 on Build GRCh38
rs371602457 Chr.17: 58194101-58194101 on Build GRCh38
rs777256507 Chr.17: 58193990-58193990 on Build GRCh38
rs756906529 Chr.17: 58194087-58194087 on Build GRCh38
rs747112771 Chr.17: 58194073-58194073 on Build GRCh38
rs750513069 Chr.17: 58193961-58193961 on Build GRCh38
rs765262486 Chr.17: 58193945-58193945 on Build GRCh38
rs200350821 Chr.17: 58194019-58194019 on Build GRCh38
rs139070509 Chr.17: 58194018-58194018 on Build GRCh38
rs753242381 Chr.17: 58194097-58194097 on Build GRCh38
rs776444431 Chr.17: 58194038-58194038 on Build GRCh38
rs569015114 Chr.17: 58193974-58193974 on Build GRCh38
rs765990684 Chr.17: 58194011-58194017 on Build GRCh38
rs765448449 Chr.17: 58194041-58194041 on Build GRCh38
rs971831852 Chr.17: 58193996-58193996 on Build GRCh38
rs150719520 Chr.17: 58194015-58194015 on Build GRCh38
rs760589017 Chr.17: 58193933-58193933 on Build GRCh38
rs566700181 Chr.17: 58194047-58194047 on Build GRCh38
rs775084013 Chr.17: 58193930-58193930 on Build GRCh38
rs748539431 Chr.17: 58194081-58194081 on Build GRCh38
rs1001276555 Chr.17: 58193992-58193992 on Build GRCh38
rs144998259 Chr.17: 58194070-58194070 on Build GRCh38
rs745362577 Chr.17: 58194098-58194098 on Build GRCh38
rs778778855 Chr.17: 58194093-58194093 on Build GRCh38
rs372501481 Chr.17: 58194022-58194022 on Build GRCh38
rs755469362 Chr.17: 58194072-58194072 on Build GRCh38
rs767507520 Chr.17: 58193928-58193928 on Build GRCh38
rs763991886 Chr.17: 58194023-58194023 on Build GRCh38
rs761963882 Chr.17: 58194039-58194039 on Build GRCh38
rs772751329 Chr.17: 58193969-58193969 on Build GRCh38

View Full Product Details