Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132140654-132140919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX51
Entrez Gene ID
317781
Gene Name
DEAD-box helicase 51
Gene Aliases
-
UniGene
Hs.445168
Gene Chromosome Location
Chr.12: 132136594-132144335 on Build GRCh38
Transcripts
NM_175066.3 XM_011538256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6259533 c.1514C>A p.S505* Chr.12: 132140662-132140662 on Build GRCh38
6580224 c.1442A>C p.H481P Chr.12: 132140734-132140734 on Build GRCh38
356744 c.1384G>A p.E462K Chr.12: 132140887-132140887 on Build GRCh38
3968103 c.1402G>A p.G468R Chr.12: 132140869-132140869 on Build GRCh38
4575229 c.1482C>T p.V494V Chr.12: 132140694-132140694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745606463 Chr.12: 132140699-132140699 on Build GRCh38
rs758567119 Chr.12: 132140860-132140860 on Build GRCh38
rs767131732 Chr.12: 132140874-132140874 on Build GRCh38
rs748312820 Chr.12: 132140797-132140797 on Build GRCh38
rs777274604 Chr.12: 132140724-132140724 on Build GRCh38
rs763164909 Chr.12: 132140707-132140707 on Build GRCh38
rs764931502 Chr.12: 132140892-132140892 on Build GRCh38
rs970982165 Chr.12: 132140789-132140789 on Build GRCh38
rs200705249 Chr.12: 132140872-132140872 on Build GRCh38
rs770047914 Chr.12: 132140870-132140870 on Build GRCh38
rs755284245 Chr.12: 132140781-132140781 on Build GRCh38
rs767583936 Chr.12: 132140869-132140869 on Build GRCh38
rs201060990 Chr.12: 132140831-132140831 on Build GRCh38
rs368116347 Chr.12: 132140786-132140786 on Build GRCh38
rs370515176 Chr.12: 132140712-132140712 on Build GRCh38
rs775830920 Chr.12: 132140702-132140702 on Build GRCh38
rs756698472 Chr.12: 132140767-132140767 on Build GRCh38
rs181712364 Chr.12: 132140862-132140862 on Build GRCh38
rs761072618 Chr.12: 132140757-132140757 on Build GRCh38
rs756125722 Chr.12: 132140832-132140832 on Build GRCh38
rs750697459 Chr.12: 132140681-132140681 on Build GRCh38
rs766475777 Chr.12: 132140886-132140886 on Build GRCh38
rs757630220 Chr.12: 132140675-132140675 on Build GRCh38
rs556412592 Chr.12: 132140687-132140687 on Build GRCh38
rs528080405 Chr.12: 132140697-132140697 on Build GRCh38
rs754664470 Chr.12: 132140682-132140682 on Build GRCh38
rs376428090 Chr.12: 132140701-132140701 on Build GRCh38
rs762395342 Chr.12: 132140714-132140714 on Build GRCh38
rs9787982 Chr.12: 132140773-132140773 on Build GRCh38
rs541798025 Chr.12: 132140693-132140693 on Build GRCh38
rs749025730 Chr.12: 132140780-132140780 on Build GRCh38
rs201297755 Chr.12: 132140732-132140732 on Build GRCh38
rs374520857 Chr.12: 132140765-132140765 on Build GRCh38
rs770711752 Chr.12: 132140694-132140694 on Build GRCh38
rs200438999 Chr.12: 132140728-132140728 on Build GRCh38
rs145968698 Chr.12: 132140849-132140849 on Build GRCh38
rs773572429 Chr.12: 132140756-132140756 on Build GRCh38
rs199645774 Chr.12: 132140893-132140893 on Build GRCh38
rs759444805 Chr.12: 132140891-132140891 on Build GRCh38
rs773842316 Chr.12: 132140826-132140826 on Build GRCh38
rs777096852 Chr.12: 132140867-132140867 on Build GRCh38
rs372914269 Chr.12: 132140840-132140840 on Build GRCh38
rs376327637 Chr.12: 132140766-132140766 on Build GRCh38
rs190948122 Chr.12: 132140771-132140771 on Build GRCh38
rs58876661 Chr.12: 132140816-132140816 on Build GRCh38
rs760901644 Chr.12: 132140721-132140721 on Build GRCh38
rs772880022 Chr.12: 132140884-132140884 on Build GRCh38
rs774494879 Chr.12: 132140750-132140750 on Build GRCh38
rs541123673 Chr.12: 132140742-132140742 on Build GRCh38
rs767911012 Chr.12: 132140679-132140679 on Build GRCh38
rs377232805 Chr.12: 132140861-132140861 on Build GRCh38
rs369866312 Chr.12: 132140889-132140889 on Build GRCh38
rs199655060 Chr.12: 132140851-132140851 on Build GRCh38
rs546587047 Chr.12: 132140745-132140745 on Build GRCh38
rs373479741 Chr.12: 132140690-132140690 on Build GRCh38
rs369566115 Chr.12: 132140684-132140684 on Build GRCh38
rs748318175 Chr.12: 132140852-132140852 on Build GRCh38
rs367574820 Chr.12: 132140834-132140834 on Build GRCh38
rs750963623 Chr.12: 132140726-132140726 on Build GRCh38
rs761384473 Chr.12: 132140873-132140873 on Build GRCh38
rs958805647 Chr.12: 132140838-132140838 on Build GRCh38
rs762594877 Chr.12: 132140764-132140764 on Build GRCh38
rs752319002 Chr.12: 132140723-132140723 on Build GRCh38
rs111226738 Chr.12: 132140817-132140817 on Build GRCh38
rs766923584 Chr.12: 132140829-132140829 on Build GRCh38
rs772659636 Chr.12: 132140754-132140754 on Build GRCh38
rs371028064 Chr.12: 132140746-132140746 on Build GRCh38
rs777999364 Chr.12: 132140897-132140897 on Build GRCh38
rs186508341 Chr.12: 132140748-132140748 on Build GRCh38
rs1025607651 Chr.12: 132140795-132140795 on Build GRCh38
rs754140871 Chr.12: 132140759-132140759 on Build GRCh38
rs763853237 Chr.12: 132140823-132140823 on Build GRCh38
rs773956797 Chr.12: 132140895-132140896 on Build GRCh38
rs772017603 Chr.12: 132140812-132140812 on Build GRCh38
rs1009264699 Chr.12: 132140778-132140778 on Build GRCh38
rs764920253 Chr.12: 132140722-132140722 on Build GRCh38
rs777006833 Chr.12: 132140819-132140819 on Build GRCh38
rs781227273 Chr.12: 132140769-132140769 on Build GRCh38
rs780939753 Chr.12: 132140731-132140731 on Build GRCh38
rs576061161 Chr.12: 132140691-132140691 on Build GRCh38
rs750322812 Chr.12: 132140770-132140770 on Build GRCh38
rs759842323 Chr.12: 132140822-132140822 on Build GRCh38
rs750831881 Chr.12: 132140715-132140715 on Build GRCh38

View Full Product Details