Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53169963-53170234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSAD
Entrez Gene ID
51380
Gene Name
cysteine sulfinic acid decarboxylase
Gene Aliases
CSD, PCAP
UniGene
Hs.279815
Gene Chromosome Location
Chr.12: 53157663-53180909 on Build GRCh38
Transcripts
NM_001244705.1 NM_015989.4 XM_011538442.2 XM_011538446.2 XM_011538449.1 XM_011538451.1 XM_017019416.1 XM_017019417.1 XM_017019418.1 XM_017019419.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4961265 c.691G>A p.A231T Chr.12: 53170083-53170083 on Build GRCh38
5375455 c.751C>A p.L251M Chr.12: 53170104-53170104 on Build GRCh38
3462601 c.743C>T p.P248L Chr.12: 53170112-53170112 on Build GRCh38
3416950 c.754G>A p.E252K Chr.12: 53170101-53170101 on Build GRCh38
3998853 c.693C>T p.A231A Chr.12: 53170081-53170081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778800317 Chr.12: 53170116-53170116 on Build GRCh38
rs780678209 Chr.12: 53170100-53170100 on Build GRCh38
rs759550752 Chr.12: 53170063-53170063 on Build GRCh38
rs879138011 Chr.12: 53170073-53170073 on Build GRCh38
rs375625717 Chr.12: 53170111-53170111 on Build GRCh38
rs373113301 Chr.12: 53170042-53170042 on Build GRCh38
rs371198598 Chr.12: 53170156-53170156 on Build GRCh38
rs558408776 Chr.12: 53170080-53170080 on Build GRCh38
rs761747194 Chr.12: 53170076-53170081 on Build GRCh38
rs778008516 Chr.12: 53170141-53170141 on Build GRCh38
rs936169265 Chr.12: 53170071-53170071 on Build GRCh38
rs568874300 Chr.12: 53170140-53170140 on Build GRCh38
rs199570525 Chr.12: 53170033-53170033 on Build GRCh38
rs11170453 Chr.12: 53170081-53170081 on Build GRCh38
rs770869402 Chr.12: 53170054-53170054 on Build GRCh38
rs765016890 Chr.12: 53170130-53170130 on Build GRCh38
rs758613723 Chr.12: 53170139-53170139 on Build GRCh38
rs756687811 Chr.12: 53170029-53170029 on Build GRCh38
rs746872400 Chr.12: 53170050-53170050 on Build GRCh38
rs984251059 Chr.12: 53169987-53169987 on Build GRCh38
rs111553950 Chr.12: 53170144-53170144 on Build GRCh38
rs146549996 Chr.12: 53170150-53170150 on Build GRCh38
rs962577528 Chr.12: 53170166-53170166 on Build GRCh38
rs199564889 Chr.12: 53170051-53170051 on Build GRCh38
rs766786071 Chr.12: 53170108-53170108 on Build GRCh38
rs139395448 Chr.12: 53170083-53170083 on Build GRCh38
rs757985717 Chr.12: 53170053-53170053 on Build GRCh38
rs1026003823 Chr.12: 53170202-53170202 on Build GRCh38
rs762684568 Chr.12: 53170088-53170088 on Build GRCh38
rs754960637 Chr.12: 53170037-53170037 on Build GRCh38
rs760991926 Chr.12: 53170106-53170106 on Build GRCh38
rs776385790 Chr.12: 53170055-53170055 on Build GRCh38
rs950215186 Chr.12: 53170035-53170035 on Build GRCh38
rs550597583 Chr.12: 53170117-53170117 on Build GRCh38
rs995408147 Chr.12: 53170201-53170201 on Build GRCh38
rs983535413 Chr.12: 53169991-53169991 on Build GRCh38
rs750787966 Chr.12: 53170092-53170092 on Build GRCh38
rs745735128 Chr.12: 53170154-53170154 on Build GRCh38
rs770131666 Chr.12: 53170195-53170195 on Build GRCh38
rs149431326 Chr.12: 53170110-53170110 on Build GRCh38
rs770593127 Chr.12: 53170145-53170145 on Build GRCh38
rs772295163 Chr.12: 53170043-53170043 on Build GRCh38
rs112284599 Chr.12: 53170190-53170190 on Build GRCh38
rs115571635 Chr.12: 53170008-53170008 on Build GRCh38
rs77656161 Chr.12: 53170044-53170044 on Build GRCh38
rs763908853 Chr.12: 53170089-53170089 on Build GRCh38
rs958469021 Chr.12: 53170020-53170020 on Build GRCh38

View Full Product Details