Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156844056-156844329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSRR
Entrez Gene ID
3645
Gene Name
insulin receptor related receptor
Gene Aliases
IRR
UniGene
Hs.248138
Gene Chromosome Location
Chr.1: 156840873-156858920 on Build GRCh38
Transcripts
NM_014215.2 NM_001007792.1

Gene Symbol
NTRK1
Entrez Gene ID
4914
Gene Name
neurotrophic receptor tyrosine kinase 1
Gene Aliases
MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA
UniGene
Hs.406293
Gene Chromosome Location
Chr.1: 156815750-156881850 on Build GRCh38
Transcripts
NM_014215.2 NM_001007792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146642 c.2783C>T p.P928L Chr.1: 156844235-156844235 on Build GRCh38
4023427 c.2797C>T p.L933L Chr.1: 156844221-156844221 on Build GRCh38
6686330 c.2806G>A p.V936I Chr.1: 156844212-156844212 on Build GRCh38
5052657 c.2783delC p.P928fs*29 Chr.1: 156844235-156844235 on Build GRCh38
3475430 c.2782C>T p.P928S Chr.1: 156844236-156844236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983104346 Chr.1: 156844260-156844260 on Build GRCh38
rs976482883 Chr.1: 156844079-156844079 on Build GRCh38
rs763667728 Chr.1: 156844228-156844228 on Build GRCh38
rs748173098 Chr.1: 156844290-156844290 on Build GRCh38
rs771422531 Chr.1: 156844203-156844203 on Build GRCh38
rs756289726 Chr.1: 156844280-156844280 on Build GRCh38
rs757161901 Chr.1: 156844162-156844162 on Build GRCh38
rs953173759 Chr.1: 156844081-156844081 on Build GRCh38
rs555373809 Chr.1: 156844185-156844185 on Build GRCh38
rs759870796 Chr.1: 156844216-156844216 on Build GRCh38
rs909007603 Chr.1: 156844107-156844107 on Build GRCh38
rs893347254 Chr.1: 156844137-156844137 on Build GRCh38
rs575113619 Chr.1: 156844269-156844269 on Build GRCh38
rs56252149 Chr.1: 156844235-156844235 on Build GRCh38
rs372329556 Chr.1: 156844261-156844261 on Build GRCh38
rs921041417 Chr.1: 156844084-156844084 on Build GRCh38
rs965635838 Chr.1: 156844240-156844240 on Build GRCh38
rs139924091 Chr.1: 156844281-156844281 on Build GRCh38
rs578124397 Chr.1: 156844296-156844296 on Build GRCh38
rs200756843 Chr.1: 156844151-156844151 on Build GRCh38
rs374540478 Chr.1: 156844156-156844156 on Build GRCh38
rs760327047 Chr.1: 156844223-156844223 on Build GRCh38
rs113403460 Chr.1: 156844157-156844157 on Build GRCh38
rs749373554 Chr.1: 156844298-156844298 on Build GRCh38
rs779943504 Chr.1: 156844265-156844265 on Build GRCh38
rs374247650 Chr.1: 156844153-156844153 on Build GRCh38
rs779001847 Chr.1: 156844163-156844163 on Build GRCh38
rs112863279 Chr.1: 156844148-156844148 on Build GRCh38
rs997138313 Chr.1: 156844295-156844295 on Build GRCh38
rs772259106 Chr.1: 156844261-156844261 on Build GRCh38
rs368569743 Chr.1: 156844219-156844219 on Build GRCh38
rs758328689 Chr.1: 156844180-156844180 on Build GRCh38
rs755066293 Chr.1: 156844106-156844106 on Build GRCh38
rs145379996 Chr.1: 156844186-156844186 on Build GRCh38
rs765255663 Chr.1: 156844256-156844256 on Build GRCh38
rs1051050101 Chr.1: 156844130-156844130 on Build GRCh38
rs539255716 Chr.1: 156844114-156844114 on Build GRCh38
rs753311076 Chr.1: 156844160-156844160 on Build GRCh38
rs141174426 Chr.1: 156844139-156844139 on Build GRCh38
rs372319737 Chr.1: 156844200-156844200 on Build GRCh38
rs745658294 Chr.1: 156844168-156844168 on Build GRCh38
rs775750741 Chr.1: 156844222-156844222 on Build GRCh38
rs767911715 Chr.1: 156844220-156844220 on Build GRCh38
rs965031886 Chr.1: 156844284-156844284 on Build GRCh38
rs374329265 Chr.1: 156844212-156844212 on Build GRCh38
rs377318206 Chr.1: 156844242-156844242 on Build GRCh38
rs746253332 Chr.1: 156844234-156844234 on Build GRCh38
rs984573138 Chr.1: 156844097-156844097 on Build GRCh38

View Full Product Details