Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74749961-74750218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1A2
Entrez Gene ID
1544
Gene Name
cytochrome P450 family 1 subfamily A member 2
Gene Aliases
CP12, P3-450, P450(PA)
UniGene
Hs.1361
Gene Chromosome Location
Chr.15: 74748843-74756600 on Build GRCh38
Transcripts
NM_000761.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
471060 c.289G>A p.A97T Chr.15: 74750027-74750027 on Build GRCh38
243936 c.407G>A p.R136H Chr.15: 74750145-74750145 on Build GRCh38
471059 c.249G>A p.T83T Chr.15: 74749987-74749987 on Build GRCh38
256720 c.348C>T p.L116L Chr.15: 74750086-74750086 on Build GRCh38
1374560 c.269G>A p.R90H Chr.15: 74750007-74750007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200550773 Chr.15: 74750105-74750105 on Build GRCh38
rs768884287 Chr.15: 74750179-74750179 on Build GRCh38
rs755802066 Chr.15: 74750013-74750013 on Build GRCh38
rs71541923 Chr.15: 74750086-74750086 on Build GRCh38
rs936160269 Chr.15: 74750079-74750079 on Build GRCh38
rs780591326 Chr.15: 74750013-74750019 on Build GRCh38
rs753307975 Chr.15: 74750134-74750134 on Build GRCh38
rs998243684 Chr.15: 74750124-74750124 on Build GRCh38
rs1038912692 Chr.15: 74750166-74750166 on Build GRCh38
rs774966785 Chr.15: 74750181-74750181 on Build GRCh38
rs201820772 Chr.15: 74750078-74750078 on Build GRCh38
rs752542857 Chr.15: 74750146-74750146 on Build GRCh38
rs768231616 Chr.15: 74750069-74750070 on Build GRCh38
rs201551575 Chr.15: 74750007-74750007 on Build GRCh38
rs762845440 Chr.15: 74750197-74750197 on Build GRCh38
rs962518660 Chr.15: 74749996-74749996 on Build GRCh38
rs192799115 Chr.15: 74750186-74750186 on Build GRCh38
rs770091203 Chr.15: 74750099-74750099 on Build GRCh38
rs773366123 Chr.15: 74750031-74750031 on Build GRCh38
rs368187861 Chr.15: 74750144-74750144 on Build GRCh38
rs137918189 Chr.15: 74750038-74750038 on Build GRCh38
rs763535401 Chr.15: 74750045-74750045 on Build GRCh38
rs537906318 Chr.15: 74750194-74750194 on Build GRCh38
rs758370714 Chr.15: 74750074-74750074 on Build GRCh38
rs745883159 Chr.15: 74750153-74750153 on Build GRCh38
rs749152552 Chr.15: 74750021-74750021 on Build GRCh38
rs148769292 Chr.15: 74750050-74750050 on Build GRCh38
rs34067076 Chr.15: 74750048-74750048 on Build GRCh38
rs770142375 Chr.15: 74750159-74750159 on Build GRCh38
rs141543251 Chr.15: 74750113-74750113 on Build GRCh38
rs34153904 Chr.15: 74750131-74750131 on Build GRCh38
rs556218799 Chr.15: 74750006-74750006 on Build GRCh38
rs1031616307 Chr.15: 74750054-74750054 on Build GRCh38
rs752279053 Chr.15: 74750195-74750195 on Build GRCh38
rs780416016 Chr.15: 74750163-74750163 on Build GRCh38
rs754811709 Chr.15: 74750066-74750066 on Build GRCh38
rs772521404 Chr.15: 74750188-74750188 on Build GRCh38
rs17861154 Chr.15: 74750044-74750044 on Build GRCh38
rs145266863 Chr.15: 74750036-74750036 on Build GRCh38
rs138652540 Chr.15: 74749986-74749986 on Build GRCh38
rs377141209 Chr.15: 74750084-74750084 on Build GRCh38
rs551386466 Chr.15: 74750149-74750149 on Build GRCh38
rs746122928 Chr.15: 74750097-74750097 on Build GRCh38
rs781680722 Chr.15: 74750028-74750028 on Build GRCh38
rs758981719 Chr.15: 74750192-74750192 on Build GRCh38
rs763157275 Chr.15: 74750109-74750109 on Build GRCh38
rs560690067 Chr.15: 74750032-74750032 on Build GRCh38
rs774711732 Chr.15: 74750120-74750120 on Build GRCh38
rs759912942 Chr.15: 74749991-74749991 on Build GRCh38
rs764447434 Chr.15: 74750047-74750047 on Build GRCh38
rs754758658 Chr.15: 74750022-74750022 on Build GRCh38
rs59410695 Chr.15: 74750151-74750151 on Build GRCh38
rs766082944 Chr.15: 74750060-74750060 on Build GRCh38
rs747471370 Chr.15: 74750063-74750063 on Build GRCh38
rs777148966 Chr.15: 74749990-74749990 on Build GRCh38
rs779502284 Chr.15: 74750016-74750016 on Build GRCh38
rs139412032 Chr.15: 74750148-74750148 on Build GRCh38
rs150893756 Chr.15: 74750145-74750145 on Build GRCh38
rs756865803 Chr.15: 74750002-74750002 on Build GRCh38
rs55802037 Chr.15: 74750111-74750111 on Build GRCh38
rs757176570 Chr.15: 74750150-74750150 on Build GRCh38
rs376179316 Chr.15: 74750061-74750061 on Build GRCh38
rs747181148 Chr.15: 74750080-74750080 on Build GRCh38
rs763825636 Chr.15: 74750198-74750198 on Build GRCh38
rs17861153 Chr.15: 74749987-74749987 on Build GRCh38
rs760284408 Chr.15: 74750130-74750130 on Build GRCh38
rs376107702 Chr.15: 74749984-74749984 on Build GRCh38
rs45442197 Chr.15: 74750069-74750069 on Build GRCh38
rs747891878 Chr.15: 74750073-74750073 on Build GRCh38
rs753232890 Chr.15: 74749995-74749995 on Build GRCh38
rs201650099 Chr.15: 74750037-74750037 on Build GRCh38
rs545726003 Chr.15: 74750027-74750027 on Build GRCh38
rs770950827 Chr.15: 74750190-74750190 on Build GRCh38
rs758124536 Chr.15: 74750147-74750147 on Build GRCh38
rs749715866 Chr.15: 74750167-74750167 on Build GRCh38
rs766652315 Chr.15: 74749988-74749988 on Build GRCh38

View Full Product Details