Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 99849728-99850001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMSS1
Entrez Gene ID
84319
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Gene Aliases
C3orf26
UniGene
Hs.581320
Gene Chromosome Location
Chr.3: 99817834-100178632 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Gene Symbol
FILIP1L
Entrez Gene ID
11259
Gene Name
filamin A interacting protein 1 like
Gene Aliases
DOC-1, DOC1, GIP130, GIP90
UniGene
Hs.104672
Gene Chromosome Location
Chr.3: 99833144-100114513 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594140 c.1718C>T p.A573V Chr.3: 99849958-99849958 on Build GRCh38
7175987 c.1678G>C p.V560L Chr.3: 99849998-99849998 on Build GRCh38
1594141 c.1738G>T p.D580Y Chr.3: 99849938-99849938 on Build GRCh38
7175988 c.1678G>C p.V560L Chr.3: 99849998-99849998 on Build GRCh38
6327068 c.1938G>A p.E646E Chr.3: 99849738-99849738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565958020 Chr.3: 99849877-99849877 on Build GRCh38
rs774617850 Chr.3: 99849958-99849958 on Build GRCh38
rs759109740 Chr.3: 99849819-99849819 on Build GRCh38
rs757389027 Chr.3: 99849752-99849752 on Build GRCh38
rs776317854 Chr.3: 99849808-99849808 on Build GRCh38
rs751830028 Chr.3: 99849841-99849841 on Build GRCh38
rs200756899 Chr.3: 99849947-99849947 on Build GRCh38
rs989923549 Chr.3: 99849913-99849913 on Build GRCh38
rs777482850 Chr.3: 99849903-99849903 on Build GRCh38
rs980786696 Chr.3: 99849969-99849969 on Build GRCh38
rs765900679 Chr.3: 99849820-99849820 on Build GRCh38
rs374282857 Chr.3: 99849928-99849928 on Build GRCh38
rs374615472 Chr.3: 99849886-99849886 on Build GRCh38
rs756131330 Chr.3: 99849883-99849883 on Build GRCh38
rs766306040 Chr.3: 99849949-99849951 on Build GRCh38
rs536583796 Chr.3: 99849887-99849887 on Build GRCh38
rs763379772 Chr.3: 99849827-99849827 on Build GRCh38
rs998790461 Chr.3: 99849862-99849862 on Build GRCh38
rs778828328 Chr.3: 99849888-99849888 on Build GRCh38
rs374810121 Chr.3: 99849750-99849750 on Build GRCh38
rs576553372 Chr.3: 99849937-99849937 on Build GRCh38
rs553705584 Chr.3: 99849845-99849845 on Build GRCh38
rs747990242 Chr.3: 99849780-99849780 on Build GRCh38
rs767739889 Chr.3: 99849978-99849978 on Build GRCh38
rs745924720 Chr.3: 99849759-99849759 on Build GRCh38
rs781198757 Chr.3: 99849757-99849757 on Build GRCh38
rs757891591 Chr.3: 99849853-99849853 on Build GRCh38
rs771698937 Chr.3: 99849781-99849781 on Build GRCh38
rs370561748 Chr.3: 99849858-99849858 on Build GRCh38
rs1018798337 Chr.3: 99849844-99849844 on Build GRCh38
rs932977160 Chr.3: 99849851-99849851 on Build GRCh38
rs1037462013 Chr.3: 99849940-99849940 on Build GRCh38
rs773676808 Chr.3: 99849916-99849916 on Build GRCh38
rs200994891 Chr.3: 99849832-99849832 on Build GRCh38
rs372304983 Chr.3: 99849895-99849895 on Build GRCh38
rs754976384 Chr.3: 99849760-99849760 on Build GRCh38
rs775799851 Chr.3: 99849823-99849823 on Build GRCh38
rs751665212 Chr.3: 99849751-99849751 on Build GRCh38
rs113764818 Chr.3: 99849767-99849767 on Build GRCh38
rs770770044 Chr.3: 99849924-99849924 on Build GRCh38
rs746792143 Chr.3: 99849906-99849906 on Build GRCh38
rs150956085 Chr.3: 99849783-99849783 on Build GRCh38
rs957299782 Chr.3: 99849912-99849912 on Build GRCh38
rs9843741 Chr.3: 99849893-99849893 on Build GRCh38
rs776095371 Chr.3: 99849939-99849939 on Build GRCh38
rs769184193 Chr.3: 99849957-99849957 on Build GRCh38
rs770688390 Chr.3: 99849805-99849805 on Build GRCh38
rs368624908 Chr.3: 99849935-99849935 on Build GRCh38
rs79320113 Chr.3: 99849790-99849790 on Build GRCh38

View Full Product Details