Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58253967-58254238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC113
Entrez Gene ID
29070
Gene Name
coiled-coil domain containing 113
Gene Aliases
HSPC065
UniGene
Hs.11614
Gene Chromosome Location
Chr.16: 58196863-58283836 on Build GRCh38
Transcripts
NM_001142302.1 NM_014157.3 XM_011523038.2 XM_011523039.2 XM_011523040.2 XM_011523041.2 XM_011523042.2 XM_017023176.1 XM_017023177.1 XM_017023178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4669675 c.232C>T p.R78* Chr.16: 58254001-58254001 on Build GRCh38
4669676 c.346C>T p.R116* Chr.16: 58254115-58254115 on Build GRCh38
3969615 c.234A>G p.R78R Chr.16: 58254003-58254003 on Build GRCh38
4595639 c.290C>T p.A97V Chr.16: 58254059-58254059 on Build GRCh38
3510605 c.255C>T p.S85S Chr.16: 58254024-58254024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752651842 Chr.16: 58254076-58254076 on Build GRCh38
rs923923731 Chr.16: 58254041-58254041 on Build GRCh38
rs778708567 Chr.16: 58254006-58254006 on Build GRCh38
rs267604586 Chr.16: 58254024-58254024 on Build GRCh38
rs774828926 Chr.16: 58254045-58254045 on Build GRCh38
rs745931374 Chr.16: 58254030-58254030 on Build GRCh38
rs200282611 Chr.16: 58254139-58254139 on Build GRCh38
rs769267327 Chr.16: 58254138-58254138 on Build GRCh38
rs775877153 Chr.16: 58254035-58254035 on Build GRCh38
rs1028571988 Chr.16: 58254212-58254212 on Build GRCh38
rs150738036 Chr.16: 58254025-58254025 on Build GRCh38
rs113345318 Chr.16: 58254194-58254194 on Build GRCh38
rs140348706 Chr.16: 58254144-58254144 on Build GRCh38
rs774949690 Chr.16: 58254149-58254149 on Build GRCh38
rs758291430 Chr.16: 58254081-58254081 on Build GRCh38
rs745546942 Chr.16: 58254204-58254204 on Build GRCh38
rs762456767 Chr.16: 58254210-58254210 on Build GRCh38
rs765930226 Chr.16: 58254061-58254061 on Build GRCh38
rs756332667 Chr.16: 58254177-58254177 on Build GRCh38
rs372193450 Chr.16: 58254026-58254026 on Build GRCh38
rs756173682 Chr.16: 58254122-58254122 on Build GRCh38
rs775048173 Chr.16: 58254205-58254205 on Build GRCh38
rs147141699 Chr.16: 58254140-58254140 on Build GRCh38
rs538405856 Chr.16: 58254141-58254141 on Build GRCh38
rs754996587 Chr.16: 58254004-58254004 on Build GRCh38
rs757567093 Chr.16: 58254107-58254107 on Build GRCh38
rs375219394 Chr.16: 58254040-58254040 on Build GRCh38
rs144246110 Chr.16: 58254069-58254069 on Build GRCh38
rs752561841 Chr.16: 58254010-58254010 on Build GRCh38
rs148705207 Chr.16: 58254156-58254156 on Build GRCh38
rs930269617 Chr.16: 58254130-58254130 on Build GRCh38
rs1043881617 Chr.16: 58254033-58254033 on Build GRCh38
rs527971683 Chr.16: 58254029-58254029 on Build GRCh38
rs767295320 Chr.16: 58254167-58254167 on Build GRCh38
rs892258843 Chr.16: 58254163-58254163 on Build GRCh38
rs764014177 Chr.16: 58254157-58254157 on Build GRCh38
rs781277591 Chr.16: 58254115-58254115 on Build GRCh38
rs769847630 Chr.16: 58254034-58254034 on Build GRCh38
rs750644359 Chr.16: 58254174-58254174 on Build GRCh38
rs751432636 Chr.16: 58254161-58254161 on Build GRCh38
rs776269301 Chr.16: 58254068-58254068 on Build GRCh38
rs766093198 Chr.16: 58254095-58254096 on Build GRCh38
rs776159355 Chr.16: 58254142-58254142 on Build GRCh38
rs143378814 Chr.16: 58254096-58254096 on Build GRCh38
rs749854850 Chr.16: 58254135-58254135 on Build GRCh38
rs764744452 Chr.16: 58254071-58254071 on Build GRCh38
rs778158876 Chr.16: 58254200-58254200 on Build GRCh38
rs760448521 Chr.16: 58254060-58254060 on Build GRCh38
rs753262070 Chr.16: 58254001-58254001 on Build GRCh38
rs1034980690 Chr.16: 58254002-58254002 on Build GRCh38
rs762748203 Chr.16: 58254071-58254072 on Build GRCh38
rs996784642 Chr.16: 58254211-58254211 on Build GRCh38
rs573970589 Chr.16: 58254178-58254178 on Build GRCh38
rs765957496 Chr.16: 58253996-58253996 on Build GRCh38
rs751342865 Chr.16: 58254104-58254104 on Build GRCh38
rs201879453 Chr.16: 58254134-58254134 on Build GRCh38
rs755606419 Chr.16: 58254183-58254183 on Build GRCh38
rs201397794 Chr.16: 58254196-58254196 on Build GRCh38
rs759769664 Chr.16: 58254014-58254014 on Build GRCh38
rs757017463 Chr.16: 58254165-58254165 on Build GRCh38
rs200330631 Chr.16: 58254116-58254116 on Build GRCh38
rs772974950 Chr.16: 58254019-58254019 on Build GRCh38
rs199694141 Chr.16: 58254188-58254188 on Build GRCh38

View Full Product Details