Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2856037-2856284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS22
Entrez Gene ID
64063
Gene Name
protease, serine 22
Gene Aliases
BSSP-4, SP001LA, hBSSP-4
UniGene
Hs.459709
Gene Chromosome Location
Chr.16: 2852727-2858630 on Build GRCh38
Transcripts
NM_022119.3 XM_005255473.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
246812 c.145C>T p.R49W Chr.16: 2856218-2856218 on Build GRCh38
6838944 c.172G>A p.D58N Chr.16: 2856191-2856191 on Build GRCh38
235244 c.279G>A p.K93K Chr.16: 2856084-2856084 on Build GRCh38
5850664 c.230G>A p.G77D Chr.16: 2856133-2856133 on Build GRCh38
7296693 c.177C>T p.S59S Chr.16: 2856186-2856186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886349794 Chr.16: 2856130-2856130 on Build GRCh38
rs143105283 Chr.16: 2856158-2856158 on Build GRCh38
rs753532703 Chr.16: 2856242-2856242 on Build GRCh38
rs199522327 Chr.16: 2856087-2856087 on Build GRCh38
rs752861534 Chr.16: 2856067-2856067 on Build GRCh38
rs759759990 Chr.16: 2856115-2856115 on Build GRCh38
rs1006372344 Chr.16: 2856060-2856060 on Build GRCh38
rs74508509 Chr.16: 2856205-2856205 on Build GRCh38
rs764538745 Chr.16: 2856124-2856124 on Build GRCh38
rs777640779 Chr.16: 2856183-2856183 on Build GRCh38
rs769672751 Chr.16: 2856096-2856096 on Build GRCh38
rs780881542 Chr.16: 2856092-2856092 on Build GRCh38
rs767443986 Chr.16: 2856100-2856100 on Build GRCh38
rs376369354 Chr.16: 2856218-2856218 on Build GRCh38
rs754532142 Chr.16: 2856164-2856164 on Build GRCh38
rs753423446 Chr.16: 2856217-2856217 on Build GRCh38
rs749272435 Chr.16: 2856101-2856101 on Build GRCh38
rs374618225 Chr.16: 2856069-2856069 on Build GRCh38
rs754480261 Chr.16: 2856083-2856083 on Build GRCh38
rs149331401 Chr.16: 2856231-2856231 on Build GRCh38
rs369180451 Chr.16: 2856196-2856196 on Build GRCh38
rs367543461 Chr.16: 2856136-2856136 on Build GRCh38
rs754397226 Chr.16: 2856070-2856070 on Build GRCh38
rs139100187 Chr.16: 2856116-2856116 on Build GRCh38
rs150572986 Chr.16: 2856207-2856207 on Build GRCh38
rs760766476 Chr.16: 2856122-2856122 on Build GRCh38
rs757155159 Chr.16: 2856247-2856247 on Build GRCh38
rs767078604 Chr.16: 2856225-2856225 on Build GRCh38
rs757709018 Chr.16: 2856076-2856076 on Build GRCh38
rs879257097 Chr.16: 2856206-2856206 on Build GRCh38
rs1017376493 Chr.16: 2856188-2856188 on Build GRCh38
rs779551978 Chr.16: 2856077-2856077 on Build GRCh38
rs200802675 Chr.16: 2856170-2856170 on Build GRCh38
rs755798377 Chr.16: 2856237-2856237 on Build GRCh38
rs200614493 Chr.16: 2856063-2856063 on Build GRCh38
rs776906108 Chr.16: 2856210-2856210 on Build GRCh38
rs745764784 Chr.16: 2856198-2856198 on Build GRCh38
rs774526540 Chr.16: 2856062-2856062 on Build GRCh38
rs745819821 Chr.16: 2856257-2856257 on Build GRCh38
rs188850412 Chr.16: 2856203-2856203 on Build GRCh38
rs755674238 Chr.16: 2856176-2856176 on Build GRCh38
rs770937406 Chr.16: 2856107-2856107 on Build GRCh38
rs760344265 Chr.16: 2856147-2856147 on Build GRCh38
rs754092755 Chr.16: 2856125-2856125 on Build GRCh38
rs144584103 Chr.16: 2856252-2856252 on Build GRCh38
rs574367142 Chr.16: 2856186-2856186 on Build GRCh38
rs898306357 Chr.16: 2856125-2856137 on Build GRCh38
rs770849689 Chr.16: 2856195-2856195 on Build GRCh38
rs760515703 Chr.16: 2856143-2856143 on Build GRCh38
rs999479171 Chr.16: 2856216-2856216 on Build GRCh38
rs200464790 Chr.16: 2856204-2856204 on Build GRCh38
rs766598839 Chr.16: 2856061-2856061 on Build GRCh38
rs762771579 Chr.16: 2856059-2856059 on Build GRCh38
rs746447517 Chr.16: 2856080-2856080 on Build GRCh38
rs200127582 Chr.16: 2856138-2856138 on Build GRCh38
rs138580039 Chr.16: 2856224-2856224 on Build GRCh38
rs777359026 Chr.16: 2856241-2856241 on Build GRCh38
rs111229833 Chr.16: 2856137-2856137 on Build GRCh38
rs774581504 Chr.16: 2856108-2856108 on Build GRCh38

View Full Product Details