Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 20707816-20708041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PACRGL
Entrez Gene ID
133015
Gene Name
PARK2 coregulated like
Gene Aliases
C4orf28
UniGene
Hs.479298
Gene Chromosome Location
Chr.4: 20696282-20757766 on Build GRCh38
Transcripts
NM_001130727.1 NM_001258345.1 NM_001258346.1 NM_001317849.1 NM_145048.3 NR_047661.1 NR_133933.1 XM_011513780.1 XM_011513781.1 XM_011513784.1 XM_011513785.1 XM_011513786.2 XM_011513787.1 XM_011513788.1 XM_011513791.1 XM_011513792.1 XM_011513794.1 XM_011513795.1 XM_011513796.1 XM_011513797.2 XM_011513798.2 XM_011513804.1 XM_011513805.2 XM_017007747.1 XM_017007748.1 XM_017007749.1 XM_017007750.1 XM_017007751.1 XM_017007752.1 XM_017007753.1 XM_017007754.1 XM_017007755.1 XM_017007756.1 XM_017007757.1 XM_017007758.1 XM_017007759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4124067 c.265A>G p.I89V Chr.4: 20707860-20707860 on Build GRCh38
6736726 c.241G>A p.A81T Chr.4: 20707836-20707836 on Build GRCh38
6736727 c.241G>A p.A81T Chr.4: 20707836-20707836 on Build GRCh38
4124065 c.224G>A p.R75Q Chr.4: 20707819-20707819 on Build GRCh38
6383043 c.269C>T p.P90L Chr.4: 20707864-20707864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015989597 Chr.4: 20707986-20707986 on Build GRCh38
rs764692816 Chr.4: 20707903-20707903 on Build GRCh38
rs976478045 Chr.4: 20707974-20707974 on Build GRCh38
rs752678513 Chr.4: 20707908-20707908 on Build GRCh38
rs769161605 Chr.4: 20707885-20707885 on Build GRCh38
rs1034265691 Chr.4: 20707951-20707951 on Build GRCh38
rs988347375 Chr.4: 20707950-20707950 on Build GRCh38
rs961741992 Chr.4: 20708001-20708001 on Build GRCh38
rs758288528 Chr.4: 20707911-20707911 on Build GRCh38
rs771089789 Chr.4: 20707857-20707857 on Build GRCh38
rs746769622 Chr.4: 20707849-20707849 on Build GRCh38
rs942670434 Chr.4: 20707879-20707879 on Build GRCh38
rs370331344 Chr.4: 20707842-20707842 on Build GRCh38
rs765956844 Chr.4: 20707887-20707887 on Build GRCh38
rs759110461 Chr.4: 20707901-20707901 on Build GRCh38
rs746003821 Chr.4: 20707865-20707865 on Build GRCh38
rs1024954997 Chr.4: 20707871-20707871 on Build GRCh38
rs201041637 Chr.4: 20707917-20707917 on Build GRCh38
rs757061889 Chr.4: 20707919-20707919 on Build GRCh38
rs958397367 Chr.4: 20707968-20707968 on Build GRCh38
rs769991983 Chr.4: 20707867-20707867 on Build GRCh38
rs1048500504 Chr.4: 20707988-20707988 on Build GRCh38
rs749792579 Chr.4: 20707883-20707883 on Build GRCh38
rs781418174 Chr.4: 20707861-20707861 on Build GRCh38
rs1005828245 Chr.4: 20707982-20707982 on Build GRCh38
rs762174523 Chr.4: 20707886-20707886 on Build GRCh38
rs553257564 Chr.4: 20707912-20707912 on Build GRCh38
rs888371970 Chr.4: 20707990-20707990 on Build GRCh38
rs777406505 Chr.4: 20707846-20707846 on Build GRCh38
rs943795670 Chr.4: 20707962-20707962 on Build GRCh38
rs776249279 Chr.4: 20707897-20707897 on Build GRCh38
rs568709719 Chr.4: 20707921-20707921 on Build GRCh38
rs4696965 Chr.4: 20707929-20707929 on Build GRCh38
rs754468843 Chr.4: 20707925-20707925 on Build GRCh38
rs972885697 Chr.4: 20708011-20708011 on Build GRCh38
rs181253053 Chr.4: 20707880-20707880 on Build GRCh38

View Full Product Details