Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119341433-119341670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF5
Entrez Gene ID
114902
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
CTRP5
UniGene
Hs.632102
Gene Chromosome Location
Chr.11: 119338934-119346673 on Build GRCh38
Transcripts
NM_015645.4 NM_031433.3

Gene Symbol
MFRP
Entrez Gene ID
83552
Gene Name
membrane frizzled-related protein
Gene Aliases
MCOP5, NNO2, RD6
UniGene
Hs.632102
Gene Chromosome Location
Chr.11: 119338934-119346673 on Build GRCh38
Transcripts
NM_015645.4 NM_031433.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415309 c.1619C>G p.S540C Chr.11: 119341669-119341669 on Build GRCh38
3791252 c.1714G>A p.D572N Chr.11: 119341574-119341574 on Build GRCh38
466454 c.1714G>T p.D572Y Chr.11: 119341574-119341574 on Build GRCh38
2149467 c.1637A>C p.E546A Chr.11: 119341651-119341651 on Build GRCh38
1214984 c.1625G>A p.C542Y Chr.11: 119341663-119341663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752071088 Chr.11: 119341487-119341487 on Build GRCh38
rs562519861 Chr.11: 119341488-119341488 on Build GRCh38
rs781100758 Chr.11: 119341606-119341606 on Build GRCh38
rs138370910 Chr.11: 119341653-119341653 on Build GRCh38
rs766376249 Chr.11: 119341622-119341623 on Build GRCh38
rs933941325 Chr.11: 119341591-119341591 on Build GRCh38
rs762547647 Chr.11: 119341642-119341642 on Build GRCh38
rs757296756 Chr.11: 119341569-119341569 on Build GRCh38
rs548437645 Chr.11: 119341515-119341515 on Build GRCh38
rs768531221 Chr.11: 119341513-119341513 on Build GRCh38
rs769523574 Chr.11: 119341520-119341520 on Build GRCh38
rs374604094 Chr.11: 119341482-119341482 on Build GRCh38
rs762577318 Chr.11: 119341534-119341534 on Build GRCh38
rs761534091 Chr.11: 119341516-119341516 on Build GRCh38
rs773902427 Chr.11: 119341651-119341651 on Build GRCh38
rs768762152 Chr.11: 119341581-119341581 on Build GRCh38
rs184075096 Chr.11: 119341503-119341503 on Build GRCh38
rs748020341 Chr.11: 119341631-119341631 on Build GRCh38
rs762892262 Chr.11: 119341610-119341610 on Build GRCh38
rs779984838 Chr.11: 119341504-119341504 on Build GRCh38
rs746892085 Chr.11: 119341579-119341579 on Build GRCh38
rs778116339 Chr.11: 119341558-119341558 on Build GRCh38
rs997493141 Chr.11: 119341477-119341477 on Build GRCh38
rs769313757 Chr.11: 119341637-119341637 on Build GRCh38
rs886047828 Chr.11: 119341539-119341539 on Build GRCh38
rs772859907 Chr.11: 119341641-119341641 on Build GRCh38
rs778890127 Chr.11: 119341576-119341576 on Build GRCh38
rs746947108 Chr.11: 119341509-119341509 on Build GRCh38
rs566842182 Chr.11: 119341517-119341517 on Build GRCh38
rs777685330 Chr.11: 119341501-119341501 on Build GRCh38
rs752068989 Chr.11: 119341537-119341537 on Build GRCh38
rs754021172 Chr.11: 119341564-119341564 on Build GRCh38
rs772590953 Chr.11: 119341529-119341529 on Build GRCh38
rs756325658 Chr.11: 119341553-119341553 on Build GRCh38
rs770430181 Chr.11: 119341649-119341649 on Build GRCh38
rs767848584 Chr.11: 119341543-119341543 on Build GRCh38
rs992632789 Chr.11: 119341580-119341580 on Build GRCh38
rs112157682 Chr.11: 119341502-119341502 on Build GRCh38
rs3814765 Chr.11: 119341536-119341536 on Build GRCh38
rs760062834 Chr.11: 119341538-119341538 on Build GRCh38
rs753103667 Chr.11: 119341547-119341547 on Build GRCh38
rs532493008 Chr.11: 119341469-119341469 on Build GRCh38
rs577487940 Chr.11: 119341622-119341622 on Build GRCh38
rs971356489 Chr.11: 119341481-119341481 on Build GRCh38

View Full Product Details