Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 40224957-40225230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRK2
Entrez Gene ID
120892
Gene Name
leucine rich repeat kinase 2
Gene Aliases
AURA17, DARDARIN, PARK8, RIPK7, ROCO2
UniGene
Hs.187636
Gene Chromosome Location
Chr.12: 40224895-40369285 on Build GRCh38
Transcripts
NM_198578.3 XM_005268629.3 XM_011537877.2 XM_011537881.2 XM_011537882.2 XM_017018786.1 XM_017018789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
187369 c.28G>A p.E10K Chr.12: 40225159-40225159 on Build GRCh38
7322542 c.46C>A p.L16M Chr.12: 40225177-40225177 on Build GRCh38
2065463 c.31G>T p.E11* Chr.12: 40225162-40225162 on Build GRCh38
6582327 c.25T>G p.C9G Chr.12: 40225156-40225156 on Build GRCh38
6246094 c.86G>T p.G29V Chr.12: 40225217-40225217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771312063 Chr.12: 40225146-40225146 on Build GRCh38
rs952314083 Chr.12: 40225059-40225059 on Build GRCh38
rs201321281 Chr.12: 40225045-40225045 on Build GRCh38
rs1036234153 Chr.12: 40224999-40224999 on Build GRCh38
rs199955454 Chr.12: 40225011-40225011 on Build GRCh38
rs41286470 Chr.12: 40225111-40225111 on Build GRCh38
rs200688492 Chr.12: 40225156-40225156 on Build GRCh38
rs767521458 Chr.12: 40225174-40225174 on Build GRCh38
rs767609591 Chr.12: 40225103-40225103 on Build GRCh38
rs1006894548 Chr.12: 40225027-40225027 on Build GRCh38
rs144990030 Chr.12: 40225100-40225100 on Build GRCh38
rs71449756 Chr.12: 40225117-40225117 on Build GRCh38
rs778295181 Chr.12: 40225136-40225136 on Build GRCh38
rs529078812 Chr.12: 40225060-40225060 on Build GRCh38
rs1011697978 Chr.12: 40225043-40225043 on Build GRCh38
rs778678298 Chr.12: 40225202-40225202 on Build GRCh38
rs904983879 Chr.12: 40224981-40224981 on Build GRCh38
rs765232448 Chr.12: 40225088-40225088 on Build GRCh38
rs910549475 Chr.12: 40225063-40225063 on Build GRCh38
rs71449755 Chr.12: 40225024-40225024 on Build GRCh38
rs1020433882 Chr.12: 40225047-40225047 on Build GRCh38
rs72546314 Chr.12: 40225102-40225102 on Build GRCh38
rs112643657 Chr.12: 40225008-40225008 on Build GRCh38
rs41286472 Chr.12: 40225079-40225079 on Build GRCh38
rs201065719 Chr.12: 40224990-40224990 on Build GRCh38
rs754575389 Chr.12: 40225198-40225198 on Build GRCh38
rs747754465 Chr.12: 40225126-40225126 on Build GRCh38
rs199909271 Chr.12: 40224993-40224993 on Build GRCh38
rs957511525 Chr.12: 40225041-40225041 on Build GRCh38
rs766282693 Chr.12: 40225185-40225185 on Build GRCh38
rs747338046 Chr.12: 40225142-40225142 on Build GRCh38
rs754445903 Chr.12: 40225112-40225112 on Build GRCh38
rs778283388 Chr.12: 40225113-40225113 on Build GRCh38
rs775407458 Chr.12: 40225170-40225170 on Build GRCh38
rs976169203 Chr.12: 40225087-40225087 on Build GRCh38
rs981632250 Chr.12: 40225153-40225153 on Build GRCh38
rs756004024 Chr.12: 40225107-40225107 on Build GRCh38
rs1027013700 Chr.12: 40225046-40225046 on Build GRCh38
rs142399623 Chr.12: 40225176-40225176 on Build GRCh38
rs908967057 Chr.12: 40224980-40224980 on Build GRCh38
rs1052256144 Chr.12: 40225154-40225154 on Build GRCh38
rs281865040 Chr.12: 40225159-40225159 on Build GRCh38
rs776873808 Chr.12: 40225084-40225084 on Build GRCh38
rs964674334 Chr.12: 40225071-40225071 on Build GRCh38
rs760489048 Chr.12: 40225182-40225182 on Build GRCh38
rs960876110 Chr.12: 40225035-40225035 on Build GRCh38
rs151283507 Chr.12: 40225197-40225197 on Build GRCh38
rs759598159 Chr.12: 40225086-40225086 on Build GRCh38
rs1388595 Chr.12: 40225001-40225001 on Build GRCh38
rs200678517 Chr.12: 40225061-40225061 on Build GRCh38
rs752886290 Chr.12: 40225098-40225098 on Build GRCh38
rs538494701 Chr.12: 40225037-40225037 on Build GRCh38
rs201002117 Chr.12: 40224997-40224997 on Build GRCh38
rs772438516 Chr.12: 40225130-40225130 on Build GRCh38
rs759670012 Chr.12: 40225160-40225160 on Build GRCh38
rs750211229 Chr.12: 40225105-40225105 on Build GRCh38
rs779804338 Chr.12: 40225109-40225109 on Build GRCh38
rs201899998 Chr.12: 40225000-40225000 on Build GRCh38

View Full Product Details