Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75097142-75097415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
UniGene
Hs.101302
Gene Chromosome Location
Chr.6: 75084326-75206262 on Build GRCh38
Transcripts
NM_004370.5 NM_080645.2 XM_011535434.1 XM_011535435.1 XM_011535436.2 XM_017010252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4356914 c.8567C>T p.P2856L Chr.6: 75097263-75097263 on Build GRCh38
5771276 c.8560G>C p.G2854R Chr.6: 75097270-75097270 on Build GRCh38
4006554 c.8565A>C p.P2855P Chr.6: 75097265-75097265 on Build GRCh38
3175093 c.8577G>T p.P2859P Chr.6: 75097253-75097253 on Build GRCh38
1081644 c.8577G>A p.P2859P Chr.6: 75097253-75097253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375797670 Chr.6: 75097218-75097218 on Build GRCh38
rs988202553 Chr.6: 75097393-75097393 on Build GRCh38
rs200375837 Chr.6: 75097276-75097276 on Build GRCh38
rs370191776 Chr.6: 75097260-75097260 on Build GRCh38
rs865873107 Chr.6: 75097257-75097265 on Build GRCh38
rs775776703 Chr.6: 75097239-75097239 on Build GRCh38
rs771432112 Chr.6: 75097356-75097356 on Build GRCh38
rs755782718 Chr.6: 75097309-75097309 on Build GRCh38
rs199505163 Chr.6: 75097288-75097288 on Build GRCh38
rs777596531 Chr.6: 75097312-75097312 on Build GRCh38
rs925242934 Chr.6: 75097342-75097342 on Build GRCh38
rs550955309 Chr.6: 75097374-75097374 on Build GRCh38
rs749595244 Chr.6: 75097351-75097351 on Build GRCh38
rs747817657 Chr.6: 75097300-75097300 on Build GRCh38
rs942767876 Chr.6: 75097337-75097337 on Build GRCh38
rs559357342 Chr.6: 75097268-75097268 on Build GRCh38
rs759599939 Chr.6: 75097219-75097219 on Build GRCh38
rs767547262 Chr.6: 75097223-75097223 on Build GRCh38
rs974657164 Chr.6: 75097250-75097250 on Build GRCh38
rs935433799 Chr.6: 75097387-75097388 on Build GRCh38
rs764992627 Chr.6: 75097257-75097257 on Build GRCh38
rs574147225 Chr.6: 75097263-75097263 on Build GRCh38
rs750933003 Chr.6: 75097309-75097310 on Build GRCh38
rs760925606 Chr.6: 75097245-75097245 on Build GRCh38
rs1016039503 Chr.6: 75097211-75097211 on Build GRCh38
rs267601121 Chr.6: 75097301-75097301 on Build GRCh38
rs369104842 Chr.6: 75097282-75097282 on Build GRCh38
rs202239067 Chr.6: 75097265-75097265 on Build GRCh38
rs562243182 Chr.6: 75097190-75097190 on Build GRCh38
rs544161815 Chr.6: 75097189-75097189 on Build GRCh38
rs975443675 Chr.6: 75097363-75097363 on Build GRCh38

View Full Product Details