Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75235299-75235536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAR1
Entrez Gene ID
9564
Gene Name
BCAR1, Cas family scaffolding protein
Gene Aliases
CAS, CAS1, CASS1, CRKAS, P130Cas
UniGene
Hs.479747
Gene Chromosome Location
Chr.16: 75229030-75268053 on Build GRCh38
Transcripts
NM_001170714.1 NM_001170715.1 NM_001170716.1 NM_001170717.1 NM_001170718.1 NM_001170719.1 NM_001170720.1 NM_001170721.1 NM_014567.3 XM_005256258.4 XM_005256259.4 XM_005256260.4 XM_011523475.2 XM_017023897.1 XM_017023898.1 XM_017023899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3680018 c.1573G>A p.A525T Chr.16: 75235326-75235326 on Build GRCh38
4890600 c.1592G>T p.G531V Chr.16: 75235445-75235445 on Build GRCh38
5981032 c.1507G>A p.G503S Chr.16: 75235446-75235446 on Build GRCh38
6610311 c.1581C>T p.A527A Chr.16: 75235372-75235372 on Build GRCh38
7101463 c.1738G>A p.D580N Chr.16: 75235299-75235299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746053585 Chr.16: 75235406-75235406 on Build GRCh38
rs754929679 Chr.16: 75235486-75235486 on Build GRCh38
rs371801925 Chr.16: 75235401-75235401 on Build GRCh38
rs775196763 Chr.16: 75235501-75235501 on Build GRCh38
rs767903911 Chr.16: 75235371-75235371 on Build GRCh38
rs200154768 Chr.16: 75235402-75235402 on Build GRCh38
rs758457850 Chr.16: 75235449-75235449 on Build GRCh38
rs16957559 Chr.16: 75235480-75235480 on Build GRCh38
rs531807345 Chr.16: 75235447-75235447 on Build GRCh38
rs753840727 Chr.16: 75235351-75235351 on Build GRCh38
rs753693100 Chr.16: 75235332-75235332 on Build GRCh38
rs200709299 Chr.16: 75235350-75235350 on Build GRCh38
rs764816297 Chr.16: 75235498-75235498 on Build GRCh38
rs755023555 Chr.16: 75235461-75235461 on Build GRCh38
rs776382344 Chr.16: 75235487-75235487 on Build GRCh38
rs767260706 Chr.16: 75235513-75235513 on Build GRCh38
rs919436260 Chr.16: 75235394-75235394 on Build GRCh38
rs575292263 Chr.16: 75235333-75235333 on Build GRCh38
rs1001160115 Chr.16: 75235476-75235476 on Build GRCh38
rs540154029 Chr.16: 75235414-75235414 on Build GRCh38
rs769085637 Chr.16: 75235366-75235366 on Build GRCh38
rs765985898 Chr.16: 75235428-75235428 on Build GRCh38
rs764036962 Chr.16: 75235504-75235504 on Build GRCh38
rs61763001 Chr.16: 75235342-75235342 on Build GRCh38
rs780145839 Chr.16: 75235413-75235413 on Build GRCh38
rs3206884 Chr.16: 75235420-75235420 on Build GRCh38
rs773594754 Chr.16: 75235483-75235483 on Build GRCh38
rs371179579 Chr.16: 75235419-75235419 on Build GRCh38
rs367866361 Chr.16: 75235391-75235391 on Build GRCh38
rs751304195 Chr.16: 75235454-75235454 on Build GRCh38
rs767099888 Chr.16: 75235319-75235319 on Build GRCh38
rs61738400 Chr.16: 75235357-75235357 on Build GRCh38
rs375275705 Chr.16: 75235378-75235378 on Build GRCh38
rs756426743 Chr.16: 75235412-75235412 on Build GRCh38
rs760427410 Chr.16: 75235380-75235380 on Build GRCh38
rs772700360 Chr.16: 75235481-75235481 on Build GRCh38
rs200175369 Chr.16: 75235505-75235505 on Build GRCh38
rs16957558 Chr.16: 75235427-75235427 on Build GRCh38
rs754535159 Chr.16: 75235399-75235399 on Build GRCh38
rs145073440 Chr.16: 75235423-75235423 on Build GRCh38
rs774802956 Chr.16: 75235421-75235421 on Build GRCh38
rs758226581 Chr.16: 75235403-75235403 on Build GRCh38
rs765789858 Chr.16: 75235390-75235390 on Build GRCh38
rs369072927 Chr.16: 75235324-75235324 on Build GRCh38
rs141024553 Chr.16: 75235442-75235442 on Build GRCh38
rs757015622 Chr.16: 75235455-75235455 on Build GRCh38
rs776157526 Chr.16: 75235441-75235441 on Build GRCh38
rs775699055 Chr.16: 75235359-75235359 on Build GRCh38
rs148628374 Chr.16: 75235325-75235325 on Build GRCh38
rs1009993556 Chr.16: 75235372-75235372 on Build GRCh38
rs117943983 Chr.16: 75235326-75235326 on Build GRCh38
rs746131932 Chr.16: 75235356-75235356 on Build GRCh38
rs971493325 Chr.16: 75235509-75235509 on Build GRCh38
rs761629396 Chr.16: 75235506-75235506 on Build GRCh38
rs764871357 Chr.16: 75235446-75235446 on Build GRCh38
rs150753098 Chr.16: 75235361-75235361 on Build GRCh38
rs759302886 Chr.16: 75235489-75235489 on Build GRCh38
rs749546698 Chr.16: 75235456-75235456 on Build GRCh38
rs763794116 Chr.16: 75235453-75235453 on Build GRCh38
rs3169330 Chr.16: 75235369-75235369 on Build GRCh38
rs754042658 Chr.16: 75235515-75235515 on Build GRCh38
rs1054274218 Chr.16: 75235503-75235503 on Build GRCh38
rs950206545 Chr.16: 75235389-75235389 on Build GRCh38
rs145822132 Chr.16: 75235485-75235485 on Build GRCh38
rs777661393 Chr.16: 75235404-75235404 on Build GRCh38
rs774853836 Chr.16: 75235368-75235368 on Build GRCh38

View Full Product Details