Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54142790-54143047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT3
Entrez Gene ID
4849
Gene Name
CCR4-NOT transcription complex subunit 3
Gene Aliases
LENG2, NOT3, NOT3H
UniGene
Hs.343571
Gene Chromosome Location
Chr.19: 54137689-54155708 on Build GRCh38
Transcripts
NM_014516.3 XM_005278279.1 XM_005278280.2 XM_005278281.1 XM_005278282.2 XM_011526992.1 XM_011526993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7209680 c.7G>C p.D3H Chr.19: 54142985-54142985 on Build GRCh38
4673190 c.5C>T p.A2V Chr.19: 54142983-54142983 on Build GRCh38
6999192 c.17A>C p.K6T Chr.19: 54142995-54142995 on Build GRCh38
3736980 c.13_14insG p.K6fs*5 Chr.19: 54142991-54142992 on Build GRCh38
4081186 c.14G>A p.R5H Chr.19: 54142992-54142992 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs904553955 Chr.19: 54142821-54142821 on Build GRCh38
rs376079665 Chr.19: 54143022-54143022 on Build GRCh38
rs139199651 Chr.19: 54142934-54142934 on Build GRCh38
rs150786079 Chr.19: 54142930-54142930 on Build GRCh38
rs751940189 Chr.19: 54143024-54143024 on Build GRCh38
rs1014443948 Chr.19: 54142881-54142881 on Build GRCh38
rs776663615 Chr.19: 54143002-54143002 on Build GRCh38
rs748792767 Chr.19: 54142929-54142935 on Build GRCh38
rs189411008 Chr.19: 54142960-54142960 on Build GRCh38
rs765336754 Chr.19: 54142929-54142929 on Build GRCh38
rs781666421 Chr.19: 54142943-54142943 on Build GRCh38
rs375303823 Chr.19: 54142895-54142895 on Build GRCh38
rs62144174 Chr.19: 54142874-54142874 on Build GRCh38
rs78784872 Chr.19: 54142867-54142867 on Build GRCh38
rs764312790 Chr.19: 54143019-54143019 on Build GRCh38
rs587659920 Chr.19: 54143010-54143010 on Build GRCh38
rs1053196012 Chr.19: 54142861-54142861 on Build GRCh38
rs934643287 Chr.19: 54142829-54142829 on Build GRCh38
rs763292898 Chr.19: 54143014-54143014 on Build GRCh38
rs1006252767 Chr.19: 54142938-54142938 on Build GRCh38
rs111692608 Chr.19: 54142920-54142920 on Build GRCh38
rs587763081 Chr.19: 54142870-54142870 on Build GRCh38
rs748702578 Chr.19: 54142955-54142955 on Build GRCh38
rs768778608 Chr.19: 54142976-54142976 on Build GRCh38
rs587739523 Chr.19: 54143029-54143029 on Build GRCh38
rs767519678 Chr.19: 54142831-54142831 on Build GRCh38
rs181011894 Chr.19: 54143007-54143007 on Build GRCh38
rs752046752 Chr.19: 54142940-54142940 on Build GRCh38
rs987219148 Chr.19: 54143011-54143011 on Build GRCh38
rs752832862 Chr.19: 54143013-54143013 on Build GRCh38
rs370627054 Chr.19: 54142958-54142958 on Build GRCh38
rs377359775 Chr.19: 54142961-54142961 on Build GRCh38
rs757702818 Chr.19: 54142942-54142942 on Build GRCh38

View Full Product Details