Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63698151-63698400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTEL1-TNFRSF6B
Entrez Gene ID
100533107
Gene Name
RTEL1-TNFRSF6B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.745057
Gene Chromosome Location
Chr.20: 63657810-63698698 on Build GRCh38
Transcripts
NR_037882.1 NM_003823.3

Gene Symbol
TNFRSF6B
Entrez Gene ID
8771
Gene Name
TNF receptor superfamily member 6b
Gene Aliases
DCR3, DJ583P15.1.1, M68, M68E, TR6
UniGene
Hs.434878
Gene Chromosome Location
Chr.20: 63696651-63698698 on Build GRCh38
Transcripts
NR_037882.1 NM_003823.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7297231 c.726G>C p.W242C Chr.20: 63698386-63698386 on Build GRCh38
1240469 c.699G>A p.L233L Chr.20: 63698359-63698359 on Build GRCh38
347196 c.639T>C p.R213R Chr.20: 63698299-63698299 on Build GRCh38
364559 c.620G>C p.G207A Chr.20: 63698280-63698280 on Build GRCh38
6805082 c.708C>T p.L236L Chr.20: 63698368-63698368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001842434 Chr.20: 63698197-63698197 on Build GRCh38
rs780798948 Chr.20: 63698311-63698311 on Build GRCh38
rs979568659 Chr.20: 63698227-63698227 on Build GRCh38
rs113115338 Chr.20: 63698226-63698226 on Build GRCh38
rs1035079072 Chr.20: 63698209-63698209 on Build GRCh38
rs770991648 Chr.20: 63698318-63698318 on Build GRCh38
rs754782569 Chr.20: 63698309-63698309 on Build GRCh38
rs776796431 Chr.20: 63698335-63698335 on Build GRCh38
rs147543198 Chr.20: 63698352-63698352 on Build GRCh38
rs905088525 Chr.20: 63698191-63698191 on Build GRCh38
rs768990640 Chr.20: 63698382-63698382 on Build GRCh38
rs1048131800 Chr.20: 63698184-63698184 on Build GRCh38
rs761928512 Chr.20: 63698356-63698380 on Build GRCh38
rs1020199316 Chr.20: 63698328-63698328 on Build GRCh38
rs143300172 Chr.20: 63698351-63698351 on Build GRCh38
rs752376808 Chr.20: 63698364-63698364 on Build GRCh38
rs959995461 Chr.20: 63698195-63698195 on Build GRCh38
rs746787861 Chr.20: 63698254-63698254 on Build GRCh38
rs754828981 Chr.20: 63698360-63698360 on Build GRCh38
rs1034041926 Chr.20: 63698194-63698194 on Build GRCh38
rs150296740 Chr.20: 63698308-63698308 on Build GRCh38
rs576754898 Chr.20: 63698252-63698252 on Build GRCh38
rs780071498 Chr.20: 63698376-63698376 on Build GRCh38
rs772115276 Chr.20: 63698373-63698373 on Build GRCh38
rs764972813 Chr.20: 63698338-63698338 on Build GRCh38
rs201139619 Chr.20: 63698303-63698303 on Build GRCh38
rs771206685 Chr.20: 63698272-63698272 on Build GRCh38
rs748004646 Chr.20: 63698314-63698314 on Build GRCh38
rs377550212 Chr.20: 63698189-63698189 on Build GRCh38
rs960514816 Chr.20: 63698211-63698211 on Build GRCh38
rs923508643 Chr.20: 63698381-63698381 on Build GRCh38
rs762255968 Chr.20: 63698336-63698336 on Build GRCh38
rs764950396 Chr.20: 63698298-63698298 on Build GRCh38
rs761132943 Chr.20: 63698329-63698329 on Build GRCh38
rs944717583 Chr.20: 63698236-63698236 on Build GRCh38
rs745405361 Chr.20: 63698235-63698235 on Build GRCh38
rs375903710 Chr.20: 63698379-63698379 on Build GRCh38
rs941209719 Chr.20: 63698290-63698290 on Build GRCh38
rs199875804 Chr.20: 63698256-63698256 on Build GRCh38
rs774575588 Chr.20: 63698304-63698306 on Build GRCh38
rs772506716 Chr.20: 63698283-63698283 on Build GRCh38
rs559139845 Chr.20: 63698257-63698257 on Build GRCh38
rs1016814428 Chr.20: 63698190-63698190 on Build GRCh38
rs745930765 Chr.20: 63698323-63698323 on Build GRCh38
rs769239517 Chr.20: 63698330-63698330 on Build GRCh38
rs776031713 Chr.20: 63698291-63698291 on Build GRCh38
rs967199084 Chr.20: 63698208-63698208 on Build GRCh38
rs548373058 Chr.20: 63698368-63698368 on Build GRCh38
rs759615436 Chr.20: 63698282-63698282 on Build GRCh38
rs781192378 Chr.20: 63698361-63698361 on Build GRCh38
rs779100044 Chr.20: 63698369-63698369 on Build GRCh38
rs756756944 Chr.20: 63698229-63698229 on Build GRCh38
rs775941814 Chr.20: 63698327-63698327 on Build GRCh38
rs747119927 Chr.20: 63698377-63698377 on Build GRCh38
rs749898916 Chr.20: 63698300-63698300 on Build GRCh38
rs555561862 Chr.20: 63698196-63698196 on Build GRCh38
rs763392579 Chr.20: 63698242-63698242 on Build GRCh38
rs750168237 Chr.20: 63698346-63698346 on Build GRCh38
rs886940476 Chr.20: 63698174-63698174 on Build GRCh38
rs943584621 Chr.20: 63698237-63698237 on Build GRCh38
rs772203190 Chr.20: 63698326-63698326 on Build GRCh38
rs1020665536 Chr.20: 63698214-63698214 on Build GRCh38
rs778240359 Chr.20: 63698233-63698233 on Build GRCh38
rs746126394 Chr.20: 63698371-63698371 on Build GRCh38
rs575499250 Chr.20: 63698212-63698212 on Build GRCh38
rs137950052 Chr.20: 63698319-63698319 on Build GRCh38
rs754254086 Chr.20: 63698297-63698297 on Build GRCh38
rs764404347 Chr.20: 63698294-63698294 on Build GRCh38
rs749523185 Chr.20: 63698262-63698262 on Build GRCh38
rs112186341 Chr.20: 63698302-63698302 on Build GRCh38
rs369505171 Chr.20: 63698251-63698251 on Build GRCh38
rs544481762 Chr.20: 63698213-63698213 on Build GRCh38
rs751138050 Chr.20: 63698353-63698353 on Build GRCh38
rs769164134 Chr.20: 63698290-63698291 on Build GRCh38
rs1017774705 Chr.20: 63698363-63698363 on Build GRCh38

View Full Product Details