Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4340837-4341078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO7-PAM16
Entrez Gene ID
100529144
Gene Name
CORO7-PAM16 readthrough
Gene Aliases
-
UniGene
Hs.437957
Gene Chromosome Location
Chr.16: 4340251-4416961 on Build GRCh38
Transcripts
NM_001201479.1 NM_016069.9

Gene Symbol
PAM16
Entrez Gene ID
51025
Gene Name
presequence translocase-associated motor 16 homolog (S. cerevisiae)
Gene Aliases
CGI-136, MAGMAS, SMDMDM, TIM16, TIMM16
UniGene
Hs.730693
Gene Chromosome Location
Chr.16: 4340251-4351372 on Build GRCh38
Transcripts
NM_001201479.1 NM_016069.9

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6021404 c.226-1G>A p.? Chr.16: 4340986-4340986 on Build GRCh38
2920352 c.262G>A p.V88M Chr.16: 4340949-4340949 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761297017 Chr.16: 4340994-4340995 on Build GRCh38
rs201929603 Chr.16: 4340891-4340891 on Build GRCh38
rs745798161 Chr.16: 4340975-4340975 on Build GRCh38
rs530133377 Chr.16: 4340981-4340981 on Build GRCh38
rs769076825 Chr.16: 4341013-4341014 on Build GRCh38
rs376875983 Chr.16: 4340942-4340942 on Build GRCh38
rs113338335 Chr.16: 4340895-4340895 on Build GRCh38
rs147028858 Chr.16: 4341040-4341040 on Build GRCh38
rs776838285 Chr.16: 4341001-4341001 on Build GRCh38
rs543939113 Chr.16: 4340988-4340988 on Build GRCh38
rs748842835 Chr.16: 4340949-4340949 on Build GRCh38
rs374565033 Chr.16: 4340950-4340950 on Build GRCh38
rs753337345 Chr.16: 4341036-4341036 on Build GRCh38
rs747753092 Chr.16: 4340916-4340916 on Build GRCh38
rs780424910 Chr.16: 4340883-4340883 on Build GRCh38
rs2072151 Chr.16: 4341039-4341039 on Build GRCh38
rs936317312 Chr.16: 4341029-4341029 on Build GRCh38
rs750750280 Chr.16: 4340898-4340898 on Build GRCh38
rs764462507 Chr.16: 4340994-4340994 on Build GRCh38
rs770183989 Chr.16: 4341010-4341010 on Build GRCh38
rs775854504 Chr.16: 4341037-4341037 on Build GRCh38
rs770892963 Chr.16: 4340869-4340869 on Build GRCh38
rs377372488 Chr.16: 4341018-4341018 on Build GRCh38
rs776092733 Chr.16: 4340945-4340945 on Build GRCh38
rs771842302 Chr.16: 4340980-4340980 on Build GRCh38
rs767628886 Chr.16: 4340875-4340875 on Build GRCh38
rs755797690 Chr.16: 4340928-4340928 on Build GRCh38
rs777231583 Chr.16: 4340938-4340938 on Build GRCh38
rs966632132 Chr.16: 4340860-4340860 on Build GRCh38
rs762051507 Chr.16: 4341003-4341003 on Build GRCh38
rs751962838 Chr.16: 4341025-4341025 on Build GRCh38
rs765236526 Chr.16: 4341004-4341004 on Build GRCh38
rs995312938 Chr.16: 4340969-4340969 on Build GRCh38
rs116657149 Chr.16: 4340910-4340910 on Build GRCh38
rs786203989 Chr.16: 4340985-4340985 on Build GRCh38
rs377245232 Chr.16: 4340864-4340864 on Build GRCh38
rs552312159 Chr.16: 4341060-4341060 on Build GRCh38
rs765832103 Chr.16: 4340896-4340896 on Build GRCh38
rs760990351 Chr.16: 4340881-4340881 on Build GRCh38
rs777456079 Chr.16: 4341031-4341031 on Build GRCh38
rs142976385 Chr.16: 4340913-4340913 on Build GRCh38
rs759428567 Chr.16: 4340874-4340874 on Build GRCh38
rs370436559 Chr.16: 4341027-4341027 on Build GRCh38
rs772095473 Chr.16: 4340911-4340912 on Build GRCh38
rs775713557 Chr.16: 4340876-4340876 on Build GRCh38
rs73507245 Chr.16: 4340872-4340872 on Build GRCh38
rs755738556 Chr.16: 4341030-4341030 on Build GRCh38
rs984797070 Chr.16: 4340912-4340912 on Build GRCh38

View Full Product Details