Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122940072-122940321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB9
Entrez Gene ID
23457
Gene Name
ATP binding cassette subfamily B member 9
Gene Aliases
EST122234, TAPL
UniGene
Hs.511951
Gene Chromosome Location
Chr.12: 122918708-122975160 on Build GRCh38
Transcripts
NM_001243013.1 NM_001243014.1 NM_019624.3 NM_019625.3 NM_203444.3 XM_005253558.2 XM_011538095.2 XM_011538096.2 XM_011538098.2 XM_011538099.2 XM_017019103.1 XM_017019104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3792298 c.1635G>A p.K545K Chr.12: 122940219-122940219 on Build GRCh38
6430249 c.1708A>T p.I570F Chr.12: 122940146-122940146 on Build GRCh38
3384113 c.1625G>A p.G542D Chr.12: 122940229-122940229 on Build GRCh38
5374479 c.1670C>A p.P557H Chr.12: 122940184-122940184 on Build GRCh38
7190874 c.1651A>G p.I551V Chr.12: 122940203-122940203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750183772 Chr.12: 122940173-122940173 on Build GRCh38
rs772326323 Chr.12: 122940187-122940187 on Build GRCh38
rs768098199 Chr.12: 122940216-122940216 on Build GRCh38
rs867276986 Chr.12: 122940154-122940154 on Build GRCh38
rs368501864 Chr.12: 122940264-122940264 on Build GRCh38
rs758704634 Chr.12: 122940267-122940267 on Build GRCh38
rs776586667 Chr.12: 122940237-122940253 on Build GRCh38
rs774679651 Chr.12: 122940115-122940115 on Build GRCh38
rs756155243 Chr.12: 122940098-122940098 on Build GRCh38
rs140303524 Chr.12: 122940262-122940262 on Build GRCh38
rs780352581 Chr.12: 122940100-122940100 on Build GRCh38
rs181063039 Chr.12: 122940183-122940183 on Build GRCh38
rs769044559 Chr.12: 122940104-122940104 on Build GRCh38
rs767066912 Chr.12: 122940243-122940243 on Build GRCh38
rs557925509 Chr.12: 122940197-122940197 on Build GRCh38
rs115844940 Chr.12: 122940218-122940218 on Build GRCh38
rs369328306 Chr.12: 122940135-122940135 on Build GRCh38
rs779022972 Chr.12: 122940185-122940185 on Build GRCh38
rs755065420 Chr.12: 122940184-122940184 on Build GRCh38
rs186347857 Chr.12: 122940246-122940246 on Build GRCh38
rs747405658 Chr.12: 122940284-122940284 on Build GRCh38
rs775039652 Chr.12: 122940146-122940146 on Build GRCh38
rs116742318 Chr.12: 122940202-122940202 on Build GRCh38
rs371490151 Chr.12: 122940194-122940194 on Build GRCh38
rs1020345009 Chr.12: 122940170-122940170 on Build GRCh38
rs544019943 Chr.12: 122940231-122940231 on Build GRCh38
rs753777058 Chr.12: 122940261-122940261 on Build GRCh38
rs556170242 Chr.12: 122940101-122940101 on Build GRCh38
rs761175587 Chr.12: 122940140-122940140 on Build GRCh38
rs1004903252 Chr.12: 122940229-122940229 on Build GRCh38
rs777082505 Chr.12: 122940196-122940196 on Build GRCh38
rs116208583 Chr.12: 122940156-122940156 on Build GRCh38
rs1029410200 Chr.12: 122940270-122940270 on Build GRCh38
rs376511047 Chr.12: 122940110-122940110 on Build GRCh38
rs756182750 Chr.12: 122940176-122940176 on Build GRCh38
rs568373792 Chr.12: 122940134-122940134 on Build GRCh38
rs116062676 Chr.12: 122940277-122940277 on Build GRCh38
rs564444198 Chr.12: 122940260-122940260 on Build GRCh38
rs768309979 Chr.12: 122940104-122940110 on Build GRCh38
rs771236759 Chr.12: 122940296-122940296 on Build GRCh38
rs893169215 Chr.12: 122940159-122940159 on Build GRCh38
rs745764051 Chr.12: 122940094-122940094 on Build GRCh38
rs553768721 Chr.12: 122940141-122940141 on Build GRCh38
rs747320759 Chr.12: 122940193-122940193 on Build GRCh38
rs748341612 Chr.12: 122940186-122940186 on Build GRCh38
rs916807343 Chr.12: 122940144-122940144 on Build GRCh38
rs374426615 Chr.12: 122940249-122940249 on Build GRCh38
rs748489928 Chr.12: 122940118-122940118 on Build GRCh38
rs775308654 Chr.12: 122940169-122940169 on Build GRCh38

View Full Product Details