Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26916145-26916384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP4-AS1
Entrez Gene ID
147429
Gene Name
AQP4 antisense RNA 1
Gene Aliases
C18orf16, CHST9-AS1
UniGene
Hs.559280
Gene Chromosome Location
Chr.18: 26865308-26935946 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_005258362.3 XM_017026033.1 XM_017026034.1

Gene Symbol
CHST9
Entrez Gene ID
83539
Gene Name
carbohydrate sulfotransferase 9
Gene Aliases
GALNAC4ST-2, GalNAc4ST2
UniGene
Hs.44584
Gene Chromosome Location
Chr.18: 26908754-27185364 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_005258362.3 XM_017026033.1 XM_017026034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1303645 c.1024G>A p.D342N Chr.18: 26916312-26916312 on Build GRCh38
473711 c.1222C>G p.Q408E Chr.18: 26916369-26916369 on Build GRCh38
6900647 c.1245G>A p.K415K Chr.18: 26916346-26916346 on Build GRCh38
386872 c.1228G>T p.V410L Chr.18: 26916363-26916363 on Build GRCh38
473709 c.1226T>C p.V409A Chr.18: 26916365-26916365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764070699 Chr.18: 26916316-26916316 on Build GRCh38
rs201405526 Chr.18: 26916295-26916295 on Build GRCh38
rs752832621 Chr.18: 26916297-26916297 on Build GRCh38
rs751601546 Chr.18: 26916321-26916321 on Build GRCh38
rs753119683 Chr.18: 26916280-26916286 on Build GRCh38
rs754740628 Chr.18: 26916356-26916356 on Build GRCh38
rs777620473 Chr.18: 26916263-26916263 on Build GRCh38
rs777312052 Chr.18: 26916274-26916274 on Build GRCh38
rs568388273 Chr.18: 26916232-26916232 on Build GRCh38
rs534240641 Chr.18: 26916272-26916272 on Build GRCh38
rs747751233 Chr.18: 26916359-26916359 on Build GRCh38
rs1009079071 Chr.18: 26916185-26916185 on Build GRCh38
rs758764758 Chr.18: 26916317-26916320 on Build GRCh38
rs373838914 Chr.18: 26916346-26916346 on Build GRCh38
rs185798102 Chr.18: 26916270-26916270 on Build GRCh38
rs778719992 Chr.18: 26916358-26916358 on Build GRCh38
rs746718413 Chr.18: 26916320-26916320 on Build GRCh38
rs773941111 Chr.18: 26916238-26916238 on Build GRCh38
rs771451674 Chr.18: 26916256-26916256 on Build GRCh38
rs1006562540 Chr.18: 26916173-26916173 on Build GRCh38
rs749162270 Chr.18: 26916300-26916300 on Build GRCh38
rs749242498 Chr.18: 26916353-26916353 on Build GRCh38
rs747879318 Chr.18: 26916243-26916243 on Build GRCh38
rs759933019 Chr.18: 26916281-26916281 on Build GRCh38
rs757330342 Chr.18: 26916333-26916333 on Build GRCh38
rs905692489 Chr.18: 26916228-26916228 on Build GRCh38
rs772845513 Chr.18: 26916269-26916269 on Build GRCh38
rs181527568 Chr.18: 26916198-26916198 on Build GRCh38
rs370973380 Chr.18: 26916234-26916234 on Build GRCh38
rs548529042 Chr.18: 26916236-26916236 on Build GRCh38
rs765470557 Chr.18: 26916222-26916224 on Build GRCh38
rs779983593 Chr.18: 26916221-26916221 on Build GRCh38

View Full Product Details