Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 31578197-31578466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC129
Entrez Gene ID
223075
Gene Name
coiled-coil domain containing 129
Gene Aliases
-
UniGene
Hs.224269
Gene Chromosome Location
Chr.7: 31514071-31658720 on Build GRCh38
Transcripts
NM_001257967.1 NM_001257968.1 NM_194300.3 NR_047565.1 XM_011515213.1 XM_017011871.1 XM_017011872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6356307 c.1143G>T p.G381G Chr.7: 31578329-31578329 on Build GRCh38
5561983 c.1028C>A p.P343Q Chr.7: 31578292-31578292 on Build GRCh38
3880604 c.1030G>A p.A344T Chr.7: 31578294-31578294 on Build GRCh38
336273 c.1005G>T p.K335N Chr.7: 31578269-31578269 on Build GRCh38
4004456 c.1029G>T p.P343P Chr.7: 31578293-31578293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867645901 Chr.7: 31578291-31578291 on Build GRCh38
rs747089263 Chr.7: 31578367-31578367 on Build GRCh38
rs200119465 Chr.7: 31578230-31578230 on Build GRCh38
rs758300969 Chr.7: 31578306-31578306 on Build GRCh38
rs760836286 Chr.7: 31578279-31578279 on Build GRCh38
rs764047248 Chr.7: 31578399-31578399 on Build GRCh38
rs373377448 Chr.7: 31578437-31578437 on Build GRCh38
rs780320062 Chr.7: 31578278-31578278 on Build GRCh38
rs752561168 Chr.7: 31578303-31578303 on Build GRCh38
rs375575625 Chr.7: 31578337-31578337 on Build GRCh38
rs780431959 Chr.7: 31578229-31578229 on Build GRCh38
rs573493648 Chr.7: 31578391-31578391 on Build GRCh38
rs778729726 Chr.7: 31578238-31578238 on Build GRCh38
rs1004054813 Chr.7: 31578349-31578354 on Build GRCh38
rs776985269 Chr.7: 31578379-31578379 on Build GRCh38
rs776617154 Chr.7: 31578293-31578293 on Build GRCh38
rs372420153 Chr.7: 31578292-31578292 on Build GRCh38
rs942902924 Chr.7: 31578385-31578385 on Build GRCh38
rs10252720 Chr.7: 31578394-31578394 on Build GRCh38
rs369963496 Chr.7: 31578339-31578339 on Build GRCh38
rs868170414 Chr.7: 31578313-31578313 on Build GRCh38
rs565388006 Chr.7: 31578435-31578435 on Build GRCh38
rs748329275 Chr.7: 31578269-31578269 on Build GRCh38
rs981922439 Chr.7: 31578241-31578241 on Build GRCh38
rs972779944 Chr.7: 31578304-31578304 on Build GRCh38
rs756869391 Chr.7: 31578222-31578222 on Build GRCh38
rs779456068 Chr.7: 31578347-31578347 on Build GRCh38
rs778563629 Chr.7: 31578419-31578419 on Build GRCh38
rs769202065 Chr.7: 31578257-31578257 on Build GRCh38
rs1016728324 Chr.7: 31578377-31578377 on Build GRCh38
rs751044745 Chr.7: 31578320-31578320 on Build GRCh38
rs753745587 Chr.7: 31578293-31578293 on Build GRCh38
rs759370327 Chr.7: 31578300-31578300 on Build GRCh38
rs771315477 Chr.7: 31578370-31578370 on Build GRCh38
rs772590110 Chr.7: 31578354-31578354 on Build GRCh38
rs923292314 Chr.7: 31578300-31578301 on Build GRCh38
rs765135134 Chr.7: 31578393-31578393 on Build GRCh38
rs748606184 Chr.7: 31578349-31578349 on Build GRCh38
rs1048353024 Chr.7: 31578369-31578369 on Build GRCh38
rs368760577 Chr.7: 31578416-31578416 on Build GRCh38
rs756729592 Chr.7: 31578414-31578414 on Build GRCh38
rs192536493 Chr.7: 31578328-31578328 on Build GRCh38
rs767224679 Chr.7: 31578420-31578420 on Build GRCh38
rs368169623 Chr.7: 31578267-31578267 on Build GRCh38
rs750063873 Chr.7: 31578432-31578432 on Build GRCh38
rs372373590 Chr.7: 31578356-31578356 on Build GRCh38
rs890044847 Chr.7: 31578371-31578371 on Build GRCh38
rs927776979 Chr.7: 31578246-31578246 on Build GRCh38
rs751084099 Chr.7: 31578403-31578403 on Build GRCh38
rs773187330 Chr.7: 31578366-31578366 on Build GRCh38
rs772033986 Chr.7: 31578276-31578276 on Build GRCh38
rs561225420 Chr.7: 31578329-31578329 on Build GRCh38

View Full Product Details