Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143330681-143330954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN1
Entrez Gene ID
1180
Gene Name
chloride voltage-gated channel 1
Gene Aliases
CLC1
UniGene
Hs.121483
Gene Chromosome Location
Chr.7: 143316126-143352008 on Build GRCh38
Transcripts
NM_000083.2 NR_046453.1 XM_017011739.1 XM_017011740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3258062 c.898C>T p.R300W Chr.7: 143330816-143330816 on Build GRCh38
4672615 c.930G>A p.T310T Chr.7: 143330848-143330848 on Build GRCh38
3635183 c.899G>A p.R300Q Chr.7: 143330817-143330817 on Build GRCh38
6109326 c.896T>G p.V299G Chr.7: 143330814-143330814 on Build GRCh38
3635182 c.854G>A p.G285E Chr.7: 143330772-143330772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149578972 Chr.7: 143330818-143330818 on Build GRCh38
rs775703359 Chr.7: 143330792-143330792 on Build GRCh38
rs80356701 Chr.7: 143330838-143330838 on Build GRCh38
rs770854110 Chr.7: 143330931-143330931 on Build GRCh38
rs202179484 Chr.7: 143330813-143330813 on Build GRCh38
rs80356689 Chr.7: 143330775-143330775 on Build GRCh38
rs760667989 Chr.7: 143330909-143330909 on Build GRCh38
rs113154958 Chr.7: 143330765-143330765 on Build GRCh38
rs766109574 Chr.7: 143330743-143330743 on Build GRCh38
rs201115855 Chr.7: 143330764-143330764 on Build GRCh38
rs201738127 Chr.7: 143330821-143330821 on Build GRCh38
rs375563386 Chr.7: 143330794-143330794 on Build GRCh38
rs202218267 Chr.7: 143330791-143330791 on Build GRCh38
rs201043261 Chr.7: 143330889-143330889 on Build GRCh38
rs982703615 Chr.7: 143330853-143330853 on Build GRCh38
rs774396430 Chr.7: 143330897-143330897 on Build GRCh38
rs975612923 Chr.7: 143330895-143330902 on Build GRCh38
rs780150093 Chr.7: 143330880-143330880 on Build GRCh38
rs766116662 Chr.7: 143330837-143330848 on Build GRCh38
rs368254261 Chr.7: 143330746-143330746 on Build GRCh38
rs747534917 Chr.7: 143330929-143330929 on Build GRCh38
rs771454498 Chr.7: 143330779-143330779 on Build GRCh38
rs773151679 Chr.7: 143330815-143330815 on Build GRCh38
rs777104046 Chr.7: 143330782-143330782 on Build GRCh38
rs368280521 Chr.7: 143330816-143330816 on Build GRCh38
rs748396910 Chr.7: 143330742-143330743 on Build GRCh38
rs750290712 Chr.7: 143330737-143330737 on Build GRCh38
rs767348977 Chr.7: 143330724-143330724 on Build GRCh38
rs764100025 Chr.7: 143330810-143330810 on Build GRCh38
rs752153471 Chr.7: 143330854-143330854 on Build GRCh38
rs757871092 Chr.7: 143330848-143330848 on Build GRCh38
rs150885084 Chr.7: 143330772-143330772 on Build GRCh38
rs370524986 Chr.7: 143330763-143330763 on Build GRCh38
rs377300277 Chr.7: 143330902-143330902 on Build GRCh38
rs761866474 Chr.7: 143330722-143330722 on Build GRCh38
rs80356691 Chr.7: 143330847-143330847 on Build GRCh38
rs764804597 Chr.7: 143330830-143330830 on Build GRCh38
rs80356692 Chr.7: 143330855-143330855 on Build GRCh38
rs80356702 Chr.7: 143330868-143330868 on Build GRCh38
rs781728156 Chr.7: 143330871-143330871 on Build GRCh38
rs749473626 Chr.7: 143330881-143330881 on Build GRCh38
rs768750688 Chr.7: 143330885-143330885 on Build GRCh38
rs893313179 Chr.7: 143330723-143330723 on Build GRCh38
rs755912761 Chr.7: 143330739-143330739 on Build GRCh38
rs121912805 Chr.7: 143330789-143330789 on Build GRCh38
rs746212684 Chr.7: 143330875-143330875 on Build GRCh38
rs80356690 Chr.7: 143330788-143330788 on Build GRCh38
rs951855372 Chr.7: 143330888-143330888 on Build GRCh38
rs1003273679 Chr.7: 143330768-143330768 on Build GRCh38
rs753533377 Chr.7: 143330745-143330745 on Build GRCh38
rs762943928 Chr.7: 143330796-143330796 on Build GRCh38
rs202177584 Chr.7: 143330769-143330769 on Build GRCh38
rs747609097 Chr.7: 143330749-143330749 on Build GRCh38
rs777378229 Chr.7: 143330851-143330851 on Build GRCh38
rs118066140 Chr.7: 143330817-143330817 on Build GRCh38

View Full Product Details