Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48342427-48342662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM143
Entrez Gene ID
55260
Gene Name
transmembrane protein 143
Gene Aliases
-
UniGene
Hs.351335
Gene Chromosome Location
Chr.19: 48332356-48364237 on Build GRCh38
Transcripts
NM_001303538.1 NM_001303539.1 NM_001303540.1 NM_018273.3 NR_130317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7216325 c.645+10C>T p.? Chr.19: 48342520-48342520 on Build GRCh38
5985033 c.513C>T p.L171L Chr.19: 48342662-48342662 on Build GRCh38
3536480 c.861C>T p.V287V Chr.19: 48342644-48342644 on Build GRCh38
4857395 c.596C>A p.S199Y Chr.19: 48342579-48342579 on Build GRCh38
474982 c.926C>A p.S309Y Chr.19: 48342579-48342579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781318508 Chr.19: 48342638-48342638 on Build GRCh38
rs563890773 Chr.19: 48342463-48342463 on Build GRCh38
rs983644811 Chr.19: 48342461-48342462 on Build GRCh38
rs141230407 Chr.19: 48342581-48342581 on Build GRCh38
rs757828183 Chr.19: 48342599-48342599 on Build GRCh38
rs748377336 Chr.19: 48342593-48342593 on Build GRCh38
rs4275952 Chr.19: 48342520-48342520 on Build GRCh38
rs372725525 Chr.19: 48342535-48342535 on Build GRCh38
rs371776234 Chr.19: 48342599-48342599 on Build GRCh38
rs758919637 Chr.19: 48342494-48342494 on Build GRCh38
rs909342383 Chr.19: 48342468-48342469 on Build GRCh38
rs147401463 Chr.19: 48342499-48342499 on Build GRCh38
rs543994814 Chr.19: 48342447-48342447 on Build GRCh38
rs781227617 Chr.19: 48342592-48342592 on Build GRCh38
rs368208555 Chr.19: 48342559-48342559 on Build GRCh38
rs182966733 Chr.19: 48342574-48342574 on Build GRCh38
rs1025362475 Chr.19: 48342634-48342634 on Build GRCh38
rs752237474 Chr.19: 48342620-48342620 on Build GRCh38
rs375809150 Chr.19: 48342512-48342512 on Build GRCh38
rs200364296 Chr.19: 48342536-48342536 on Build GRCh38
rs778652459 Chr.19: 48342627-48342627 on Build GRCh38
rs574511147 Chr.19: 48342640-48342640 on Build GRCh38
rs760088406 Chr.19: 48342615-48342615 on Build GRCh38
rs548897789 Chr.19: 48342641-48342641 on Build GRCh38
rs909374602 Chr.19: 48342493-48342493 on Build GRCh38
rs777482957 Chr.19: 48342564-48342564 on Build GRCh38
rs895232661 Chr.19: 48342619-48342619 on Build GRCh38
rs781456638 Chr.19: 48342547-48342547 on Build GRCh38
rs763733273 Chr.19: 48342623-48342623 on Build GRCh38
rs757158559 Chr.19: 48342572-48342572 on Build GRCh38
rs891054856 Chr.19: 48342573-48342573 on Build GRCh38
rs767182009 Chr.19: 48342617-48342617 on Build GRCh38
rs768766993 Chr.19: 48342582-48342582 on Build GRCh38
rs752435541 Chr.19: 48342562-48342562 on Build GRCh38
rs763437652 Chr.19: 48342544-48342544 on Build GRCh38
rs767308742 Chr.19: 48342560-48342560 on Build GRCh38
rs778599096 Chr.19: 48342563-48342564 on Build GRCh38
rs1044628360 Chr.19: 48342502-48342502 on Build GRCh38
rs753600045 Chr.19: 48342625-48342625 on Build GRCh38
rs138171230 Chr.19: 48342569-48342569 on Build GRCh38
rs776797547 Chr.19: 48342530-48342530 on Build GRCh38
rs553369848 Chr.19: 48342633-48342633 on Build GRCh38
rs541280160 Chr.19: 48342621-48342621 on Build GRCh38
rs754682054 Chr.19: 48342510-48342510 on Build GRCh38
rs145038055 Chr.19: 48342598-48342598 on Build GRCh38
rs983240816 Chr.19: 48342484-48342484 on Build GRCh38
rs771585590 Chr.19: 48342605-48342605 on Build GRCh38
rs774959405 Chr.19: 48342608-48342608 on Build GRCh38
rs201478905 Chr.19: 48342586-48342586 on Build GRCh38
rs751111250 Chr.19: 48342501-48342501 on Build GRCh38
rs773613522 Chr.19: 48342541-48342541 on Build GRCh38
rs755825011 Chr.19: 48342513-48342513 on Build GRCh38
rs929054837 Chr.19: 48342480-48342480 on Build GRCh38
rs372908272 Chr.19: 48342511-48342511 on Build GRCh38
rs747702752 Chr.19: 48342564-48342566 on Build GRCh38
rs777589978 Chr.19: 48342519-48342519 on Build GRCh38
rs371521776 Chr.19: 48342563-48342563 on Build GRCh38
rs747037503 Chr.19: 48342525-48342525 on Build GRCh38
rs148331223 Chr.19: 48342637-48342637 on Build GRCh38
rs757039002 Chr.19: 48342626-48342626 on Build GRCh38

View Full Product Details