Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40646828-40647079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf38
Entrez Gene ID
159013
Gene Name
chromosome X open reading frame 38
Gene Aliases
-
UniGene
Hs.495961
Gene Chromosome Location
Chr.X: 40626921-40647678 on Build GRCh38
Transcripts
NM_144970.2 XM_005272589.2 XM_006724527.3 XM_006724528.2 XM_017029302.1 XM_017029303.1 XM_017029304.1 XM_017029305.1

SNP Detail

SNP ID Chromosome Location
rs918543928 Chr.X: 40646987-40646987 on Build GRCh38
rs754990948 Chr.X: 40646944-40646944 on Build GRCh38
rs766870219 Chr.X: 40646898-40646898 on Build GRCh38
rs772426274 Chr.X: 40647013-40647013 on Build GRCh38
rs183513576 Chr.X: 40646961-40646961 on Build GRCh38
rs1011448233 Chr.X: 40646954-40646954 on Build GRCh38
rs899908254 Chr.X: 40647058-40647058 on Build GRCh38
rs765505908 Chr.X: 40647056-40647056 on Build GRCh38
rs889548974 Chr.X: 40646980-40646980 on Build GRCh38
rs34507774 Chr.X: 40646994-40646994 on Build GRCh38
rs896003226 Chr.X: 40646922-40646922 on Build GRCh38
rs747733567 Chr.X: 40646970-40646970 on Build GRCh38
rs746291662 Chr.X: 40646989-40646989 on Build GRCh38
rs930238118 Chr.X: 40646926-40646926 on Build GRCh38
rs748364646 Chr.X: 40646984-40646984 on Build GRCh38
rs769635699 Chr.X: 40646972-40646972 on Build GRCh38
rs768666799 Chr.X: 40647035-40647035 on Build GRCh38
rs1047805232 Chr.X: 40646942-40646942 on Build GRCh38
rs762021259 Chr.X: 40647052-40647052 on Build GRCh38
rs1057303459 Chr.X: 40646915-40646915 on Build GRCh38
rs761110465 Chr.X: 40647030-40647030 on Build GRCh38
rs751615992 Chr.X: 40646909-40646909 on Build GRCh38
rs28608911 Chr.X: 40646985-40646985 on Build GRCh38
rs776699513 Chr.X: 40647050-40647050 on Build GRCh38
rs780859899 Chr.X: 40646964-40646964 on Build GRCh38
rs776025918 Chr.X: 40647018-40647018 on Build GRCh38

View Full Product Details