Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75763307-75763562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALK1
Entrez Gene ID
2584
Gene Name
galactokinase 1
Gene Aliases
GALK, GK1, HEL-S-19
UniGene
Hs.407966
Gene Chromosome Location
Chr.17: 75757937-75765199 on Build GRCh38
Transcripts
NM_000154.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377645 c.419C>A p.S140Y Chr.17: 75763376-75763376 on Build GRCh38
5732039 c.415C>A p.L139M Chr.17: 75763380-75763380 on Build GRCh38
1386044 c.408G>A p.G136G Chr.17: 75763387-75763387 on Build GRCh38
6556671 c.443C>A p.A148D Chr.17: 75763352-75763352 on Build GRCh38
5387323 c.472C>T p.P158S Chr.17: 75763323-75763323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140148059 Chr.17: 75763353-75763353 on Build GRCh38
rs770495951 Chr.17: 75763479-75763479 on Build GRCh38
rs905165111 Chr.17: 75763347-75763347 on Build GRCh38
rs184364091 Chr.17: 75763513-75763513 on Build GRCh38
rs781024507 Chr.17: 75763514-75763514 on Build GRCh38
rs560180991 Chr.17: 75763448-75763448 on Build GRCh38
rs758422873 Chr.17: 75763436-75763436 on Build GRCh38
rs752401700 Chr.17: 75763483-75763483 on Build GRCh38
rs765744088 Chr.17: 75763348-75763348 on Build GRCh38
rs777059513 Chr.17: 75763370-75763370 on Build GRCh38
rs527422715 Chr.17: 75763489-75763489 on Build GRCh38
rs147800408 Chr.17: 75763336-75763336 on Build GRCh38
rs1051199859 Chr.17: 75763472-75763472 on Build GRCh38
rs780587160 Chr.17: 75763354-75763354 on Build GRCh38
rs761316738 Chr.17: 75763378-75763378 on Build GRCh38
rs769550271 Chr.17: 75763379-75763379 on Build GRCh38
rs1027992509 Chr.17: 75763510-75763510 on Build GRCh38
rs1003512759 Chr.17: 75763334-75763334 on Build GRCh38
rs747536956 Chr.17: 75763355-75763355 on Build GRCh38
rs766401384 Chr.17: 75763399-75763399 on Build GRCh38
rs968025480 Chr.17: 75763477-75763477 on Build GRCh38
rs374886943 Chr.17: 75763450-75763450 on Build GRCh38
rs751497736 Chr.17: 75763403-75763403 on Build GRCh38
rs201110055 Chr.17: 75763396-75763396 on Build GRCh38
rs759530195 Chr.17: 75763480-75763480 on Build GRCh38
rs563197353 Chr.17: 75763519-75763519 on Build GRCh38
rs769303513 Chr.17: 75763470-75763470 on Build GRCh38
rs988018523 Chr.17: 75763368-75763368 on Build GRCh38
rs749400702 Chr.17: 75763345-75763345 on Build GRCh38
rs767156604 Chr.17: 75763413-75763413 on Build GRCh38
rs757414831 Chr.17: 75763431-75763431 on Build GRCh38
rs912339678 Chr.17: 75763528-75763528 on Build GRCh38
rs915070690 Chr.17: 75763385-75763385 on Build GRCh38
rs767556614 Chr.17: 75763481-75763481 on Build GRCh38
rs759546533 Chr.17: 75763410-75763410 on Build GRCh38
rs779234943 Chr.17: 75763432-75763432 on Build GRCh38
rs753104550 Chr.17: 75763349-75763349 on Build GRCh38
rs7209235 Chr.17: 75763471-75763471 on Build GRCh38
rs150323432 Chr.17: 75763363-75763363 on Build GRCh38
rs1036587004 Chr.17: 75763492-75763492 on Build GRCh38
rs767329054 Chr.17: 75763384-75763385 on Build GRCh38
rs762721387 Chr.17: 75763475-75763475 on Build GRCh38
rs138057626 Chr.17: 75763387-75763387 on Build GRCh38
rs750478379 Chr.17: 75763433-75763433 on Build GRCh38
rs752592319 Chr.17: 75763424-75763424 on Build GRCh38
rs780140518 Chr.17: 75763445-75763445 on Build GRCh38
rs748615704 Chr.17: 75763459-75763459 on Build GRCh38
rs781400629 Chr.17: 75763458-75763458 on Build GRCh38

View Full Product Details