Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45926684-45926935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARGE2
Entrez Gene ID
120071
Gene Name
LARGE xylosyl- and glucuronyltransferase 2
Gene Aliases
GYLTL1B, PP5656
UniGene
Hs.86543
Gene Chromosome Location
Chr.11: 45921621-45929096 on Build GRCh38
Transcripts
NM_001300721.1 NM_001300722.1 NM_152312.4 XM_005252787.2 XM_006718140.1 XM_006718141.3 XM_011519886.1 XM_011519888.1 XM_011519889.1 XM_011519890.1 XM_011519891.1 XM_011519892.2 XM_011519893.1 XM_017017173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613816 c.456_457insC p.R155fs*68 Chr.11: 45926821-45926822 on Build GRCh38
4574371 c.354G>T p.Q118H Chr.11: 45926719-45926719 on Build GRCh38
4032924 c.1201C>A p.P401T Chr.11: 45926747-45926747 on Build GRCh38
1979861 c.1283G>A p.R428Q Chr.11: 45926829-45926829 on Build GRCh38
6873968 c.1314G>A p.L438L Chr.11: 45926860-45926860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766190878 Chr.11: 45926836-45926836 on Build GRCh38
rs780880591 Chr.11: 45926848-45926848 on Build GRCh38
rs140734305 Chr.11: 45926811-45926811 on Build GRCh38
rs775493980 Chr.11: 45926871-45926871 on Build GRCh38
rs768689762 Chr.11: 45926738-45926738 on Build GRCh38
rs779831215 Chr.11: 45926800-45926800 on Build GRCh38
rs1055384599 Chr.11: 45926702-45926702 on Build GRCh38
rs776917611 Chr.11: 45926726-45926727 on Build GRCh38
rs762259118 Chr.11: 45926825-45926825 on Build GRCh38
rs372819981 Chr.11: 45926816-45926816 on Build GRCh38
rs758146826 Chr.11: 45926799-45926799 on Build GRCh38
rs150922857 Chr.11: 45926807-45926807 on Build GRCh38
rs999531167 Chr.11: 45926766-45926766 on Build GRCh38
rs974538917 Chr.11: 45926762-45926762 on Build GRCh38
rs955658322 Chr.11: 45926794-45926794 on Build GRCh38
rs763149503 Chr.11: 45926746-45926746 on Build GRCh38
rs778688625 Chr.11: 45926709-45926709 on Build GRCh38
rs182112944 Chr.11: 45926913-45926913 on Build GRCh38
rs760913898 Chr.11: 45926764-45926764 on Build GRCh38
rs370592670 Chr.11: 45926722-45926722 on Build GRCh38
rs763493989 Chr.11: 45926906-45926906 on Build GRCh38
rs756089595 Chr.11: 45926864-45926864 on Build GRCh38
rs374046612 Chr.11: 45926899-45926899 on Build GRCh38
rs370654703 Chr.11: 45926837-45926837 on Build GRCh38
rs767674272 Chr.11: 45926761-45926761 on Build GRCh38
rs142702567 Chr.11: 45926806-45926806 on Build GRCh38
rs765790962 Chr.11: 45926827-45926827 on Build GRCh38
rs568405351 Chr.11: 45926787-45926787 on Build GRCh38
rs776426729 Chr.11: 45926744-45926744 on Build GRCh38
rs534243389 Chr.11: 45926829-45926829 on Build GRCh38
rs146272277 Chr.11: 45926756-45926756 on Build GRCh38
rs770928472 Chr.11: 45926747-45926747 on Build GRCh38
rs775688198 Chr.11: 45926818-45926818 on Build GRCh38
rs780027260 Chr.11: 45926725-45926725 on Build GRCh38
rs867397092 Chr.11: 45926726-45926726 on Build GRCh38
rs1030152727 Chr.11: 45926893-45926894 on Build GRCh38
rs765886447 Chr.11: 45926894-45926894 on Build GRCh38
rs777653934 Chr.11: 45926865-45926865 on Build GRCh38
rs772127801 Chr.11: 45926870-45926870 on Build GRCh38
rs555737474 Chr.11: 45926789-45926789 on Build GRCh38
rs746753031 Chr.11: 45926735-45926735 on Build GRCh38
rs376236828 Chr.11: 45926822-45926822 on Build GRCh38
rs3802757 Chr.11: 45926812-45926812 on Build GRCh38
rs574202333 Chr.11: 45926908-45926908 on Build GRCh38
rs771783938 Chr.11: 45926723-45926723 on Build GRCh38
rs759647029 Chr.11: 45926821-45926822 on Build GRCh38
rs749912976 Chr.11: 45926791-45926791 on Build GRCh38
rs774149433 Chr.11: 45926737-45926737 on Build GRCh38
rs747924189 Chr.11: 45926858-45926858 on Build GRCh38
rs746011556 Chr.11: 45926868-45926868 on Build GRCh38
rs375584007 Chr.11: 45926888-45926888 on Build GRCh38
rs531591286 Chr.11: 45926763-45926763 on Build GRCh38
rs147376210 Chr.11: 45926817-45926817 on Build GRCh38
rs768340577 Chr.11: 45926828-45926828 on Build GRCh38
rs768887826 Chr.11: 45926878-45926878 on Build GRCh38
rs568895884 Chr.11: 45926758-45926758 on Build GRCh38
rs576030248 Chr.11: 45926872-45926872 on Build GRCh38
rs902440657 Chr.11: 45926910-45926910 on Build GRCh38
rs765134280 Chr.11: 45926822-45926822 on Build GRCh38
rs764383861 Chr.11: 45926823-45926823 on Build GRCh38
rs148470206 Chr.11: 45926786-45926786 on Build GRCh38

View Full Product Details