Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13365161-13365392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFOD1
Entrez Gene ID
54438
Gene Name
glucose-fructose oxidoreductase domain containing 1
Gene Aliases
ADG-90, C6orf114
UniGene
Hs.484686
Gene Chromosome Location
Chr.6: 13357830-13487637 on Build GRCh38
Transcripts
NM_001242628.1 NM_001242630.1 NM_018988.3 XM_011514699.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4912358 c.746T>C p.V249A Chr.6: 13365170-13365170 on Build GRCh38
3697510 c.606C>T p.F202F Chr.6: 13365310-13365310 on Build GRCh38
4894934 c.524C>T p.S175F Chr.6: 13365392-13365392 on Build GRCh38
1642870 c.651C>T p.S217S Chr.6: 13365265-13365265 on Build GRCh38
6357986 c.629A>G p.K210R Chr.6: 13365287-13365287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113459835 Chr.6: 13365334-13365334 on Build GRCh38
rs777729814 Chr.6: 13365335-13365335 on Build GRCh38
rs376523971 Chr.6: 13365339-13365339 on Build GRCh38
rs544970773 Chr.6: 13365358-13365358 on Build GRCh38
rs376773882 Chr.6: 13365186-13365186 on Build GRCh38
rs1027308876 Chr.6: 13365214-13365214 on Build GRCh38
rs765478647 Chr.6: 13365251-13365251 on Build GRCh38
rs950373787 Chr.6: 13365226-13365226 on Build GRCh38
rs753113596 Chr.6: 13365270-13365270 on Build GRCh38
rs554597063 Chr.6: 13365190-13365190 on Build GRCh38
rs746014593 Chr.6: 13365353-13365353 on Build GRCh38
rs775043626 Chr.6: 13365373-13365373 on Build GRCh38
rs747995227 Chr.6: 13365195-13365195 on Build GRCh38
rs775707679 Chr.6: 13365361-13365361 on Build GRCh38
rs749076810 Chr.6: 13365189-13365189 on Build GRCh38
rs868423927 Chr.6: 13365350-13365350 on Build GRCh38
rs371891921 Chr.6: 13365292-13365292 on Build GRCh38
rs149994474 Chr.6: 13365187-13365187 on Build GRCh38
rs567147003 Chr.6: 13365205-13365205 on Build GRCh38
rs776888360 Chr.6: 13365215-13365215 on Build GRCh38
rs556070235 Chr.6: 13365351-13365351 on Build GRCh38
rs201485802 Chr.6: 13365370-13365370 on Build GRCh38
rs199694596 Chr.6: 13365362-13365362 on Build GRCh38
rs759827691 Chr.6: 13365238-13365238 on Build GRCh38
rs755723408 Chr.6: 13365291-13365291 on Build GRCh38
rs965857859 Chr.6: 13365322-13365322 on Build GRCh38
rs747987858 Chr.6: 13365328-13365328 on Build GRCh38
rs760800643 Chr.6: 13365203-13365203 on Build GRCh38
rs999514859 Chr.6: 13365278-13365278 on Build GRCh38
rs773217142 Chr.6: 13365196-13365196 on Build GRCh38
rs763314839 Chr.6: 13365271-13365271 on Build GRCh38
rs764758991 Chr.6: 13365279-13365279 on Build GRCh38
rs147742016 Chr.6: 13365283-13365283 on Build GRCh38
rs142660025 Chr.6: 13365298-13365298 on Build GRCh38
rs778504183 Chr.6: 13365317-13365317 on Build GRCh38
rs118178478 Chr.6: 13365352-13365352 on Build GRCh38
rs753380403 Chr.6: 13365297-13365297 on Build GRCh38
rs368584392 Chr.6: 13365193-13365193 on Build GRCh38

View Full Product Details