Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176189294-176189559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD1
Entrez Gene ID
3231
Gene Name
homeobox D1
Gene Aliases
HOX4, HOX4G, Hox-4.7
UniGene
Hs.83465
Gene Chromosome Location
Chr.2: 176188579-176190907 on Build GRCh38
Transcripts
NM_024501.2 XM_005246507.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
717630 c.648G>C p.K216N Chr.2: 176189449-176189449 on Build GRCh38
6213994 c.332A>C p.D111A Chr.2: 176189316-176189316 on Build GRCh38
6431042 c.529G>A p.A177T Chr.2: 176189330-176189330 on Build GRCh38
5736268 c.376A>G p.T126A Chr.2: 176189360-176189360 on Build GRCh38
6398259 c.493G>A p.A165T Chr.2: 176189294-176189294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966665939 Chr.2: 176189385-176189385 on Build GRCh38
rs764503036 Chr.2: 176189401-176189401 on Build GRCh38
rs1056098610 Chr.2: 176189395-176189395 on Build GRCh38
rs758576720 Chr.2: 176189352-176189352 on Build GRCh38
rs764592971 Chr.2: 176189347-176189347 on Build GRCh38
rs942257367 Chr.2: 176189386-176189387 on Build GRCh38
rs759239001 Chr.2: 176189487-176189487 on Build GRCh38
rs774976157 Chr.2: 176189419-176189420 on Build GRCh38
rs773984075 Chr.2: 176189471-176189471 on Build GRCh38
rs762937700 Chr.2: 176189510-176189510 on Build GRCh38
rs762329948 Chr.2: 176189497-176189497 on Build GRCh38
rs896234785 Chr.2: 176189426-176189426 on Build GRCh38
rs919230567 Chr.2: 176189390-176189390 on Build GRCh38
rs781181014 Chr.2: 176189441-176189441 on Build GRCh38
rs779941495 Chr.2: 176189364-176189364 on Build GRCh38
rs13390503 Chr.2: 176189326-176189326 on Build GRCh38
rs1030370590 Chr.2: 176189482-176189482 on Build GRCh38
rs750417702 Chr.2: 176189346-176189347 on Build GRCh38
rs967924598 Chr.2: 176189461-176189461 on Build GRCh38
rs768647619 Chr.2: 176189368-176189368 on Build GRCh38
rs781270965 Chr.2: 176189372-176189372 on Build GRCh38
rs965029737 Chr.2: 176189475-176189475 on Build GRCh38
rs765318396 Chr.2: 176189412-176189412 on Build GRCh38
rs527392319 Chr.2: 176189386-176189386 on Build GRCh38
rs1056679616 Chr.2: 176189504-176189504 on Build GRCh38
rs748080098 Chr.2: 176189445-176189445 on Build GRCh38
rs754804825 Chr.2: 176189438-176189438 on Build GRCh38
rs756556456 Chr.2: 176189342-176189342 on Build GRCh38
rs746068296 Chr.2: 176189351-176189351 on Build GRCh38
rs758584269 Chr.2: 176189429-176189429 on Build GRCh38
rs757348869 Chr.2: 176189350-176189350 on Build GRCh38
rs778437925 Chr.2: 176189540-176189540 on Build GRCh38
rs746810219 Chr.2: 176189367-176189367 on Build GRCh38
rs780426544 Chr.2: 176189430-176189430 on Build GRCh38
rs753083153 Chr.2: 176189335-176189335 on Build GRCh38
rs767374189 Chr.2: 176189517-176189517 on Build GRCh38
rs912164562 Chr.2: 176189384-176189384 on Build GRCh38
rs766623397 Chr.2: 176189512-176189512 on Build GRCh38
rs769317363 Chr.2: 176189375-176189375 on Build GRCh38
rs983557681 Chr.2: 176189505-176189505 on Build GRCh38
rs770498706 Chr.2: 176189466-176189466 on Build GRCh38
rs147120298 Chr.2: 176189387-176189387 on Build GRCh38
rs1056045777 Chr.2: 176189538-176189538 on Build GRCh38
rs138188388 Chr.2: 176189509-176189509 on Build GRCh38
rs748753500 Chr.2: 176189463-176189463 on Build GRCh38
rs536497105 Chr.2: 176189533-176189533 on Build GRCh38
rs756013347 Chr.2: 176189527-176189527 on Build GRCh38
rs770503358 Chr.2: 176189389-176189389 on Build GRCh38
rs986671511 Chr.2: 176189361-176189361 on Build GRCh38
rs751808553 Chr.2: 176189515-176189515 on Build GRCh38
rs768981359 Chr.2: 176189488-176189488 on Build GRCh38
rs754413010 Chr.2: 176189349-176189349 on Build GRCh38
rs909344039 Chr.2: 176189507-176189507 on Build GRCh38
rs752448559 Chr.2: 176189518-176189518 on Build GRCh38
rs150112597 Chr.2: 176189453-176189453 on Build GRCh38
rs904823859 Chr.2: 176189388-176189388 on Build GRCh38
rs964753963 Chr.2: 176189318-176189318 on Build GRCh38
rs375477049 Chr.2: 176189392-176189392 on Build GRCh38
rs777570887 Chr.2: 176189528-176189528 on Build GRCh38
rs758361361 Chr.2: 176189447-176189450 on Build GRCh38
rs548572236 Chr.2: 176189539-176189539 on Build GRCh38
rs551896412 Chr.2: 176189404-176189404 on Build GRCh38
rs751896324 Chr.2: 176189433-176189433 on Build GRCh38
rs923351699 Chr.2: 176189346-176189346 on Build GRCh38
rs761027678 Chr.2: 176189393-176189393 on Build GRCh38
rs762553801 Chr.2: 176189329-176189329 on Build GRCh38
rs370143100 Chr.2: 176189496-176189496 on Build GRCh38
rs976078763 Chr.2: 176189476-176189476 on Build GRCh38
rs748093143 Chr.2: 176189373-176189373 on Build GRCh38
rs750487671 Chr.2: 176189416-176189416 on Build GRCh38
rs550724624 Chr.2: 176189508-176189508 on Build GRCh38
rs1001160152 Chr.2: 176189462-176189462 on Build GRCh38
rs372456292 Chr.2: 176189330-176189330 on Build GRCh38
rs141997863 Chr.2: 176189411-176189411 on Build GRCh38

View Full Product Details