Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18978502-18978769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83G
Entrez Gene ID
644815
Gene Name
family with sequence similarity 83 member G
Gene Aliases
PAWS1
UniGene
Hs.710727
Gene Chromosome Location
Chr.17: 18968789-19006144 on Build GRCh38
Transcripts
NM_001039999.2 XM_017024953.1 NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Gene Symbol
SLC5A10
Entrez Gene ID
125206
Gene Name
solute carrier family 5 member 10
Gene Aliases
SGLT-5, SGLT5
UniGene
Hs.462418
Gene Chromosome Location
Chr.17: 18950676-19020691 on Build GRCh38
Transcripts
NM_001039999.2 XM_017024953.1 NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
336500 c.1124G>T p.G375V Chr.17: 18978542-18978542 on Build GRCh38
6660244 c.1075G>A p.E359K Chr.17: 18978591-18978591 on Build GRCh38
4151496 c.926G>T p.S309I Chr.17: 18978740-18978740 on Build GRCh38
7367531 c.1133G>A p.G378D Chr.17: 18978533-18978533 on Build GRCh38
7176157 c.936C>A p.L312L Chr.17: 18978730-18978730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978351846 Chr.17: 18978607-18978607 on Build GRCh38
rs762745473 Chr.17: 18978521-18978521 on Build GRCh38
rs756740100 Chr.17: 18978585-18978585 on Build GRCh38
rs563466170 Chr.17: 18978594-18978594 on Build GRCh38
rs932870371 Chr.17: 18978643-18978643 on Build GRCh38
rs772149780 Chr.17: 18978716-18978716 on Build GRCh38
rs759934737 Chr.17: 18978650-18978650 on Build GRCh38
rs773359331 Chr.17: 18978717-18978717 on Build GRCh38
rs766632884 Chr.17: 18978620-18978620 on Build GRCh38
rs375487977 Chr.17: 18978648-18978648 on Build GRCh38
rs779652084 Chr.17: 18978596-18978596 on Build GRCh38
rs753263341 Chr.17: 18978581-18978581 on Build GRCh38
rs770977614 Chr.17: 18978734-18978734 on Build GRCh38
rs201626154 Chr.17: 18978657-18978657 on Build GRCh38
rs371196107 Chr.17: 18978605-18978605 on Build GRCh38
rs755377653 Chr.17: 18978563-18978563 on Build GRCh38
rs772239714 Chr.17: 18978612-18978612 on Build GRCh38
rs376786995 Chr.17: 18978736-18978736 on Build GRCh38
rs761010779 Chr.17: 18978541-18978541 on Build GRCh38
rs768473258 Chr.17: 18978699-18978699 on Build GRCh38
rs755655474 Chr.17: 18978690-18978690 on Build GRCh38
rs561481480 Chr.17: 18978693-18978693 on Build GRCh38
rs559388888 Chr.17: 18978655-18978655 on Build GRCh38
rs367658769 Chr.17: 18978658-18978658 on Build GRCh38
rs761977884 Chr.17: 18978527-18978527 on Build GRCh38
rs779473423 Chr.17: 18978577-18978577 on Build GRCh38
rs201631502 Chr.17: 18978742-18978742 on Build GRCh38
rs766603981 Chr.17: 18978544-18978544 on Build GRCh38
rs139570154 Chr.17: 18978598-18978598 on Build GRCh38
rs371247127 Chr.17: 18978634-18978634 on Build GRCh38
rs200817890 Chr.17: 18978533-18978533 on Build GRCh38
rs929912150 Chr.17: 18978748-18978748 on Build GRCh38
rs1026758058 Chr.17: 18978525-18978525 on Build GRCh38
rs759719309 Chr.17: 18978741-18978741 on Build GRCh38
rs748949766 Chr.17: 18978697-18978697 on Build GRCh38
rs776041051 Chr.17: 18978656-18978656 on Build GRCh38
rs959408707 Chr.17: 18978564-18978564 on Build GRCh38
rs760752072 Chr.17: 18978619-18978619 on Build GRCh38
rs780718186 Chr.17: 18978591-18978591 on Build GRCh38
rs749944687 Chr.17: 18978668-18978668 on Build GRCh38
rs1041180600 Chr.17: 18978624-18978624 on Build GRCh38
rs748625657 Chr.17: 18978703-18978703 on Build GRCh38
rs745480019 Chr.17: 18978592-18978592 on Build GRCh38
rs768651101 Chr.17: 18978526-18978526 on Build GRCh38
rs372386054 Chr.17: 18978733-18978733 on Build GRCh38
rs748051047 Chr.17: 18978708-18978708 on Build GRCh38
rs375449100 Chr.17: 18978610-18978610 on Build GRCh38
rs199992271 Chr.17: 18978656-18978656 on Build GRCh38
rs376887547 Chr.17: 18978618-18978618 on Build GRCh38
rs199697114 Chr.17: 18978597-18978597 on Build GRCh38

View Full Product Details