Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87060060-87060333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLUD1
Entrez Gene ID
2746
Gene Name
glutamate dehydrogenase 1
Gene Aliases
GDH, GDH1, GLUD
UniGene
Hs.500409
Gene Chromosome Location
Chr.10: 87050202-87095047 on Build GRCh38
Transcripts
NM_001318900.1 NM_001318901.1 NM_001318902.1 NM_001318904.1 NM_001318905.1 NM_001318906.1 NM_005271.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6871602 c.1277C>T p.P426L Chr.10: 87060162-87060162 on Build GRCh38
6066743 c.1240G>C p.D414H Chr.10: 87060199-87060199 on Build GRCh38
3441252 c.1200C>T p.I400I Chr.10: 87060239-87060239 on Build GRCh38
3967195 c.1247T>G p.I416S Chr.10: 87060192-87060192 on Build GRCh38
7147935 c.1207G>A p.E403K Chr.10: 87060232-87060232 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1002559788 Chr.10: 87060117-87060117 on Build GRCh38
rs781337235 Chr.10: 87060256-87060256 on Build GRCh38
rs368113183 Chr.10: 87060273-87060273 on Build GRCh38
rs1011853247 Chr.10: 87060302-87060302 on Build GRCh38
rs367584341 Chr.10: 87060120-87060120 on Build GRCh38
rs138169430 Chr.10: 87060197-87060197 on Build GRCh38
rs915231206 Chr.10: 87060174-87060174 on Build GRCh38
rs773192562 Chr.10: 87060129-87060129 on Build GRCh38
rs780203435 Chr.10: 87060248-87060248 on Build GRCh38
rs769624528 Chr.10: 87060264-87060264 on Build GRCh38
rs753448342 Chr.10: 87060247-87060248 on Build GRCh38
rs371850058 Chr.10: 87060262-87060262 on Build GRCh38
rs754256930 Chr.10: 87060176-87060176 on Build GRCh38
rs549333564 Chr.10: 87060266-87060266 on Build GRCh38
rs765821684 Chr.10: 87060196-87060196 on Build GRCh38
rs201421730 Chr.10: 87060250-87060250 on Build GRCh38
rs200782393 Chr.10: 87060184-87060184 on Build GRCh38
rs758374060 Chr.10: 87060215-87060215 on Build GRCh38
rs759252785 Chr.10: 87060166-87060166 on Build GRCh38
rs1038982710 Chr.10: 87060227-87060227 on Build GRCh38
rs759041919 Chr.10: 87060282-87060282 on Build GRCh38
rs764304337 Chr.10: 87060175-87060175 on Build GRCh38
rs1022918034 Chr.10: 87060136-87060136 on Build GRCh38
rs767190881 Chr.10: 87060283-87060283 on Build GRCh38
rs765953369 Chr.10: 87060145-87060145 on Build GRCh38
rs1036270884 Chr.10: 87060090-87060090 on Build GRCh38
rs773833824 Chr.10: 87060161-87060161 on Build GRCh38
rs942694203 Chr.10: 87060213-87060213 on Build GRCh38
rs762684232 Chr.10: 87060272-87060281 on Build GRCh38
rs769718066 Chr.10: 87060125-87060125 on Build GRCh38
rs1011326507 Chr.10: 87060133-87060133 on Build GRCh38
rs776499948 Chr.10: 87060112-87060112 on Build GRCh38
rs202076447 Chr.10: 87060276-87060276 on Build GRCh38
rs191637320 Chr.10: 87060143-87060143 on Build GRCh38
rs182913619 Chr.10: 87060272-87060272 on Build GRCh38
rs757758478 Chr.10: 87060188-87060188 on Build GRCh38

View Full Product Details