Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 131052442-131052675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMC3
Entrez Gene ID
10319
Gene Name
laminin subunit gamma 3
Gene Aliases
OCCM
UniGene
Hs.201805
Gene Chromosome Location
Chr.9: 131009117-131093059 on Build GRCh38
Transcripts
NM_006059.3 XM_006716921.2 XM_011518121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6699441 c.1791C>T p.A597A Chr.9: 131052651-131052651 on Build GRCh38
3982754 c.1675A>G p.I559V Chr.9: 131052535-131052535 on Build GRCh38
3329411 c.1662T>C p.Y554Y Chr.9: 131052522-131052522 on Build GRCh38
334915 c.1782C>A p.P594P Chr.9: 131052642-131052642 on Build GRCh38
7417630 c.1717G>A p.V573I Chr.9: 131052577-131052577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762811443 Chr.9: 131052524-131052524 on Build GRCh38
rs756246325 Chr.9: 131052544-131052544 on Build GRCh38
rs774054516 Chr.9: 131052562-131052562 on Build GRCh38
rs774146147 Chr.9: 131052530-131052530 on Build GRCh38
rs116422150 Chr.9: 131052652-131052652 on Build GRCh38
rs767746796 Chr.9: 131052534-131052534 on Build GRCh38
rs760910007 Chr.9: 131052592-131052592 on Build GRCh38
rs761972927 Chr.9: 131052532-131052532 on Build GRCh38
rs150171154 Chr.9: 131052548-131052548 on Build GRCh38
rs771902055 Chr.9: 131052651-131052651 on Build GRCh38
rs202042709 Chr.9: 131052601-131052601 on Build GRCh38
rs749724991 Chr.9: 131052478-131052478 on Build GRCh38
rs369683961 Chr.9: 131052558-131052558 on Build GRCh38
rs776047232 Chr.9: 131052504-131052504 on Build GRCh38
rs373106726 Chr.9: 131052605-131052605 on Build GRCh38
rs10901333 Chr.9: 131052491-131052491 on Build GRCh38
rs138637827 Chr.9: 131052556-131052556 on Build GRCh38
rs148377508 Chr.9: 131052535-131052535 on Build GRCh38
rs868403677 Chr.9: 131052508-131052508 on Build GRCh38
rs775904966 Chr.9: 131052552-131052553 on Build GRCh38
rs199764333 Chr.9: 131052547-131052547 on Build GRCh38
rs753933045 Chr.9: 131052606-131052606 on Build GRCh38
rs773561591 Chr.9: 131052587-131052587 on Build GRCh38
rs772238823 Chr.9: 131052583-131052583 on Build GRCh38
rs780681856 Chr.9: 131052474-131052474 on Build GRCh38
rs770231356 Chr.9: 131052472-131052474 on Build GRCh38
rs779180808 Chr.9: 131052553-131052553 on Build GRCh38
rs760164373 Chr.9: 131052607-131052607 on Build GRCh38
rs755480548 Chr.9: 131052550-131052550 on Build GRCh38
rs755715827 Chr.9: 131052624-131052624 on Build GRCh38
rs769783100 Chr.9: 131052512-131052512 on Build GRCh38
rs756725535 Chr.9: 131052473-131052473 on Build GRCh38
rs763891300 Chr.9: 131052525-131052525 on Build GRCh38
rs749663325 Chr.9: 131052617-131052617 on Build GRCh38
rs140815210 Chr.9: 131052516-131052516 on Build GRCh38
rs755386300 Chr.9: 131052482-131052482 on Build GRCh38
rs779570435 Chr.9: 131052626-131052626 on Build GRCh38
rs761645362 Chr.9: 131052575-131052575 on Build GRCh38
rs758786678 Chr.9: 131052614-131052614 on Build GRCh38
rs200790891 Chr.9: 131052641-131052641 on Build GRCh38
rs201297038 Chr.9: 131052612-131052612 on Build GRCh38
rs142796007 Chr.9: 131052559-131052559 on Build GRCh38
rs868137647 Chr.9: 131052527-131052527 on Build GRCh38
rs568225117 Chr.9: 131052617-131052617 on Build GRCh38
rs746666733 Chr.9: 131052492-131052492 on Build GRCh38
rs770581587 Chr.9: 131052496-131052496 on Build GRCh38
rs749246721 Chr.9: 131052561-131052561 on Build GRCh38
rs746756940 Chr.9: 131052511-131052511 on Build GRCh38
rs535615357 Chr.9: 131052630-131052630 on Build GRCh38
rs768522714 Chr.9: 131052636-131052636 on Build GRCh38
rs1030229080 Chr.9: 131052580-131052580 on Build GRCh38
rs369695895 Chr.9: 131052499-131052499 on Build GRCh38
rs1037419878 Chr.9: 131052501-131052501 on Build GRCh38
rs199701268 Chr.9: 131052542-131052542 on Build GRCh38
rs570498707 Chr.9: 131052554-131052554 on Build GRCh38
rs765754622 Chr.9: 131052608-131052608 on Build GRCh38
rs374767603 Chr.9: 131052522-131052522 on Build GRCh38
rs778895040 Chr.9: 131052640-131052640 on Build GRCh38

View Full Product Details