Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180792806-180793055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT1
Entrez Gene ID
4245
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
Gene Aliases
GLCNAC-TI, GLCT1, GLYT1, GNT-1, GNT-I, GnTI, MGAT
UniGene
Hs.519818
Gene Chromosome Location
Chr.5: 180790541-180815652 on Build GRCh38
Transcripts
NM_001114617.1 NM_001114618.1 NM_001114619.1 NM_001114620.1 NM_002406.3 XM_005265915.1 XM_005265916.1 XM_006714866.1 XM_011534559.1 XM_011534560.1 XM_011534561.1 XM_011534562.1 XM_011534563.2 XM_017009486.1 XM_017009487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1436719 c.142G>A p.A48T Chr.5: 180792830-180792830 on Build GRCh38
6369748 c.58T>C p.W20R Chr.5: 180792914-180792914 on Build GRCh38
3854417 c.72G>C p.L24L Chr.5: 180792900-180792900 on Build GRCh38
3661763 c.37G>T p.G13C Chr.5: 180792935-180792935 on Build GRCh38
6038894 c.135C>T p.G45G Chr.5: 180792837-180792837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746372187 Chr.5: 180792993-180792993 on Build GRCh38
rs112471124 Chr.5: 180793031-180793031 on Build GRCh38
rs115600707 Chr.5: 180792855-180792855 on Build GRCh38
rs112929344 Chr.5: 180792983-180792983 on Build GRCh38
rs61743256 Chr.5: 180792838-180792838 on Build GRCh38
rs765828403 Chr.5: 180792977-180792977 on Build GRCh38
rs767196669 Chr.5: 180792984-180792985 on Build GRCh38
rs550919801 Chr.5: 180792915-180792915 on Build GRCh38
rs761663462 Chr.5: 180793010-180793010 on Build GRCh38
rs368997432 Chr.5: 180792849-180792849 on Build GRCh38
rs745346635 Chr.5: 180792831-180792831 on Build GRCh38
rs111535929 Chr.5: 180792982-180792982 on Build GRCh38
rs924374684 Chr.5: 180793001-180793001 on Build GRCh38
rs34344788 Chr.5: 180792842-180792842 on Build GRCh38
rs906622840 Chr.5: 180792876-180792876 on Build GRCh38
rs944353716 Chr.5: 180792920-180792920 on Build GRCh38
rs776454807 Chr.5: 180792933-180792933 on Build GRCh38
rs755731290 Chr.5: 180793022-180793022 on Build GRCh38
rs1034885428 Chr.5: 180793032-180793032 on Build GRCh38
rs773653296 Chr.5: 180792837-180792837 on Build GRCh38
rs746023209 Chr.5: 180792897-180792897 on Build GRCh38
rs72549456 Chr.5: 180792985-180792985 on Build GRCh38
rs900220461 Chr.5: 180792976-180792977 on Build GRCh38
rs778419570 Chr.5: 180792830-180792830 on Build GRCh38
rs762882398 Chr.5: 180792966-180792966 on Build GRCh38
rs758113077 Chr.5: 180792833-180792833 on Build GRCh38
rs757834418 Chr.5: 180792874-180792874 on Build GRCh38
rs749878991 Chr.5: 180792992-180793005 on Build GRCh38
rs368619912 Chr.5: 180793005-180793005 on Build GRCh38
rs779351211 Chr.5: 180792990-180792990 on Build GRCh38
rs758954674 Chr.5: 180792887-180792889 on Build GRCh38
rs573360169 Chr.5: 180792836-180792836 on Build GRCh38
rs3733751 Chr.5: 180792984-180792984 on Build GRCh38
rs994998138 Chr.5: 180792884-180792884 on Build GRCh38
rs778312623 Chr.5: 180792881-180792881 on Build GRCh38
rs768885940 Chr.5: 180792921-180792921 on Build GRCh38
rs757687045 Chr.5: 180792988-180792988 on Build GRCh38
rs544680574 Chr.5: 180792980-180792980 on Build GRCh38
rs374617470 Chr.5: 180792995-180792995 on Build GRCh38
rs72549457 Chr.5: 180792882-180792882 on Build GRCh38
rs753583224 Chr.5: 180792856-180792856 on Build GRCh38
rs373212509 Chr.5: 180792924-180792924 on Build GRCh38
rs765217561 Chr.5: 180792942-180792942 on Build GRCh38
rs201892739 Chr.5: 180792827-180792827 on Build GRCh38
rs771614172 Chr.5: 180792888-180792888 on Build GRCh38
rs146019221 Chr.5: 180792928-180792928 on Build GRCh38
rs757132216 Chr.5: 180792829-180792829 on Build GRCh38
rs751178115 Chr.5: 180792877-180792877 on Build GRCh38
rs779546331 Chr.5: 180792894-180792894 on Build GRCh38
rs757222204 Chr.5: 180792857-180792857 on Build GRCh38
rs769583419 Chr.5: 180793015-180793015 on Build GRCh38
rs780030510 Chr.5: 180793000-180793000 on Build GRCh38
rs913185557 Chr.5: 180792861-180792861 on Build GRCh38
rs765131745 Chr.5: 180792862-180792862 on Build GRCh38
rs898022374 Chr.5: 180792883-180792883 on Build GRCh38
rs761577610 Chr.5: 180792936-180792936 on Build GRCh38
rs376603224 Chr.5: 180792879-180792879 on Build GRCh38
rs748605521 Chr.5: 180792835-180792835 on Build GRCh38
rs749985633 Chr.5: 180792873-180792873 on Build GRCh38
rs199590721 Chr.5: 180792845-180792845 on Build GRCh38
rs776822064 Chr.5: 180793006-180793006 on Build GRCh38
rs1803652 Chr.5: 180793003-180793003 on Build GRCh38
rs750356145 Chr.5: 180792959-180792959 on Build GRCh38
rs767833331 Chr.5: 180792848-180792848 on Build GRCh38
rs775578616 Chr.5: 180792908-180792908 on Build GRCh38
rs556651842 Chr.5: 180792834-180792834 on Build GRCh38
rs1007188892 Chr.5: 180792960-180792960 on Build GRCh38

View Full Product Details