Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12663244-12663513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5365735 c.819T>G p.C273W Chr.19: 12663407-12663407 on Build GRCh38
5972713 c.764G>C p.G255A Chr.19: 12663462-12663462 on Build GRCh38
1390612 c.855C>T p.P285P Chr.19: 12663371-12663371 on Build GRCh38
6003816 c.909+4A>G p.? Chr.19: 12663313-12663313 on Build GRCh38
6286867 c.902C>A p.T301N Chr.19: 12663324-12663324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954167607 Chr.19: 12663285-12663285 on Build GRCh38
rs377049583 Chr.19: 12663378-12663378 on Build GRCh38
rs764312812 Chr.19: 12663386-12663386 on Build GRCh38
rs762330547 Chr.19: 12663485-12663485 on Build GRCh38
rs532183715 Chr.19: 12663271-12663271 on Build GRCh38
rs373864097 Chr.19: 12663476-12663476 on Build GRCh38
rs753038091 Chr.19: 12663309-12663309 on Build GRCh38
rs752041314 Chr.19: 12663455-12663476 on Build GRCh38
rs776303664 Chr.19: 12663286-12663286 on Build GRCh38
rs766802139 Chr.19: 12663292-12663292 on Build GRCh38
rs763820854 Chr.19: 12663282-12663282 on Build GRCh38
rs781127503 Chr.19: 12663462-12663462 on Build GRCh38
rs757302890 Chr.19: 12663325-12663325 on Build GRCh38
rs758433220 Chr.19: 12663412-12663412 on Build GRCh38
rs370129661 Chr.19: 12663288-12663288 on Build GRCh38
rs768458196 Chr.19: 12663350-12663350 on Build GRCh38
rs149484019 Chr.19: 12663437-12663437 on Build GRCh38
rs775475187 Chr.19: 12663344-12663344 on Build GRCh38
rs772572174 Chr.19: 12663486-12663486 on Build GRCh38
rs775917208 Chr.19: 12663490-12663490 on Build GRCh38
rs746756800 Chr.19: 12663347-12663347 on Build GRCh38
rs886054232 Chr.19: 12663383-12663383 on Build GRCh38
rs781465198 Chr.19: 12663293-12663293 on Build GRCh38
rs146909343 Chr.19: 12663362-12663362 on Build GRCh38
rs1054486 Chr.19: 12663394-12663394 on Build GRCh38
rs944317989 Chr.19: 12663441-12663441 on Build GRCh38
rs976940825 Chr.19: 12663447-12663447 on Build GRCh38
rs768516728 Chr.19: 12663450-12663450 on Build GRCh38
rs779091097 Chr.19: 12663396-12663396 on Build GRCh38
rs749372996 Chr.19: 12663320-12663320 on Build GRCh38
rs927024699 Chr.19: 12663364-12663364 on Build GRCh38
rs199690827 Chr.19: 12663327-12663327 on Build GRCh38
rs973412688 Chr.19: 12663346-12663346 on Build GRCh38
rs985478505 Chr.19: 12663398-12663398 on Build GRCh38
rs771874603 Chr.19: 12663337-12663337 on Build GRCh38
rs995995538 Chr.19: 12663274-12663274 on Build GRCh38
rs151037468 Chr.19: 12663331-12663331 on Build GRCh38
rs757417272 Chr.19: 12663395-12663395 on Build GRCh38
rs978130289 Chr.19: 12663379-12663379 on Build GRCh38
rs771963115 Chr.19: 12663266-12663266 on Build GRCh38
rs750447748 Chr.19: 12663404-12663404 on Build GRCh38
rs777928463 Chr.19: 12663312-12663312 on Build GRCh38
rs961621873 Chr.19: 12663321-12663321 on Build GRCh38
rs139218045 Chr.19: 12663371-12663371 on Build GRCh38
rs1017356457 Chr.19: 12663302-12663302 on Build GRCh38
rs45576136 Chr.19: 12663382-12663382 on Build GRCh38
rs141077530 Chr.19: 12663352-12663352 on Build GRCh38
rs201385272 Chr.19: 12663310-12663310 on Build GRCh38
rs557695977 Chr.19: 12663297-12663297 on Build GRCh38
rs543188883 Chr.19: 12663281-12663281 on Build GRCh38
rs772796727 Chr.19: 12663482-12663482 on Build GRCh38
rs761245440 Chr.19: 12663361-12663361 on Build GRCh38
rs746808159 Chr.19: 12663438-12663438 on Build GRCh38
rs757435344 Chr.19: 12663287-12663287 on Build GRCh38
rs747871268 Chr.19: 12663473-12663473 on Build GRCh38
rs755434737 Chr.19: 12663303-12663303 on Build GRCh38
rs767907971 Chr.19: 12663308-12663308 on Build GRCh38
rs772562587 Chr.19: 12663370-12663370 on Build GRCh38

View Full Product Details