Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39785617-39785876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEUTX
Entrez Gene ID
342900
Gene Name
leucine twenty homeobox
Gene Aliases
-
UniGene
Hs.451638
Gene Chromosome Location
Chr.19: 39776594-39786135 on Build GRCh38
Transcripts
NM_001143832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771068 c.130C>T p.R44W Chr.19: 39785758-39785758 on Build GRCh38
5557425 c.195A>G p.I65M Chr.19: 39785823-39785823 on Build GRCh38
5581538 c.224C>T p.P75L Chr.19: 39785852-39785852 on Build GRCh38
4140633 c.174G>A p.K58K Chr.19: 39785802-39785802 on Build GRCh38
228221 c.98G>A p.W33* Chr.19: 39785726-39785726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748018606 Chr.19: 39785811-39785811 on Build GRCh38
rs576568621 Chr.19: 39785660-39785660 on Build GRCh38
rs954443066 Chr.19: 39785700-39785700 on Build GRCh38
rs776270599 Chr.19: 39785804-39785804 on Build GRCh38
rs558962748 Chr.19: 39785758-39785758 on Build GRCh38
rs768484764 Chr.19: 39785792-39785792 on Build GRCh38
rs974354701 Chr.19: 39785742-39785742 on Build GRCh38
rs763039281 Chr.19: 39785674-39785674 on Build GRCh38
rs930028774 Chr.19: 39785762-39785762 on Build GRCh38
rs10410397 Chr.19: 39785683-39785683 on Build GRCh38
rs529686425 Chr.19: 39785812-39785812 on Build GRCh38
rs867912521 Chr.19: 39785704-39785704 on Build GRCh38
rs541307025 Chr.19: 39785720-39785720 on Build GRCh38
rs376692805 Chr.19: 39785740-39785740 on Build GRCh38
rs773212414 Chr.19: 39785813-39785813 on Build GRCh38
rs949468596 Chr.19: 39785836-39785836 on Build GRCh38
rs555000947 Chr.19: 39785651-39785651 on Build GRCh38
rs373629971 Chr.19: 39785840-39785840 on Build GRCh38
rs1032632220 Chr.19: 39785682-39785682 on Build GRCh38
rs921339184 Chr.19: 39785759-39785759 on Build GRCh38
rs776861037 Chr.19: 39785678-39785678 on Build GRCh38
rs985819903 Chr.19: 39785717-39785717 on Build GRCh38
rs1007829942 Chr.19: 39785849-39785849 on Build GRCh38
rs752965786 Chr.19: 39785845-39785845 on Build GRCh38
rs1030065739 Chr.19: 39785697-39785697 on Build GRCh38
rs372270167 Chr.19: 39785688-39785688 on Build GRCh38
rs759722545 Chr.19: 39785785-39785787 on Build GRCh38
rs191208768 Chr.19: 39785648-39785648 on Build GRCh38
rs766439043 Chr.19: 39785815-39785815 on Build GRCh38
rs369685173 Chr.19: 39785783-39785783 on Build GRCh38
rs751780707 Chr.19: 39785679-39785679 on Build GRCh38
rs547985951 Chr.19: 39785839-39785839 on Build GRCh38
rs577142991 Chr.19: 39785716-39785716 on Build GRCh38
rs745314487 Chr.19: 39785725-39785725 on Build GRCh38
rs979223035 Chr.19: 39785681-39785681 on Build GRCh38
rs774471747 Chr.19: 39785821-39785821 on Build GRCh38
rs762950974 Chr.19: 39785814-39785814 on Build GRCh38
rs1023457058 Chr.19: 39785644-39785644 on Build GRCh38
rs368857254 Chr.19: 39785678-39785678 on Build GRCh38
rs920170515 Chr.19: 39785760-39785760 on Build GRCh38
rs565258087 Chr.19: 39785706-39785706 on Build GRCh38

View Full Product Details