Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184112687-184112912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP6
Entrez Gene ID
133121
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Gene Aliases
NPP6
UniGene
Hs.297814
Gene Chromosome Location
Chr.4: 184088706-184217961 on Build GRCh38
Transcripts
NM_153343.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
381550 c.977G>T p.G326V Chr.4: 184112688-184112688 on Build GRCh38
3130189 c.970G>A p.D324N Chr.4: 184112695-184112695 on Build GRCh38
3825593 c.933G>T p.K311N Chr.4: 184112732-184112732 on Build GRCh38
4512708 c.912C>T p.I304I Chr.4: 184112753-184112753 on Build GRCh38
5654728 c.943T>G p.F315V Chr.4: 184112722-184112722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762758635 Chr.4: 184112788-184112788 on Build GRCh38
rs777712020 Chr.4: 184112741-184112741 on Build GRCh38
rs763495026 Chr.4: 184112709-184112709 on Build GRCh38
rs996424080 Chr.4: 184112871-184112871 on Build GRCh38
rs141497036 Chr.4: 184112805-184112805 on Build GRCh38
rs767844061 Chr.4: 184112750-184112750 on Build GRCh38
rs200617400 Chr.4: 184112768-184112768 on Build GRCh38
rs563694602 Chr.4: 184112845-184112845 on Build GRCh38
rs140684686 Chr.4: 184112723-184112723 on Build GRCh38
rs754865458 Chr.4: 184112799-184112799 on Build GRCh38
rs775993721 Chr.4: 184112772-184112772 on Build GRCh38
rs138480459 Chr.4: 184112794-184112794 on Build GRCh38
rs200000730 Chr.4: 184112739-184112739 on Build GRCh38
rs9992680 Chr.4: 184112714-184112714 on Build GRCh38
rs1037735212 Chr.4: 184112815-184112815 on Build GRCh38
rs779216094 Chr.4: 184112757-184112757 on Build GRCh38
rs777964667 Chr.4: 184112804-184112804 on Build GRCh38
rs779304242 Chr.4: 184112837-184112837 on Build GRCh38
rs150195560 Chr.4: 184112767-184112767 on Build GRCh38
rs572185024 Chr.4: 184112735-184112735 on Build GRCh38
rs201995765 Chr.4: 184112797-184112797 on Build GRCh38
rs749588199 Chr.4: 184112744-184112744 on Build GRCh38
rs750239546 Chr.4: 184112727-184112727 on Build GRCh38
rs948603264 Chr.4: 184112762-184112762 on Build GRCh38
rs145892678 Chr.4: 184112730-184112730 on Build GRCh38
rs757290881 Chr.4: 184112822-184112822 on Build GRCh38
rs746235678 Chr.4: 184112764-184112764 on Build GRCh38
rs79633596 Chr.4: 184112862-184112862 on Build GRCh38
rs750038439 Chr.4: 184112786-184112786 on Build GRCh38
rs967079828 Chr.4: 184112732-184112732 on Build GRCh38
rs753590688 Chr.4: 184112711-184112711 on Build GRCh38
rs756277302 Chr.4: 184112737-184112737 on Build GRCh38
rs761330652 Chr.4: 184112773-184112773 on Build GRCh38
rs758148477 Chr.4: 184112728-184112728 on Build GRCh38

View Full Product Details