Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25383723-25383996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTNB
Entrez Gene ID
1838
Gene Name
dystrobrevin beta
Gene Aliases
-
UniGene
Hs.307720
Gene Chromosome Location
Chr.2: 25377220-25673647 on Build GRCh38
Transcripts
NM_001256303.1 NM_001256304.1 NM_001256308.2 NM_001320932.1 NM_001320933.1 NM_001320934.1 NM_001320935.1 NM_021907.4 NM_033147.3 NM_033148.3 NM_183360.2 NM_183361.2 XM_005264182.1 XM_011532671.1 XM_011532673.1 XM_011532674.1 XM_011532675.1 XM_011532676.1 XM_011532677.2 XM_011532680.2 XM_011532681.2 XM_017003529.1 XM_017003530.1 XM_017003532.1 XM_017003533.1 XM_017003534.1 XM_017003535.1 XM_017003537.1 XM_017003538.1 XM_017003539.1 XM_017003540.1 XM_017003541.1 XM_017003542.1 XM_017003543.1 XM_017003544.1 XM_017003545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650033 c.1761_1763delAGA p.E588delE Chr.2: 25383862-25383864 on Build GRCh38
384928 c.1826-1G>T p.? Chr.2: 25383890-25383890 on Build GRCh38
4402667 c.1851A>C p.E617D Chr.2: 25383864-25383864 on Build GRCh38
6157732 c.1741G>A p.E581K Chr.2: 25383884-25383884 on Build GRCh38
392652 c.1866_1867insT p.G623fs*>6 Chr.2: 25383848-25383849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024241806 Chr.2: 25383864-25383864 on Build GRCh38
rs753625406 Chr.2: 25383743-25383743 on Build GRCh38
rs993334169 Chr.2: 25383910-25383910 on Build GRCh38
rs914517529 Chr.2: 25383906-25383906 on Build GRCh38
rs750294877 Chr.2: 25383959-25383959 on Build GRCh38
rs961850767 Chr.2: 25383884-25383884 on Build GRCh38
rs777527361 Chr.2: 25383758-25383758 on Build GRCh38
rs763444605 Chr.2: 25383802-25383802 on Build GRCh38
rs761563947 Chr.2: 25383911-25383911 on Build GRCh38
rs748243276 Chr.2: 25383905-25383905 on Build GRCh38
rs763542328 Chr.2: 25383835-25383846 on Build GRCh38
rs747060600 Chr.2: 25383781-25383781 on Build GRCh38
rs766624173 Chr.2: 25383926-25383926 on Build GRCh38
rs755984212 Chr.2: 25383826-25383826 on Build GRCh38
rs1053349244 Chr.2: 25383872-25383872 on Build GRCh38
rs889457090 Chr.2: 25383927-25383927 on Build GRCh38
rs778816990 Chr.2: 25383852-25383852 on Build GRCh38
rs770385591 Chr.2: 25383912-25383912 on Build GRCh38
rs375553958 Chr.2: 25383925-25383925 on Build GRCh38
rs775977629 Chr.2: 25383862-25383864 on Build GRCh38
rs76887521 Chr.2: 25383766-25383766 on Build GRCh38
rs776559230 Chr.2: 25383888-25383888 on Build GRCh38
rs772213423 Chr.2: 25383775-25383775 on Build GRCh38
rs773880374 Chr.2: 25383915-25383915 on Build GRCh38
rs754702944 Chr.2: 25383822-25383822 on Build GRCh38
rs766936077 Chr.2: 25383823-25383823 on Build GRCh38
rs776811575 Chr.2: 25383785-25383785 on Build GRCh38
rs200087086 Chr.2: 25383866-25383866 on Build GRCh38
rs368074319 Chr.2: 25383792-25383792 on Build GRCh38
rs751153504 Chr.2: 25383820-25383820 on Build GRCh38
rs752314040 Chr.2: 25383825-25383825 on Build GRCh38
rs200606748 Chr.2: 25383809-25383809 on Build GRCh38
rs200318184 Chr.2: 25383907-25383907 on Build GRCh38
rs147861559 Chr.2: 25383776-25383776 on Build GRCh38
rs763773059 Chr.2: 25383749-25383749 on Build GRCh38
rs774850158 Chr.2: 25383932-25383932 on Build GRCh38
rs768594139 Chr.2: 25383783-25383783 on Build GRCh38
rs749091185 Chr.2: 25383774-25383774 on Build GRCh38
rs775338933 Chr.2: 25383768-25383769 on Build GRCh38
rs773645238 Chr.2: 25383793-25383793 on Build GRCh38
rs777688713 Chr.2: 25383838-25383838 on Build GRCh38
rs757063061 Chr.2: 25383839-25383839 on Build GRCh38
rs760208216 Chr.2: 25383832-25383833 on Build GRCh38
rs768990011 Chr.2: 25383877-25383877 on Build GRCh38
rs762404766 Chr.2: 25383787-25383787 on Build GRCh38

View Full Product Details