Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20653536-20653801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDOST
Entrez Gene ID
1650
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
Gene Aliases
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1
UniGene
Hs.523145
Gene Chromosome Location
Chr.1: 20651767-20661544 on Build GRCh38
Transcripts
NM_005216.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3482880 c.903G>A p.K301K Chr.1: 20653717-20653717 on Build GRCh38
1929122 c.959C>T p.A320V Chr.1: 20653661-20653661 on Build GRCh38
7119900 c.954G>C p.E318D Chr.1: 20653666-20653666 on Build GRCh38
6641372 c.928C>T p.P310S Chr.1: 20653692-20653692 on Build GRCh38
6503632 c.875C>T p.A292V Chr.1: 20653745-20653745 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs6692016 Chr.1: 20653738-20653738 on Build GRCh38
rs370563377 Chr.1: 20653632-20653632 on Build GRCh38
rs757476771 Chr.1: 20653747-20653747 on Build GRCh38
rs770571110 Chr.1: 20653781-20653781 on Build GRCh38
rs889845144 Chr.1: 20653594-20653594 on Build GRCh38
rs747502025 Chr.1: 20653767-20653767 on Build GRCh38
rs763212913 Chr.1: 20653640-20653642 on Build GRCh38
rs776007343 Chr.1: 20653693-20653693 on Build GRCh38
rs117925699 Chr.1: 20653676-20653676 on Build GRCh38
rs142402976 Chr.1: 20653677-20653677 on Build GRCh38
rs367807479 Chr.1: 20653700-20653700 on Build GRCh38
rs760030173 Chr.1: 20653597-20653597 on Build GRCh38
rs1010960426 Chr.1: 20653604-20653604 on Build GRCh38
rs772143791 Chr.1: 20653757-20653757 on Build GRCh38
rs200050311 Chr.1: 20653657-20653657 on Build GRCh38
rs1049781574 Chr.1: 20653563-20653563 on Build GRCh38
rs754561384 Chr.1: 20653730-20653730 on Build GRCh38
rs752691070 Chr.1: 20653732-20653732 on Build GRCh38
rs771053378 Chr.1: 20653593-20653593 on Build GRCh38
rs774348897 Chr.1: 20653595-20653595 on Build GRCh38
rs781331641 Chr.1: 20653671-20653671 on Build GRCh38
rs538267323 Chr.1: 20653664-20653664 on Build GRCh38
rs769111163 Chr.1: 20653772-20653772 on Build GRCh38
rs200857992 Chr.1: 20653674-20653674 on Build GRCh38
rs762664774 Chr.1: 20653717-20653717 on Build GRCh38
rs771122583 Chr.1: 20653679-20653679 on Build GRCh38
rs145327708 Chr.1: 20653675-20653675 on Build GRCh38
rs754936579 Chr.1: 20653670-20653670 on Build GRCh38
rs765989677 Chr.1: 20653719-20653719 on Build GRCh38
rs35610874 Chr.1: 20653593-20653593 on Build GRCh38
rs767525741 Chr.1: 20653731-20653731 on Build GRCh38
rs761048216 Chr.1: 20653643-20653643 on Build GRCh38
rs17849634 Chr.1: 20653669-20653669 on Build GRCh38
rs768049045 Chr.1: 20653600-20653600 on Build GRCh38
rs750526667 Chr.1: 20653613-20653614 on Build GRCh38
rs749074495 Chr.1: 20653739-20653739 on Build GRCh38
rs762134804 Chr.1: 20653777-20653777 on Build GRCh38
rs201962457 Chr.1: 20653774-20653774 on Build GRCh38
rs749291113 Chr.1: 20653586-20653586 on Build GRCh38
rs369105590 Chr.1: 20653754-20653754 on Build GRCh38
rs745906369 Chr.1: 20653755-20653755 on Build GRCh38
rs756060619 Chr.1: 20653733-20653733 on Build GRCh38
rs371751724 Chr.1: 20653701-20653701 on Build GRCh38
rs745995127 Chr.1: 20653688-20653688 on Build GRCh38
rs1004428863 Chr.1: 20653780-20653780 on Build GRCh38
rs774456070 Chr.1: 20653685-20653685 on Build GRCh38
rs368054727 Chr.1: 20653583-20653583 on Build GRCh38
rs144834631 Chr.1: 20653666-20653666 on Build GRCh38
rs780481039 Chr.1: 20653766-20653766 on Build GRCh38
rs770121804 Chr.1: 20653578-20653578 on Build GRCh38
rs998783210 Chr.1: 20653668-20653668 on Build GRCh38
rs772517304 Chr.1: 20653692-20653692 on Build GRCh38
rs751094890 Chr.1: 20653726-20653726 on Build GRCh38
rs764755742 Chr.1: 20653652-20653652 on Build GRCh38
rs750093594 Chr.1: 20653659-20653659 on Build GRCh38

View Full Product Details