Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48904893-48905118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35A2
Entrez Gene ID
7355
Gene Name
solute carrier family 35 member A2
Gene Aliases
CDG2M, CDGX, UDP-Gal-Tr, UGALT, UGAT, UGT, UGT1, UGT2, UGTL
UniGene
Hs.21899
Gene Chromosome Location
Chr.X: 48903180-48911958 on Build GRCh38
Transcripts
NM_001032289.2 NM_001042498.2 NM_001282647.1 NM_001282648.1 NM_001282649.1 NM_001282650.1 NM_001282651.1 NM_005660.2

SNP Detail

SNP ID Chromosome Location
rs373788556 Chr.X: 48904943-48904943 on Build GRCh38
rs781917175 Chr.X: 48905036-48905036 on Build GRCh38
rs782737391 Chr.X: 48905083-48905083 on Build GRCh38
rs781825695 Chr.X: 48905087-48905087 on Build GRCh38
rs782603817 Chr.X: 48904928-48904928 on Build GRCh38
rs781992603 Chr.X: 48904969-48904969 on Build GRCh38
rs781996492 Chr.X: 48905069-48905069 on Build GRCh38
rs782406133 Chr.X: 48905066-48905066 on Build GRCh38
rs782485193 Chr.X: 48904927-48904927 on Build GRCh38
rs782113085 Chr.X: 48905081-48905081 on Build GRCh38
rs782321864 Chr.X: 48905035-48905035 on Build GRCh38
rs587776961 Chr.X: 48904918-48904918 on Build GRCh38
rs376745488 Chr.X: 48904964-48904964 on Build GRCh38
rs35926671 Chr.X: 48904957-48904958 on Build GRCh38
rs143797243 Chr.X: 48904967-48904967 on Build GRCh38
rs587777435 Chr.X: 48904937-48904937 on Build GRCh38
rs782209341 Chr.X: 48905006-48905006 on Build GRCh38
rs140079332 Chr.X: 48904951-48904951 on Build GRCh38
rs920606773 Chr.X: 48904968-48904968 on Build GRCh38
rs782161189 Chr.X: 48905057-48905057 on Build GRCh38
rs146079657 Chr.X: 48904919-48904919 on Build GRCh38
rs782068680 Chr.X: 48905094-48905094 on Build GRCh38
rs782675945 Chr.X: 48904952-48904952 on Build GRCh38

View Full Product Details