Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7922004-7922277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAPC2
Entrez Gene ID
6618
Gene Name
small nuclear RNA activating complex polypeptide 2
Gene Aliases
PTFDELTA, SNAP45
UniGene
Hs.631860
Gene Chromosome Location
Chr.19: 7920309-7923251 on Build GRCh38
Transcripts
NM_003083.3 NR_030717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1711726 c.607G>C p.A203P Chr.19: 7922269-7922269 on Build GRCh38
7222033 c.446G>T p.G149V Chr.19: 7922108-7922108 on Build GRCh38
2729722 c.408C>T p.L136L Chr.19: 7922070-7922070 on Build GRCh38
1680585 c.385G>A p.A129T Chr.19: 7922047-7922047 on Build GRCh38
4082935 c.386C>T p.A129V Chr.19: 7922048-7922048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774284600 Chr.19: 7922250-7922250 on Build GRCh38
rs775091743 Chr.19: 7922121-7922121 on Build GRCh38
rs768823097 Chr.19: 7922192-7922192 on Build GRCh38
rs752232101 Chr.19: 7922066-7922066 on Build GRCh38
rs763886500 Chr.19: 7922127-7922127 on Build GRCh38
rs374957840 Chr.19: 7922206-7922206 on Build GRCh38
rs774770237 Chr.19: 7922105-7922105 on Build GRCh38
rs773589574 Chr.19: 7922047-7922047 on Build GRCh38
rs759488700 Chr.19: 7922251-7922251 on Build GRCh38
rs367794573 Chr.19: 7922159-7922159 on Build GRCh38
rs959213233 Chr.19: 7922188-7922188 on Build GRCh38
rs754948243 Chr.19: 7922087-7922087 on Build GRCh38
rs780676694 Chr.19: 7922225-7922225 on Build GRCh38
rs377694941 Chr.19: 7922031-7922031 on Build GRCh38
rs1026391651 Chr.19: 7922146-7922146 on Build GRCh38
rs779268421 Chr.19: 7922163-7922163 on Build GRCh38
rs771416536 Chr.19: 7922103-7922103 on Build GRCh38
rs773091798 Chr.19: 7922193-7922193 on Build GRCh38
rs934127158 Chr.19: 7922242-7922242 on Build GRCh38
rs533379070 Chr.19: 7922173-7922173 on Build GRCh38
rs536117478 Chr.19: 7922038-7922038 on Build GRCh38
rs767459345 Chr.19: 7922149-7922149 on Build GRCh38
rs374752327 Chr.19: 7922152-7922152 on Build GRCh38
rs887648608 Chr.19: 7922061-7922061 on Build GRCh38
rs200308536 Chr.19: 7922184-7922184 on Build GRCh38
rs577692069 Chr.19: 7922027-7922027 on Build GRCh38
rs768225274 Chr.19: 7922237-7922237 on Build GRCh38
rs796347480 Chr.19: 7922230-7922231 on Build GRCh38
rs201779233 Chr.19: 7922093-7922093 on Build GRCh38
rs758383443 Chr.19: 7922145-7922145 on Build GRCh38
rs763411378 Chr.19: 7922048-7922048 on Build GRCh38
rs756394094 Chr.19: 7922097-7922097 on Build GRCh38
rs370465466 Chr.19: 7922171-7922171 on Build GRCh38
rs929065703 Chr.19: 7922041-7922041 on Build GRCh38
rs757497834 Chr.19: 7922210-7922210 on Build GRCh38
rs202107483 Chr.19: 7922076-7922076 on Build GRCh38
rs746585987 Chr.19: 7922175-7922175 on Build GRCh38
rs767057828 Chr.19: 7922060-7922060 on Build GRCh38
rs778084242 Chr.19: 7922098-7922098 on Build GRCh38
rs975739511 Chr.19: 7922035-7922035 on Build GRCh38
rs770772794 Chr.19: 7922243-7922243 on Build GRCh38
rs773830865 Chr.19: 7922051-7922051 on Build GRCh38
rs754007935 Chr.19: 7922209-7922209 on Build GRCh38
rs1051311397 Chr.19: 7922111-7922111 on Build GRCh38
rs749513946 Chr.19: 7922153-7922153 on Build GRCh38
rs768417357 Chr.19: 7922182-7922182 on Build GRCh38
rs748410916 Chr.19: 7922040-7922040 on Build GRCh38
rs766391778 Chr.19: 7922199-7922199 on Build GRCh38
rs201531431 Chr.19: 7922240-7922240 on Build GRCh38
rs75251458 Chr.19: 7922125-7922125 on Build GRCh38
rs201040469 Chr.19: 7922086-7922086 on Build GRCh38
rs780326346 Chr.19: 7922090-7922090 on Build GRCh38
rs750271652 Chr.19: 7922141-7922141 on Build GRCh38
rs769999852 Chr.19: 7922042-7922042 on Build GRCh38
rs780432962 Chr.19: 7922219-7922220 on Build GRCh38
rs759033656 Chr.19: 7922056-7922056 on Build GRCh38
rs199514852 Chr.19: 7922070-7922070 on Build GRCh38
rs760813217 Chr.19: 7922202-7922202 on Build GRCh38
rs924059139 Chr.19: 7922102-7922102 on Build GRCh38
rs1013237015 Chr.19: 7922147-7922147 on Build GRCh38
rs185154761 Chr.19: 7922238-7922238 on Build GRCh38
rs761765057 Chr.19: 7922129-7922129 on Build GRCh38
rs752708385 Chr.19: 7922200-7922200 on Build GRCh38
rs758007942 Chr.19: 7922230-7922231 on Build GRCh38
rs765076101 Chr.19: 7922191-7922191 on Build GRCh38
rs151120769 Chr.19: 7922164-7922164 on Build GRCh38
rs752865998 Chr.19: 7922151-7922151 on Build GRCh38
rs761572960 Chr.19: 7922185-7922185 on Build GRCh38
rs760213636 Chr.19: 7922124-7922124 on Build GRCh38
rs145914511 Chr.19: 7922114-7922114 on Build GRCh38
rs374915726 Chr.19: 7922216-7922216 on Build GRCh38
rs753725949 Chr.19: 7922078-7922078 on Build GRCh38
rs747473913 Chr.19: 7922226-7922226 on Build GRCh38
rs746397588 Chr.19: 7922108-7922108 on Build GRCh38
rs781346651 Chr.19: 7922089-7922089 on Build GRCh38
rs774615332 Chr.19: 7922227-7922227 on Build GRCh38
rs200376385 Chr.19: 7922128-7922128 on Build GRCh38
rs771469454 Chr.19: 7922049-7922049 on Build GRCh38
rs1038649467 Chr.19: 7922139-7922139 on Build GRCh38
rs374306019 Chr.19: 7922241-7922241 on Build GRCh38
rs758817659 Chr.19: 7922219-7922219 on Build GRCh38
rs772674180 Chr.19: 7922165-7922165 on Build GRCh38
rs954391147 Chr.19: 7922211-7922211 on Build GRCh38
rs200817922 Chr.19: 7922215-7922215 on Build GRCh38
rs150246284 Chr.19: 7922217-7922217 on Build GRCh38
rs765130914 Chr.19: 7922137-7922137 on Build GRCh38
rs141029808 Chr.19: 7922194-7922194 on Build GRCh38
rs754333476 Chr.19: 7922228-7922230 on Build GRCh38
rs750817767 Chr.19: 7922083-7922083 on Build GRCh38
rs200768961 Chr.19: 7922083-7922083 on Build GRCh38
rs371309789 Chr.19: 7922101-7922101 on Build GRCh38
rs764114888 Chr.19: 7922252-7922252 on Build GRCh38

View Full Product Details